Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218782348-218782575 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP27A1
Entrez Gene ID
1593
Gene Name
cytochrome P450 family 27 subfamily A member 1
Gene Aliases
CP27, CTX, CYP27
UniGene
Hs.516700
Gene Chromosome Location
Chr.2: 218781006-218815293 on Build GRCh38
Transcripts
NM_000784.3 XM_017003488.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7168033 c.219C>T p.F73F Chr.2: 218782401-218782401 on Build GRCh38
243938 c.219C>G p.F73L Chr.2: 218782401-218782401 on Build GRCh38
4449298 c.201_203delCTT p.F70delF Chr.2: 218782383-218782385 on Build GRCh38
5679020 c.183A>T p.P61P Chr.2: 218782365-218782365 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs546155967 Chr.2: 218782547-218782547 on Build GRCh38
rs989008885 Chr.2: 218782473-218782473 on Build GRCh38
rs374941032 Chr.2: 218782437-218782437 on Build GRCh38
rs773257024 Chr.2: 218782503-218782503 on Build GRCh38
rs747003400 Chr.2: 218782396-218782396 on Build GRCh38
rs745513715 Chr.2: 218782405-218782405 on Build GRCh38
rs534844099 Chr.2: 218782402-218782402 on Build GRCh38
rs754977188 Chr.2: 218782471-218782471 on Build GRCh38
rs767761739 Chr.2: 218782451-218782451 on Build GRCh38
rs754140326 Chr.2: 218782462-218782462 on Build GRCh38
rs757282860 Chr.2: 218782482-218782482 on Build GRCh38
rs557106671 Chr.2: 218782513-218782513 on Build GRCh38
rs75897848 Chr.2: 218782425-218782425 on Build GRCh38
rs766655647 Chr.2: 218782460-218782460 on Build GRCh38
rs138189735 Chr.2: 218782397-218782397 on Build GRCh38
rs542296211 Chr.2: 218782496-218782496 on Build GRCh38
rs778416475 Chr.2: 218782494-218782495 on Build GRCh38
rs746903905 Chr.2: 218782478-218782478 on Build GRCh38
rs370498798 Chr.2: 218782423-218782423 on Build GRCh38
rs185591162 Chr.2: 218782430-218782430 on Build GRCh38
rs78706879 Chr.2: 218782524-218782524 on Build GRCh38
rs777527880 Chr.2: 218782393-218782393 on Build GRCh38
rs762433789 Chr.2: 218782446-218782446 on Build GRCh38
rs587780327 Chr.2: 218782385-218782385 on Build GRCh38
rs1003105272 Chr.2: 218782418-218782418 on Build GRCh38
rs779095207 Chr.2: 218782474-218782474 on Build GRCh38
rs1019942078 Chr.2: 218782497-218782497 on Build GRCh38
rs1011541224 Chr.2: 218782510-218782510 on Build GRCh38
rs955051821 Chr.2: 218782514-218782514 on Build GRCh38
rs774193477 Chr.2: 218782383-218782385 on Build GRCh38
rs967431301 Chr.2: 218782527-218782527 on Build GRCh38
rs867496731 Chr.2: 218782415-218782415 on Build GRCh38
rs774689564 Chr.2: 218782445-218782445 on Build GRCh38

View Full Product Details