Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 298104-298329 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AXIN1
Entrez Gene ID
8312
Gene Name
axin 1
Gene Aliases
AXIN, PPP1R49
UniGene
Hs.592082
Gene Chromosome Location
Chr.16: 287440-355226 on Build GRCh38
Transcripts
NM_003502.3 NM_181050.2 NR_134879.1 XM_011522682.2 XM_011522683.2 XM_011522684.2 XM_011522686.1 XM_017023743.1 XM_017023744.1 XM_017023745.1 XM_017023746.1 XM_017023747.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
143859 c.1336C>A p.H446N Chr.16: 298170-298170 on Build GRCh38
6981490 c.1349G>A p.R450H Chr.16: 298157-298157 on Build GRCh38
4821406 c.1344G>A p.P448P Chr.16: 298162-298162 on Build GRCh38
1609308 c.1371_1372insC p.G458fs*11 Chr.16: 298134-298135 on Build GRCh38
5043574 c.1255G>T p.E419* Chr.16: 298251-298251 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775371593 Chr.16: 298291-298291 on Build GRCh38
rs776159008 Chr.16: 298154-298154 on Build GRCh38
rs750617218 Chr.16: 298205-298205 on Build GRCh38
rs952813669 Chr.16: 298292-298292 on Build GRCh38
rs200184098 Chr.16: 298146-298146 on Build GRCh38
rs1040609642 Chr.16: 298278-298278 on Build GRCh38
rs994094846 Chr.16: 298132-298132 on Build GRCh38
rs748733363 Chr.16: 298230-298230 on Build GRCh38
rs533366295 Chr.16: 298237-298237 on Build GRCh38
rs755028316 Chr.16: 298223-298223 on Build GRCh38
rs923545793 Chr.16: 298280-298280 on Build GRCh38
rs773763817 Chr.16: 298234-298234 on Build GRCh38
rs748975424 Chr.16: 298128-298128 on Build GRCh38
rs214250 Chr.16: 298222-298222 on Build GRCh38
rs770548875 Chr.16: 298239-298239 on Build GRCh38
rs531887973 Chr.16: 298161-298161 on Build GRCh38
rs1047458992 Chr.16: 298134-298134 on Build GRCh38
rs779554309 Chr.16: 298224-298224 on Build GRCh38
rs753368709 Chr.16: 298261-298261 on Build GRCh38
rs765161459 Chr.16: 298179-298179 on Build GRCh38
rs540857771 Chr.16: 298186-298186 on Build GRCh38
rs147589868 Chr.16: 298273-298273 on Build GRCh38
rs975441164 Chr.16: 298272-298272 on Build GRCh38
rs776159575 Chr.16: 298258-298258 on Build GRCh38
rs778456607 Chr.16: 298190-298190 on Build GRCh38
rs529484795 Chr.16: 298216-298216 on Build GRCh38
rs966557459 Chr.16: 298231-298231 on Build GRCh38
rs747637123 Chr.16: 298218-298218 on Build GRCh38
rs374481133 Chr.16: 298158-298158 on Build GRCh38
rs373062859 Chr.16: 298266-298266 on Build GRCh38
rs773229103 Chr.16: 298226-298226 on Build GRCh38
rs760417274 Chr.16: 298135-298135 on Build GRCh38
rs562247902 Chr.16: 298204-298204 on Build GRCh38
rs982913334 Chr.16: 298297-298297 on Build GRCh38
rs116350678 Chr.16: 298233-298233 on Build GRCh38
rs756308108 Chr.16: 298213-298213 on Build GRCh38
rs759073941 Chr.16: 298263-298263 on Build GRCh38
rs752688778 Chr.16: 298180-298180 on Build GRCh38
rs560238796 Chr.16: 298127-298127 on Build GRCh38

View Full Product Details