Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 34856911-34857168 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AQR
Entrez Gene ID
9716
Gene Name
aquarius intron-binding spliceosomal factor
Gene Aliases
IBP160, fSAP164
UniGene
Hs.510958
Gene Chromosome Location
Chr.15: 34856351-34969794 on Build GRCh38
Transcripts
NM_014691.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1749100 c.4202A>T p.E1401V Chr.15: 34857048-34857048 on Build GRCh38
6602660 c.4285G>T p.E1429* Chr.15: 34856965-34856965 on Build GRCh38
5054254 c.4275C>T p.S1425S Chr.15: 34856975-34856975 on Build GRCh38
960896 c.4246G>C p.D1416H Chr.15: 34857004-34857004 on Build GRCh38
470532 c.4280G>A p.R1427H Chr.15: 34856970-34856970 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757489039 Chr.15: 34856941-34856941 on Build GRCh38
rs773911381 Chr.15: 34856985-34856985 on Build GRCh38
rs780657298 Chr.15: 34856966-34856966 on Build GRCh38
rs200139526 Chr.15: 34856960-34856960 on Build GRCh38
rs1032821885 Chr.15: 34857026-34857026 on Build GRCh38
rs760602707 Chr.15: 34857010-34857010 on Build GRCh38
rs780283450 Chr.15: 34857077-34857077 on Build GRCh38
rs75861957 Chr.15: 34857125-34857125 on Build GRCh38
rs750484796 Chr.15: 34856959-34856959 on Build GRCh38
rs371217912 Chr.15: 34857112-34857113 on Build GRCh38
rs375985364 Chr.15: 34856934-34856934 on Build GRCh38
rs756556527 Chr.15: 34857123-34857126 on Build GRCh38
rs1021795969 Chr.15: 34857016-34857016 on Build GRCh38
rs765458983 Chr.15: 34857058-34857058 on Build GRCh38
rs79241448 Chr.15: 34857124-34857124 on Build GRCh38
rs762695504 Chr.15: 34857111-34857111 on Build GRCh38
rs371203440 Chr.15: 34857095-34857095 on Build GRCh38
rs753645474 Chr.15: 34857028-34857028 on Build GRCh38
rs190121355 Chr.15: 34856937-34856937 on Build GRCh38
rs758444517 Chr.15: 34857070-34857070 on Build GRCh38
rs761554059 Chr.15: 34857046-34857046 on Build GRCh38
rs1011781151 Chr.15: 34857014-34857014 on Build GRCh38
rs75851493 Chr.15: 34857109-34857109 on Build GRCh38
rs768083628 Chr.15: 34857113-34857114 on Build GRCh38
rs753207769 Chr.15: 34857120-34857121 on Build GRCh38
rs560642086 Chr.15: 34857122-34857122 on Build GRCh38
rs781775827 Chr.15: 34857121-34857121 on Build GRCh38
rs764079226 Chr.15: 34857018-34857018 on Build GRCh38
rs771586153 Chr.15: 34856992-34856992 on Build GRCh38
rs573724519 Chr.15: 34856970-34856970 on Build GRCh38
rs78487073 Chr.15: 34857108-34857108 on Build GRCh38
rs750632859 Chr.15: 34857068-34857068 on Build GRCh38
rs373744153 Chr.15: 34856981-34856981 on Build GRCh38
rs778224215 Chr.15: 34857130-34857130 on Build GRCh38
rs759041598 Chr.15: 34856991-34856991 on Build GRCh38
rs778574058 Chr.15: 34857135-34857135 on Build GRCh38
rs769072924 Chr.15: 34856971-34856971 on Build GRCh38
rs755526215 Chr.15: 34857116-34857116 on Build GRCh38
rs763883158 Chr.15: 34856935-34856935 on Build GRCh38
rs200045468 Chr.15: 34857126-34857126 on Build GRCh38
rs867377756 Chr.15: 34856933-34856933 on Build GRCh38
rs975153872 Chr.15: 34857047-34857049 on Build GRCh38
rs368732448 Chr.15: 34857006-34857006 on Build GRCh38
rs765919946 Chr.15: 34857113-34857113 on Build GRCh38
rs555623203 Chr.15: 34856955-34856955 on Build GRCh38
rs200963145 Chr.15: 34856962-34856962 on Build GRCh38

View Full Product Details