Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 87872874-87873147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF4L2
Entrez Gene ID
138009
Gene Name
DDB1 and CUL4 associated factor 4 like 2
Gene Aliases
WDR21C
UniGene
Hs.371738
Gene Chromosome Location
Chr.8: 87870743-87874068 on Build GRCh38
Transcripts
NM_152418.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1743146 c.919C>A p.Q307K Chr.8: 87873053-87873053 on Build GRCh38
5559162 c.837C>A p.L279L Chr.8: 87873135-87873135 on Build GRCh38
6181842 c.1031G>T p.R344L Chr.8: 87872941-87872941 on Build GRCh38
3651598 c.966G>C p.V322V Chr.8: 87873006-87873006 on Build GRCh38
4620258 c.967A>T p.N323Y Chr.8: 87873005-87873005 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139934952 Chr.8: 87872979-87872979 on Build GRCh38
rs773285935 Chr.8: 87872988-87872988 on Build GRCh38
rs529069612 Chr.8: 87872962-87872962 on Build GRCh38
rs762077763 Chr.8: 87872901-87872901 on Build GRCh38
rs962130156 Chr.8: 87872927-87872927 on Build GRCh38
rs765137281 Chr.8: 87872902-87872902 on Build GRCh38
rs753548822 Chr.8: 87873097-87873097 on Build GRCh38
rs765613159 Chr.8: 87872987-87872987 on Build GRCh38
rs747563111 Chr.8: 87873021-87873021 on Build GRCh38
rs202221876 Chr.8: 87873112-87873112 on Build GRCh38
rs540135166 Chr.8: 87872907-87872907 on Build GRCh38
rs948580084 Chr.8: 87872912-87872912 on Build GRCh38
rs141645313 Chr.8: 87873034-87873034 on Build GRCh38
rs146030031 Chr.8: 87872964-87872964 on Build GRCh38
rs138662456 Chr.8: 87873002-87873002 on Build GRCh38
rs767496556 Chr.8: 87873067-87873067 on Build GRCh38
rs746297789 Chr.8: 87872960-87872960 on Build GRCh38
rs755701425 Chr.8: 87872941-87872941 on Build GRCh38
rs766051160 Chr.8: 87872994-87872994 on Build GRCh38
rs142735829 Chr.8: 87872919-87872919 on Build GRCh38
rs143361209 Chr.8: 87873015-87873015 on Build GRCh38
rs777447745 Chr.8: 87873006-87873006 on Build GRCh38
rs977224604 Chr.8: 87873026-87873026 on Build GRCh38
rs762847637 Chr.8: 87872991-87872991 on Build GRCh38
rs773515750 Chr.8: 87873036-87873036 on Build GRCh38
rs754549912 Chr.8: 87872934-87872934 on Build GRCh38
rs370189631 Chr.8: 87873003-87873003 on Build GRCh38
rs766389714 Chr.8: 87872911-87872911 on Build GRCh38
rs150153725 Chr.8: 87872904-87872904 on Build GRCh38
rs762130958 Chr.8: 87872984-87872984 on Build GRCh38
rs551327930 Chr.8: 87873086-87873086 on Build GRCh38
rs751935639 Chr.8: 87873109-87873109 on Build GRCh38
rs199898936 Chr.8: 87873087-87873087 on Build GRCh38
rs973382388 Chr.8: 87873022-87873022 on Build GRCh38
rs376130986 Chr.8: 87873094-87873094 on Build GRCh38
rs201502417 Chr.8: 87872999-87872999 on Build GRCh38
rs756800765 Chr.8: 87873104-87873104 on Build GRCh38
rs773954462 Chr.8: 87873054-87873054 on Build GRCh38
rs955825666 Chr.8: 87872947-87872947 on Build GRCh38
rs747432904 Chr.8: 87872978-87872978 on Build GRCh38
rs754142692 Chr.8: 87872895-87872895 on Build GRCh38
rs777265160 Chr.8: 87873027-87873027 on Build GRCh38
rs781774386 Chr.8: 87873123-87873123 on Build GRCh38
rs199636183 Chr.8: 87872932-87872932 on Build GRCh38
rs780634469 Chr.8: 87872935-87872935 on Build GRCh38
rs759325348 Chr.8: 87873059-87873059 on Build GRCh38
rs754816770 Chr.8: 87872997-87872997 on Build GRCh38
rs766213640 Chr.8: 87873053-87873053 on Build GRCh38
rs747816185 Chr.8: 87872939-87872939 on Build GRCh38
rs146716352 Chr.8: 87873028-87873028 on Build GRCh38
rs748857948 Chr.8: 87873010-87873010 on Build GRCh38
rs200613731 Chr.8: 87872995-87872995 on Build GRCh38
rs370434676 Chr.8: 87873035-87873035 on Build GRCh38
rs149891752 Chr.8: 87873126-87873126 on Build GRCh38
rs916639894 Chr.8: 87872905-87872905 on Build GRCh38
rs189636421 Chr.8: 87873125-87873125 on Build GRCh38
rs761571207 Chr.8: 87872937-87872938 on Build GRCh38
rs1045539745 Chr.8: 87872916-87872916 on Build GRCh38
rs537310751 Chr.8: 87872993-87872995 on Build GRCh38
rs769218201 Chr.8: 87873024-87873024 on Build GRCh38
rs373840760 Chr.8: 87873075-87873075 on Build GRCh38
rs763038706 Chr.8: 87873048-87873048 on Build GRCh38
rs201314325 Chr.8: 87872945-87872945 on Build GRCh38

View Full Product Details