Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 75206729-75206992 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C15orf39
Entrez Gene ID
56905
Gene Name
chromosome 15 open reading frame 39
Gene Aliases
-
UniGene
Hs.17936
Gene Chromosome Location
Chr.15: 75201880-75212169 on Build GRCh38
Transcripts
NM_015492.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6614377 c.939C>T p.T313T Chr.15: 75206987-75206987 on Build GRCh38
6614375 c.865T>C p.C289R Chr.15: 75206913-75206913 on Build GRCh38
6614381 c.704C>A p.P235H Chr.15: 75206752-75206752 on Build GRCh38
6614378 c.939C>T p.T313T Chr.15: 75206987-75206987 on Build GRCh38
5601133 c.763C>T p.Q255* Chr.15: 75206811-75206811 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762760056 Chr.15: 75206853-75206853 on Build GRCh38
rs200426504 Chr.15: 75206828-75206828 on Build GRCh38
rs770736892 Chr.15: 75206854-75206868 on Build GRCh38
rs554751240 Chr.15: 75206896-75206896 on Build GRCh38
rs35337310 Chr.15: 75206850-75206851 on Build GRCh38
rs138518753 Chr.15: 75206775-75206775 on Build GRCh38
rs754120764 Chr.15: 75206826-75206826 on Build GRCh38
rs780161865 Chr.15: 75206824-75206824 on Build GRCh38
rs201220527 Chr.15: 75206794-75206794 on Build GRCh38
rs536795576 Chr.15: 75206886-75206886 on Build GRCh38
rs965024950 Chr.15: 75206847-75206847 on Build GRCh38
rs376673047 Chr.15: 75206963-75206963 on Build GRCh38
rs368905041 Chr.15: 75206972-75206972 on Build GRCh38
rs760652184 Chr.15: 75206887-75206887 on Build GRCh38
rs757777642 Chr.15: 75206968-75206968 on Build GRCh38
rs747518018 Chr.15: 75206841-75206841 on Build GRCh38
rs201183267 Chr.15: 75206918-75206918 on Build GRCh38
rs761533466 Chr.15: 75206885-75206885 on Build GRCh38
rs768423656 Chr.15: 75206877-75206877 on Build GRCh38
rs756191912 Chr.15: 75206820-75206820 on Build GRCh38
rs762672229 Chr.15: 75206784-75206784 on Build GRCh38
rs372675349 Chr.15: 75206930-75206930 on Build GRCh38
rs138715960 Chr.15: 75206953-75206953 on Build GRCh38
rs769235707 Chr.15: 75206844-75206844 on Build GRCh38
rs71440223 Chr.15: 75206844-75206844 on Build GRCh38
rs201584125 Chr.15: 75206916-75206916 on Build GRCh38
rs772435271 Chr.15: 75206834-75206834 on Build GRCh38
rs752877035 Chr.15: 75206919-75206919 on Build GRCh38
rs908594946 Chr.15: 75206881-75206881 on Build GRCh38
rs867257282 Chr.15: 75206934-75206934 on Build GRCh38
rs766439758 Chr.15: 75206890-75206890 on Build GRCh38
rs775043070 Chr.15: 75206782-75206782 on Build GRCh38
rs761630119 Chr.15: 75206805-75206805 on Build GRCh38
rs973335139 Chr.15: 75206858-75206858 on Build GRCh38
rs147093913 Chr.15: 75206750-75206750 on Build GRCh38
rs146917321 Chr.15: 75206840-75206840 on Build GRCh38
rs748815589 Chr.15: 75206974-75206974 on Build GRCh38
rs772462766 Chr.15: 75206762-75206762 on Build GRCh38
rs539327340 Chr.15: 75206772-75206772 on Build GRCh38
rs144078010 Chr.15: 75206801-75206801 on Build GRCh38
rs747527222 Chr.15: 75206967-75206967 on Build GRCh38
rs748526077 Chr.15: 75206830-75206830 on Build GRCh38
rs919949911 Chr.15: 75206861-75206861 on Build GRCh38
rs750322458 Chr.15: 75206806-75206806 on Build GRCh38
rs775125471 Chr.15: 75206846-75206846 on Build GRCh38
rs965800328 Chr.15: 75206827-75206827 on Build GRCh38
rs984461998 Chr.15: 75206921-75206921 on Build GRCh38
rs774084534 Chr.15: 75206878-75206878 on Build GRCh38
rs982877535 Chr.15: 75206876-75206876 on Build GRCh38
rs931279210 Chr.15: 75206866-75206866 on Build GRCh38
rs769306778 Chr.15: 75206778-75206778 on Build GRCh38
rs918892046 Chr.15: 75206884-75206884 on Build GRCh38
rs753921029 Chr.15: 75206905-75206905 on Build GRCh38
rs71440224 Chr.15: 75206847-75206848 on Build GRCh38

View Full Product Details