Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 2239469-2239728 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRM2
Entrez Gene ID
29960
Gene Name
mitochondrial rRNA methyltransferase 2
Gene Aliases
FJH1, FTSJ2, HEL97, RRMJ2
UniGene
Hs.279877
Gene Chromosome Location
Chr.7: 2234291-2242198 on Build GRCh38
Transcripts
NM_013393.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6177326 c.33C>T p.S11S Chr.7: 2239683-2239683 on Build GRCh38
3637485 c.247C>T p.P83S Chr.7: 2239469-2239469 on Build GRCh38
6177327 c.63G>C p.G21G Chr.7: 2239653-2239653 on Build GRCh38
6109714 c.137C>T p.A46V Chr.7: 2239579-2239579 on Build GRCh38
3637488 c.168C>T p.S56S Chr.7: 2239548-2239548 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138187457 Chr.7: 2239649-2239649 on Build GRCh38
rs746259393 Chr.7: 2239679-2239679 on Build GRCh38
rs746029844 Chr.7: 2239604-2239604 on Build GRCh38
rs772720493 Chr.7: 2239524-2239524 on Build GRCh38
rs757715878 Chr.7: 2239675-2239675 on Build GRCh38
rs201864572 Chr.7: 2239530-2239530 on Build GRCh38
rs765231837 Chr.7: 2239540-2239540 on Build GRCh38
rs752227558 Chr.7: 2239658-2239658 on Build GRCh38
rs775504312 Chr.7: 2239627-2239627 on Build GRCh38
rs55904231 Chr.7: 2239684-2239684 on Build GRCh38
rs142976260 Chr.7: 2239575-2239575 on Build GRCh38
rs557166200 Chr.7: 2239707-2239707 on Build GRCh38
rs998979267 Chr.7: 2239688-2239688 on Build GRCh38
rs149945416 Chr.7: 2239500-2239500 on Build GRCh38
rs748600680 Chr.7: 2239515-2239515 on Build GRCh38
rs116607388 Chr.7: 2239676-2239676 on Build GRCh38
rs766133599 Chr.7: 2239633-2239633 on Build GRCh38
rs749356400 Chr.7: 2239705-2239705 on Build GRCh38
rs143741179 Chr.7: 2239492-2239492 on Build GRCh38
rs765014343 Chr.7: 2239653-2239653 on Build GRCh38
rs747620316 Chr.7: 2239670-2239670 on Build GRCh38
rs758934191 Chr.7: 2239571-2239571 on Build GRCh38
rs145102163 Chr.7: 2239548-2239548 on Build GRCh38
rs150856697 Chr.7: 2239579-2239579 on Build GRCh38
rs369728243 Chr.7: 2239631-2239631 on Build GRCh38
rs138541422 Chr.7: 2239493-2239493 on Build GRCh38
rs761136679 Chr.7: 2239529-2239529 on Build GRCh38
rs753589819 Chr.7: 2239637-2239637 on Build GRCh38
rs994971472 Chr.7: 2239573-2239573 on Build GRCh38
rs144143432 Chr.7: 2239497-2239497 on Build GRCh38
rs763061299 Chr.7: 2239628-2239628 on Build GRCh38
rs768337659 Chr.7: 2239504-2239504 on Build GRCh38
rs375028016 Chr.7: 2239704-2239704 on Build GRCh38
rs753590801 Chr.7: 2239559-2239559 on Build GRCh38
rs761685975 Chr.7: 2239545-2239545 on Build GRCh38
rs368426054 Chr.7: 2239606-2239606 on Build GRCh38
rs748357200 Chr.7: 2239505-2239505 on Build GRCh38
rs928430449 Chr.7: 2239605-2239605 on Build GRCh38
rs775557566 Chr.7: 2239542-2239542 on Build GRCh38
rs372451168 Chr.7: 2239607-2239607 on Build GRCh38
rs990592377 Chr.7: 2239557-2239557 on Build GRCh38
rs183764913 Chr.7: 2239629-2239629 on Build GRCh38
rs376300149 Chr.7: 2239552-2239552 on Build GRCh38
rs1009062342 Chr.7: 2239580-2239580 on Build GRCh38
rs139072008 Chr.7: 2239531-2239531 on Build GRCh38
rs778132157 Chr.7: 2239667-2239667 on Build GRCh38
rs934733645 Chr.7: 2239519-2239519 on Build GRCh38
rs754684144 Chr.7: 2239565-2239565 on Build GRCh38
rs778509302 Chr.7: 2239569-2239569 on Build GRCh38
rs765932500 Chr.7: 2239558-2239558 on Build GRCh38
rs747316647 Chr.7: 2239576-2239576 on Build GRCh38
rs776716401 Chr.7: 2239586-2239586 on Build GRCh38
rs371623071 Chr.7: 2239490-2239490 on Build GRCh38
rs747684401 Chr.7: 2239570-2239570 on Build GRCh38
rs758034545 Chr.7: 2239659-2239659 on Build GRCh38

View Full Product Details