Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 31408947-31409216 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DROSHA
Entrez Gene ID
29102
Gene Name
drosha ribonuclease III
Gene Aliases
ETOHI2, HSA242976, RANSE3L, RN3, RNASE3L, RNASEN
UniGene
Hs.97997
Gene Chromosome Location
Chr.5: 31400494-31532175 on Build GRCh38
Transcripts
NM_001100412.1 NM_013235.4 XM_005248291.3 XM_005248292.3 XM_005248293.3 XM_005248294.3 XM_017009399.1 XM_017009400.1 XM_017009401.1 XM_017009402.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
738165 c.3823G>T p.E1275* Chr.5: 31409087-31409087 on Build GRCh38
3014362 c.3835C>A p.P1279T Chr.5: 31409075-31409075 on Build GRCh38
4003496 c.3765T>C p.N1255N Chr.5: 31409145-31409145 on Build GRCh38
1264327 c.3819G>A p.R1273R Chr.5: 31409091-31409091 on Build GRCh38
1287647 c.3849G>C p.L1283L Chr.5: 31409061-31409061 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs76613646 Chr.5: 31409018-31409018 on Build GRCh38
rs757353501 Chr.5: 31409164-31409164 on Build GRCh38
rs1002990980 Chr.5: 31409086-31409086 on Build GRCh38
rs770406706 Chr.5: 31409129-31409129 on Build GRCh38
rs1013050576 Chr.5: 31408985-31408985 on Build GRCh38
rs541676959 Chr.5: 31408980-31408980 on Build GRCh38
rs372856536 Chr.5: 31409049-31409049 on Build GRCh38
rs751507949 Chr.5: 31409028-31409028 on Build GRCh38
rs990437331 Chr.5: 31409179-31409179 on Build GRCh38
rs989689507 Chr.5: 31409085-31409085 on Build GRCh38
rs956254570 Chr.5: 31409163-31409163 on Build GRCh38
rs747510301 Chr.5: 31409135-31409135 on Build GRCh38
rs181677016 Chr.5: 31409103-31409103 on Build GRCh38
rs773892099 Chr.5: 31409133-31409133 on Build GRCh38
rs754934125 Chr.5: 31409032-31409032 on Build GRCh38
rs7727617 Chr.5: 31409044-31409044 on Build GRCh38
rs780328063 Chr.5: 31409027-31409027 on Build GRCh38
rs186198647 Chr.5: 31409157-31409157 on Build GRCh38
rs769635822 Chr.5: 31409094-31409094 on Build GRCh38
rs781024074 Chr.5: 31409048-31409048 on Build GRCh38
rs2241337 Chr.5: 31409145-31409145 on Build GRCh38
rs644236 Chr.5: 31409008-31409008 on Build GRCh38
rs970769826 Chr.5: 31408991-31408991 on Build GRCh38
rs368139258 Chr.5: 31408997-31408997 on Build GRCh38

View Full Product Details