Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150747720-150747973 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSS
Entrez Gene ID
1520
Gene Name
cathepsin S
Gene Aliases
-
UniGene
Hs.181301
Gene Chromosome Location
Chr.1: 150730196-150765957 on Build GRCh38
Transcripts
NM_001199739.1 NM_004079.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1579699 c.862C>G p.L288V Chr.1: 150747811-150747811 on Build GRCh38
3473484 c.835G>A p.G279S Chr.1: 150747838-150747838 on Build GRCh38
3473483 c.868G>T p.G290W Chr.1: 150747805-150747805 on Build GRCh38
1579698 c.865A>T p.N289Y Chr.1: 150747808-150747808 on Build GRCh38
1579697 c.866A>G p.N289S Chr.1: 150747807-150747807 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs58471571 Chr.1: 150747851-150747851 on Build GRCh38
rs779505650 Chr.1: 150747901-150747901 on Build GRCh38
rs777879777 Chr.1: 150747865-150747865 on Build GRCh38
rs752949974 Chr.1: 150747861-150747861 on Build GRCh38
rs971759177 Chr.1: 150747764-150747764 on Build GRCh38
rs768437857 Chr.1: 150747779-150747779 on Build GRCh38
rs80210253 Chr.1: 150747787-150747787 on Build GRCh38
rs142363816 Chr.1: 150747750-150747750 on Build GRCh38
rs750380196 Chr.1: 150747811-150747811 on Build GRCh38
rs587756697 Chr.1: 150747902-150747902 on Build GRCh38
rs928608408 Chr.1: 150747837-150747837 on Build GRCh38
rs751367182 Chr.1: 150747830-150747830 on Build GRCh38
rs771589626 Chr.1: 150747899-150747899 on Build GRCh38
rs762906143 Chr.1: 150747826-150747826 on Build GRCh38
rs781335273 Chr.1: 150747860-150747860 on Build GRCh38
rs879811377 Chr.1: 150747870-150747870 on Build GRCh38
rs756196248 Chr.1: 150747862-150747862 on Build GRCh38
rs369500729 Chr.1: 150747894-150747894 on Build GRCh38
rs766251331 Chr.1: 150747827-150747827 on Build GRCh38
rs754699303 Chr.1: 150747845-150747845 on Build GRCh38
rs896960342 Chr.1: 150747945-150747945 on Build GRCh38
rs924488332 Chr.1: 150747804-150747804 on Build GRCh38
rs140408537 Chr.1: 150747785-150747785 on Build GRCh38
rs587638060 Chr.1: 150747929-150747929 on Build GRCh38
rs757613058 Chr.1: 150747898-150747899 on Build GRCh38
rs759933150 Chr.1: 150747756-150747756 on Build GRCh38
rs376919416 Chr.1: 150747752-150747752 on Build GRCh38

View Full Product Details