Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 130476818-130477051 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASS1
Entrez Gene ID
445
Gene Name
argininosuccinate synthase 1
Gene Aliases
ASS, CTLN1
UniGene
Hs.160786
Gene Chromosome Location
Chr.9: 130444707-130501274 on Build GRCh38
Transcripts
NM_000050.4 NM_054012.3 XM_005272200.3 XM_011518705.2 XM_017014729.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6202861 c.603A>T p.Q201H Chr.9: 130476876-130476876 on Build GRCh38
4504198 c.658C>T p.P220S Chr.9: 130476931-130476931 on Build GRCh38
6606097 c.647delC p.N218fs*16 Chr.9: 130476920-130476920 on Build GRCh38
6606093 c.598-9delT p.? Chr.9: 130476862-130476862 on Build GRCh38
1701819 c.613G>A p.G205S Chr.9: 130476886-130476886 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200076976 Chr.9: 130477001-130477001 on Build GRCh38
rs750947418 Chr.9: 130476960-130476960 on Build GRCh38
rs764331417 Chr.9: 130476943-130476943 on Build GRCh38
rs780742787 Chr.9: 130476856-130476857 on Build GRCh38
rs1043964127 Chr.9: 130476920-130476920 on Build GRCh38
rs749907969 Chr.9: 130476862-130476863 on Build GRCh38
rs376371866 Chr.9: 130476878-130476878 on Build GRCh38
rs966116020 Chr.9: 130476840-130476840 on Build GRCh38
rs756128489 Chr.9: 130476929-130476929 on Build GRCh38
rs62637575 Chr.9: 130476895-130476895 on Build GRCh38
rs769538241 Chr.9: 130476929-130476929 on Build GRCh38
rs766889872 Chr.9: 130476973-130476973 on Build GRCh38
rs78432485 Chr.9: 130476965-130476965 on Build GRCh38
rs58233547 Chr.9: 130476948-130476948 on Build GRCh38
rs147858743 Chr.9: 130476880-130476880 on Build GRCh38
rs757319617 Chr.9: 130476850-130476850 on Build GRCh38
rs763203138 Chr.9: 130476942-130476942 on Build GRCh38
rs373824533 Chr.9: 130477006-130477006 on Build GRCh38
rs776441071 Chr.9: 130476896-130476896 on Build GRCh38
rs759518085 Chr.9: 130476897-130476897 on Build GRCh38
rs775246400 Chr.9: 130476930-130476930 on Build GRCh38
rs1036591318 Chr.9: 130476997-130476997 on Build GRCh38
rs747850211 Chr.9: 130476879-130476879 on Build GRCh38
rs755341321 Chr.9: 130476986-130476986 on Build GRCh38
rs563922134 Chr.9: 130476874-130476874 on Build GRCh38
rs201784978 Chr.9: 130476837-130476837 on Build GRCh38
rs761813681 Chr.9: 130476949-130476949 on Build GRCh38
rs767628546 Chr.9: 130476950-130476950 on Build GRCh38
rs746536193 Chr.9: 130476886-130476886 on Build GRCh38
rs995001369 Chr.9: 130477022-130477022 on Build GRCh38
rs529023316 Chr.9: 130476985-130476985 on Build GRCh38
rs777579815 Chr.9: 130476990-130476990 on Build GRCh38
rs781572633 Chr.9: 130476868-130476868 on Build GRCh38
rs750874679 Chr.9: 130476873-130476873 on Build GRCh38
rs975805165 Chr.9: 130476851-130476851 on Build GRCh38
rs771033174 Chr.9: 130476983-130476983 on Build GRCh38
rs890497211 Chr.9: 130476971-130476971 on Build GRCh38
rs746602223 Chr.9: 130476993-130476993 on Build GRCh38

View Full Product Details