Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 40559305-40559560 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXO1
Entrez Gene ID
2308
Gene Name
forkhead box O1
Gene Aliases
FKH1, FKHR, FOXO1A
UniGene
Hs.370666
Gene Chromosome Location
Chr.13: 40555664-40666597 on Build GRCh38
Transcripts
NM_002015.3 XM_011535008.2 XM_011535010.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1300198 c.1941A>G p.T647T Chr.13: 40559550-40559550 on Build GRCh38
6666460 c.1961C>G p.S654* Chr.13: 40559530-40559530 on Build GRCh38
5108792 c.1947A>G p.T649T Chr.13: 40559544-40559544 on Build GRCh38
6933346 c.1931G>T p.S644I Chr.13: 40559560-40559560 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs568695333 Chr.13: 40559359-40559359 on Build GRCh38
rs769157838 Chr.13: 40559383-40559383 on Build GRCh38
rs1044648857 Chr.13: 40559400-40559400 on Build GRCh38
rs374930087 Chr.13: 40559506-40559506 on Build GRCh38
rs976584585 Chr.13: 40559457-40559457 on Build GRCh38
rs112585563 Chr.13: 40559374-40559374 on Build GRCh38
rs770526416 Chr.13: 40559494-40559494 on Build GRCh38
rs1047802662 Chr.13: 40559375-40559375 on Build GRCh38
rs562138604 Chr.13: 40559459-40559459 on Build GRCh38
rs1018227132 Chr.13: 40559449-40559449 on Build GRCh38
rs115476993 Chr.13: 40559368-40559368 on Build GRCh38
rs958087335 Chr.13: 40559387-40559387 on Build GRCh38
rs759124635 Chr.13: 40559448-40559448 on Build GRCh38
rs539369700 Chr.13: 40559429-40559429 on Build GRCh38
rs769354575 Chr.13: 40559534-40559534 on Build GRCh38
rs555894254 Chr.13: 40559452-40559452 on Build GRCh38
rs1013744652 Chr.13: 40559350-40559350 on Build GRCh38
rs1009738135 Chr.13: 40559470-40559470 on Build GRCh38
rs186583526 Chr.13: 40559438-40559438 on Build GRCh38
rs1032322935 Chr.13: 40559380-40559380 on Build GRCh38
rs571100538 Chr.13: 40559410-40559410 on Build GRCh38
rs987273613 Chr.13: 40559535-40559535 on Build GRCh38
rs957725137 Chr.13: 40559463-40559463 on Build GRCh38
rs778492835 Chr.13: 40559485-40559485 on Build GRCh38
rs775066641 Chr.13: 40559538-40559538 on Build GRCh38
rs781392626 Chr.13: 40559458-40559458 on Build GRCh38
rs895295212 Chr.13: 40559342-40559342 on Build GRCh38
rs548764406 Chr.13: 40559372-40559372 on Build GRCh38
rs954719653 Chr.13: 40559510-40559510 on Build GRCh38
rs181811457 Chr.13: 40559379-40559379 on Build GRCh38
rs747509005 Chr.13: 40559493-40559493 on Build GRCh38
rs749877266 Chr.13: 40559510-40559510 on Build GRCh38
rs745343501 Chr.13: 40559515-40559515 on Build GRCh38
rs1020662472 Chr.13: 40559504-40559504 on Build GRCh38

View Full Product Details