The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002462.4 | NP_002453.2 | 1 - 2 | 231 | 133 |
XM_017028351.1 | XP_016883840.1 | 1 - 2 | 174 | 133 | |
GenBank mRNA | DA351922.1 | - | 1 - 2 | 222 | 133 |