The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037872.2 | NP_001032961.1 | 18 - 19 | 3290 | 108 |
NM_001321454.1 | NP_001308383.1 | 19 - 20 | 3401 | 108 | |
NM_001321455.1 | NP_001308384.1 | 19 - 20 | 3447 | 108 | |
NM_001321458.1 | NP_001308387.1 | 19 - 20 | 3417 | 108 | |
NM_001321459.1 | NP_001308388.1 | 19 - 20 | 3493 | 108 | |
NM_001321460.1 | NP_001308389.1 | 19 - 20 | 3490 | 108 | |
NM_016316.3 | NP_057400.1 | 18 - 19 | 3293 | 108 | |
NR_135649.1 | 19 - 20 | 3490 | 108 | ||
NR_135650.1 | 19 - 20 | 3447 | 108 | ||
NR_135651.1 | 17 - 18 | 3124 | 108 | ||
NR_135652.1 | 18 - 19 | 3293 | 108 | ||
NR_135653.1 | 17 - 18 | 3127 | 108 | ||
XM_011511341.2 | XP_011509643.1 | 12 - 13 | 1910 | 108 | |
XM_017004309.1 | XP_016859798.1 | 19 - 20 | 3814 | 108 | |
XM_017004310.1 | XP_016859799.1 | 19 - 20 | 3180 | 108 | |
XM_017004311.1 | XP_016859800.1 | 19 - 20 | 3180 | 108 | |
XM_017004312.1 | XP_016859801.1 | 19 - 20 | 3393 | 108 | |
XM_017004313.1 | XP_016859802.1 | 18 - 19 | 5019 | 108 | |
XM_017004314.1 | XP_016859803.1 | 15 - 16 | 2670 | 108 | |
XM_017004315.1 | XP_016859804.1 | 15 - 16 | 2654 | 108 | |
GenBank mRNA | AB047646.1 | - | 18 - 19 | 3215 | 108 |
AF151538.1 | - | 18 - 19 | 3246 | 108 | |
AF206019.1 | - | 18 - 19 | 3255 | 108 | |
AF357886.1 | - | 18 - 19 | 3211 | 108 | |
AJ131720.1 | - | 10 - 11 | 1612 | 108 | |
AK025176.1 | - | 10 - 11 | 1572 | 108 | |
AK307719.1 | - | 18 - 19 | 3204 | 108 | |
BC037734.1 | - | 19 - 20 | 3428 | 108 | |
BC130411.1 | - | 17 - 18 | 3061 | 108 |