The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000059.3 | NP_000050.2 | 24 - 25 | 9488 | 94 |
GenBank mRNA | KP272108.1 | - | 1 - 2 | 144 | 94 |
U43746.1 | - | 24 - 25 | 9489 | 94 |