The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285439.1 | NP_001272368.1 | 13 - 14 | 1187 | 93 |
NM_001285440.1 | NP_001272369.1 | 13 - 14 | 1185 | 93 | |
NM_152756.4 | NP_689969.2 | 13 - 14 | 1187 | 93 | |
XM_006714463.3 | XP_006714526.1 | 13 - 14 | 1200 | 93 | |
XM_011514005.2 | XP_011512307.1 | 13 - 14 | 1200 | 93 | |
XM_011514006.2 | XP_011512308.1 | 12 - 13 | 1004 | 93 | |
XM_017009311.1 | XP_016864800.1 | 12 - 13 | 1120 | 93 | |
XM_017009312.1 | XP_016864801.1 | 12 - 13 | 1120 | 93 | |
XM_017009313.1 | XP_016864802.1 | 12 - 13 | 1141 | 93 | |
XM_017009314.1 | XP_016864803.1 | 12 - 13 | 1110 | 93 | |
XM_017009315.1 | XP_016864804.1 | 12 - 13 | 1110 | 93 | |
GenBank mRNA | AY515854.1 | - | 13 - 14 | 1183 | 93 |
BC137163.1 | - | 13 - 14 | 1180 | 93 | |
BC137164.1 | - | 13 - 14 | 1180 | 93 | |
BC144509.1 | - | 13 - 14 | 1180 | 93 | |
CR749280.1 | - | 13 - 14 | 1166 | 93 |