Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 75667785-75668289 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP6
Entrez Gene ID
4135
Gene Name
microtubule associated protein 6
Gene Aliases
MAP6-N, MTAP6, N-STOP, STOP
UniGene
Hs.585540
Gene Chromosome Location
Chr.11: 75586914-75671108 on Build GRCh38
Transcripts
NM_033063.1 NM_207577.1 XM_006718556.3 XM_011545039.2 XM_017017755.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5725559 c.519G>A p.W173* Chr.11: 75667851-75667851 on Build GRCh38
4700490 c.453G>A p.E151E Chr.11: 75667917-75667917 on Build GRCh38
4036752 c.101C>T p.S34L Chr.11: 75668269-75668269 on Build GRCh38
1605236 c.318G>A p.P106P Chr.11: 75668052-75668052 on Build GRCh38
6042584 c.391C>T p.Q131* Chr.11: 75667979-75667979 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752629392 Chr.11: 75667834-75667834 on Build GRCh38
rs772604171 Chr.11: 75668240-75668240 on Build GRCh38
rs943501024 Chr.11: 75667976-75667976 on Build GRCh38
rs756815241 Chr.11: 75667887-75667887 on Build GRCh38
rs550230646 Chr.11: 75668233-75668233 on Build GRCh38
rs960771886 Chr.11: 75667873-75667873 on Build GRCh38
rs759693147 Chr.11: 75668254-75668254 on Build GRCh38
rs754780916 Chr.11: 75667822-75667822 on Build GRCh38
rs988630661 Chr.11: 75668072-75668072 on Build GRCh38
rs533630603 Chr.11: 75668202-75668202 on Build GRCh38
rs747527031 Chr.11: 75668247-75668247 on Build GRCh38
rs1035520001 Chr.11: 75668067-75668067 on Build GRCh38
rs867767378 Chr.11: 75667865-75667865 on Build GRCh38
rs764942489 Chr.11: 75668004-75668004 on Build GRCh38
rs773117783 Chr.11: 75667946-75667946 on Build GRCh38
rs1026299496 Chr.11: 75667935-75667935 on Build GRCh38
rs547964028 Chr.11: 75668147-75668147 on Build GRCh38
rs762709416 Chr.11: 75668259-75668259 on Build GRCh38
rs748600606 Chr.11: 75667933-75667933 on Build GRCh38
rs905703316 Chr.11: 75668045-75668045 on Build GRCh38
rs115871621 Chr.11: 75667977-75667977 on Build GRCh38
rs976246585 Chr.11: 75668187-75668187 on Build GRCh38
rs779363523 Chr.11: 75668230-75668230 on Build GRCh38
rs751495165 Chr.11: 75668199-75668199 on Build GRCh38
rs765076493 Chr.11: 75667824-75667824 on Build GRCh38
rs370290525 Chr.11: 75667838-75667838 on Build GRCh38
rs985069751 Chr.11: 75668117-75668117 on Build GRCh38
rs755834067 Chr.11: 75668224-75668224 on Build GRCh38
rs913130402 Chr.11: 75668076-75668076 on Build GRCh38
rs771608951 Chr.11: 75668021-75668022 on Build GRCh38
rs770021891 Chr.11: 75668256-75668256 on Build GRCh38
rs1044666202 Chr.11: 75668043-75668043 on Build GRCh38
rs777211062 Chr.11: 75667867-75667867 on Build GRCh38
rs947344472 Chr.11: 75668037-75668037 on Build GRCh38
rs766655422 Chr.11: 75667808-75667808 on Build GRCh38
rs775278140 Chr.11: 75668229-75668230 on Build GRCh38
rs1026330535 Chr.11: 75668100-75668100 on Build GRCh38
rs772720420 Chr.11: 75667937-75667937 on Build GRCh38
rs993566644 Chr.11: 75668099-75668099 on Build GRCh38
rs947007990 Chr.11: 75668192-75668192 on Build GRCh38
rs1022810623 Chr.11: 75667809-75667809 on Build GRCh38
rs771037189 Chr.11: 75668251-75668251 on Build GRCh38
rs923429258 Chr.11: 75668190-75668190 on Build GRCh38
rs934394517 Chr.11: 75667994-75667994 on Build GRCh38
rs985417126 Chr.11: 75667973-75667973 on Build GRCh38
rs769132084 Chr.11: 75667931-75667931 on Build GRCh38
rs562348164 Chr.11: 75668052-75668052 on Build GRCh38
rs746555372 Chr.11: 75667878-75667878 on Build GRCh38
rs762824021 Chr.11: 75668013-75668013 on Build GRCh38
rs571182244 Chr.11: 75668191-75668191 on Build GRCh38
rs896594757 Chr.11: 75668098-75668098 on Build GRCh38
rs1018310470 Chr.11: 75668129-75668129 on Build GRCh38
rs745559278 Chr.11: 75667914-75667914 on Build GRCh38
rs1035068838 Chr.11: 75667869-75667869 on Build GRCh38
rs545418011 Chr.11: 75667812-75667812 on Build GRCh38
rs780862772 Chr.11: 75667902-75667902 on Build GRCh38
rs952356662 Chr.11: 75667941-75667941 on Build GRCh38
rs778320511 Chr.11: 75668246-75668246 on Build GRCh38
rs1003153118 Chr.11: 75668058-75668058 on Build GRCh38
rs527889112 Chr.11: 75668085-75668085 on Build GRCh38
rs775221669 Chr.11: 75668258-75668258 on Build GRCh38
rs750087400 Chr.11: 75668222-75668222 on Build GRCh38
rs1052749974 Chr.11: 75668022-75668022 on Build GRCh38
rs905669479 Chr.11: 75667836-75667836 on Build GRCh38
rs531469715 Chr.11: 75667974-75667974 on Build GRCh38
rs752498745 Chr.11: 75668007-75668007 on Build GRCh38
rs759741154 Chr.11: 75667980-75667980 on Build GRCh38
rs972096695 Chr.11: 75667930-75667930 on Build GRCh38
rs760776050 Chr.11: 75667958-75667958 on Build GRCh38
rs764054785 Chr.11: 75668114-75668114 on Build GRCh38
rs780627303 Chr.11: 75668205-75668205 on Build GRCh38
rs964989648 Chr.11: 75668183-75668183 on Build GRCh38
rs776575456 Chr.11: 75668253-75668253 on Build GRCh38

View Full Product Details