Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 95027116-95027621 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDM4E
Entrez Gene ID
390245
Gene Name
lysine demethylase 4E
Gene Aliases
JMJD2E, KDM4DL
UniGene
Hs.714692
Gene Chromosome Location
Chr.11: 95025258-95027596 on Build GRCh38
Transcripts
NM_001161630.1

SNP Detail

SNP ID Chromosome Location
rs563292612 Chr.11: 95027209-95027209 on Build GRCh38
rs960791318 Chr.11: 95027160-95027160 on Build GRCh38
rs532171688 Chr.11: 95027223-95027223 on Build GRCh38
rs892330304 Chr.11: 95027309-95027309 on Build GRCh38
rs919467321 Chr.11: 95027189-95027189 on Build GRCh38
rs77146219 Chr.11: 95027250-95027250 on Build GRCh38
rs965365578 Chr.11: 95027510-95027510 on Build GRCh38
rs781861775 Chr.11: 95027196-95027196 on Build GRCh38
rs1026412089 Chr.11: 95027458-95027458 on Build GRCh38
rs782080116 Chr.11: 95027159-95027159 on Build GRCh38
rs933729110 Chr.11: 95027584-95027584 on Build GRCh38
rs975046447 Chr.11: 95027526-95027526 on Build GRCh38
rs569061339 Chr.11: 95027340-95027340 on Build GRCh38
rs1033716117 Chr.11: 95027407-95027407 on Build GRCh38
rs180854624 Chr.11: 95027490-95027490 on Build GRCh38
rs913660000 Chr.11: 95027597-95027597 on Build GRCh38
rs534468367 Chr.11: 95027214-95027214 on Build GRCh38
rs546557158 Chr.11: 95027237-95027237 on Build GRCh38
rs773002475 Chr.11: 95027483-95027483 on Build GRCh38
rs933781467 Chr.11: 95027288-95027288 on Build GRCh38
rs903772768 Chr.11: 95027353-95027353 on Build GRCh38
rs558973966 Chr.11: 95027521-95027521 on Build GRCh38
rs12292301 Chr.11: 95027452-95027452 on Build GRCh38
rs540398082 Chr.11: 95027498-95027498 on Build GRCh38
rs920952798 Chr.11: 95027548-95027548 on Build GRCh38
rs149582732 Chr.11: 95027293-95027293 on Build GRCh38
rs144257378 Chr.11: 95027310-95027310 on Build GRCh38
rs376658611 Chr.11: 95027147-95027148 on Build GRCh38
rs78184421 Chr.11: 95027497-95027497 on Build GRCh38
rs1013703013 Chr.11: 95027457-95027457 on Build GRCh38
rs115129996 Chr.11: 95027594-95027594 on Build GRCh38
rs1043628026 Chr.11: 95027339-95027339 on Build GRCh38
rs761400681 Chr.11: 95027186-95027186 on Build GRCh38
rs909471703 Chr.11: 95027236-95027236 on Build GRCh38
rs773879161 Chr.11: 95027185-95027185 on Build GRCh38
rs782791017 Chr.11: 95027247-95027247 on Build GRCh38
rs1002177445 Chr.11: 95027398-95027398 on Build GRCh38

View Full Product Details