Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 101907549-101908056 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPTL5
Entrez Gene ID
253935
Gene Name
angiopoietin like 5
Gene Aliases
-
UniGene
Hs.318370
Gene Chromosome Location
Chr.11: 101890674-101916522 on Build GRCh38
Transcripts
NM_178127.4 XM_011542735.2 XM_017017466.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7272702 c.11C>A p.P4Q Chr.11: 101907899-101907899 on Build GRCh38
6262780 c.20C>A p.A7D Chr.11: 101907890-101907890 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs78267648 Chr.11: 101907584-101907584 on Build GRCh38
rs756694056 Chr.11: 101907921-101907922 on Build GRCh38
rs958406280 Chr.11: 101907706-101907706 on Build GRCh38
rs758791765 Chr.11: 101907831-101907831 on Build GRCh38
rs550425046 Chr.11: 101907721-101907721 on Build GRCh38
rs1046062041 Chr.11: 101908031-101908031 on Build GRCh38
rs76880133 Chr.11: 101907595-101907595 on Build GRCh38
rs371425912 Chr.11: 101907838-101907838 on Build GRCh38
rs745346414 Chr.11: 101907717-101907717 on Build GRCh38
rs778644014 Chr.11: 101907939-101907939 on Build GRCh38
rs769335412 Chr.11: 101907843-101907843 on Build GRCh38
rs765401643 Chr.11: 101907905-101907905 on Build GRCh38
rs113018418 Chr.11: 101907760-101907760 on Build GRCh38
rs757507727 Chr.11: 101907814-101907814 on Build GRCh38
rs954163656 Chr.11: 101907883-101907883 on Build GRCh38
rs772178264 Chr.11: 101907926-101907926 on Build GRCh38
rs1033858832 Chr.11: 101907676-101907676 on Build GRCh38
rs771770632 Chr.11: 101907865-101907865 on Build GRCh38
rs752610198 Chr.11: 101907783-101907783 on Build GRCh38
rs770759940 Chr.11: 101907856-101907856 on Build GRCh38
rs761904379 Chr.11: 101907899-101907899 on Build GRCh38
rs760749002 Chr.11: 101907791-101907791 on Build GRCh38
rs750737032 Chr.11: 101907828-101907828 on Build GRCh38
rs562130256 Chr.11: 101907582-101907582 on Build GRCh38
rs1002330368 Chr.11: 101907656-101907656 on Build GRCh38
rs199507393 Chr.11: 101907921-101907921 on Build GRCh38
rs746880308 Chr.11: 101908011-101908011 on Build GRCh38
rs767796607 Chr.11: 101907788-101907792 on Build GRCh38
rs969089090 Chr.11: 101907673-101907673 on Build GRCh38
rs923214842 Chr.11: 101907853-101907853 on Build GRCh38
rs571759946 Chr.11: 101908010-101908010 on Build GRCh38
rs919159037 Chr.11: 101907967-101907967 on Build GRCh38
rs374846209 Chr.11: 101907895-101907895 on Build GRCh38
rs771257892 Chr.11: 101907578-101907578 on Build GRCh38
rs760593828 Chr.11: 101907885-101907885 on Build GRCh38
rs748917714 Chr.11: 101907845-101907845 on Build GRCh38
rs755477929 Chr.11: 101907924-101907924 on Build GRCh38
rs906156343 Chr.11: 101908033-101908033 on Build GRCh38
rs770600096 Chr.11: 101907937-101907937 on Build GRCh38
rs758733465 Chr.11: 101907909-101907909 on Build GRCh38
rs779227395 Chr.11: 101907815-101907815 on Build GRCh38
rs966729852 Chr.11: 101907813-101907813 on Build GRCh38
rs956600256 Chr.11: 101907750-101907750 on Build GRCh38
rs865913680 Chr.11: 101907947-101907947 on Build GRCh38
rs746939271 Chr.11: 101907961-101907961 on Build GRCh38
rs570273151 Chr.11: 101907799-101907799 on Build GRCh38
rs777319075 Chr.11: 101907927-101907927 on Build GRCh38
rs759368494 Chr.11: 101907857-101907857 on Build GRCh38
rs764134774 Chr.11: 101907793-101907793 on Build GRCh38
rs747486175 Chr.11: 101907840-101907840 on Build GRCh38
rs750645502 Chr.11: 101907907-101907907 on Build GRCh38
rs1037613629 Chr.11: 101907956-101907956 on Build GRCh38
rs527798851 Chr.11: 101907583-101907583 on Build GRCh38
rs745570692 Chr.11: 101907948-101907948 on Build GRCh38
rs539688597 Chr.11: 101907712-101907712 on Build GRCh38
rs1661410 Chr.11: 101907979-101907979 on Build GRCh38
rs775185790 Chr.11: 101907954-101907954 on Build GRCh38
rs1011200504 Chr.11: 101907751-101907751 on Build GRCh38
rs139580863 Chr.11: 101907818-101907818 on Build GRCh38
rs1023182575 Chr.11: 101908029-101908029 on Build GRCh38
rs922570065 Chr.11: 101907870-101907870 on Build GRCh38
rs780342795 Chr.11: 101907914-101907914 on Build GRCh38
rs906303097 Chr.11: 101907620-101907620 on Build GRCh38
rs1021209503 Chr.11: 101907798-101907798 on Build GRCh38
rs976730077 Chr.11: 101907822-101907822 on Build GRCh38
rs771893542 Chr.11: 101907953-101907953 on Build GRCh38
rs753198412 Chr.11: 101907836-101907836 on Build GRCh38
rs144300304 Chr.11: 101907869-101907869 on Build GRCh38
rs748762177 Chr.11: 101907929-101907929 on Build GRCh38
rs895956970 Chr.11: 101907980-101907980 on Build GRCh38
rs978870741 Chr.11: 101907704-101907704 on Build GRCh38

View Full Product Details