Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 193195-193688 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC6A12
Entrez Gene ID
6539
Gene Name
solute carrier family 6 member 12
Gene Aliases
BGT-1, BGT1, GAT2
UniGene
Hs.437174
Gene Chromosome Location
Chr.12: 190077-214523 on Build GRCh38
Transcripts
NM_001122847.2 NM_001122848.2 NM_001206931.1 NM_003044.4 XM_005253747.4 XM_005253748.4 XM_006719005.3 XM_011521010.1 XM_017019841.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7085611 c.1464G>A p.M488I Chr.12: 193343-193343 on Build GRCh38
6462541 c.1529T>C p.L510P Chr.12: 193278-193278 on Build GRCh38
4146988 c.1518T>A p.P506P Chr.12: 193289-193289 on Build GRCh38
938617 c.1439G>A p.R480H Chr.12: 193368-193368 on Build GRCh38
693407 c.1480T>C p.W494R Chr.12: 193327-193327 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs975212490 Chr.12: 193403-193403 on Build GRCh38
rs758345561 Chr.12: 193295-193295 on Build GRCh38
rs138178078 Chr.12: 193326-193326 on Build GRCh38
rs386759334 Chr.12: 193594-193595 on Build GRCh38
rs759427029 Chr.12: 193332-193332 on Build GRCh38
rs750737377 Chr.12: 193328-193328 on Build GRCh38
rs774114087 Chr.12: 193244-193244 on Build GRCh38
rs765452457 Chr.12: 193334-193334 on Build GRCh38
rs775342805 Chr.12: 193419-193419 on Build GRCh38
rs141848384 Chr.12: 193333-193333 on Build GRCh38
rs201403020 Chr.12: 193374-193374 on Build GRCh38
rs1038846754 Chr.12: 193280-193280 on Build GRCh38
rs752453739 Chr.12: 193257-193257 on Build GRCh38
rs755490867 Chr.12: 193318-193318 on Build GRCh38
rs78715843 Chr.12: 193440-193440 on Build GRCh38
rs117257289 Chr.12: 193629-193629 on Build GRCh38
rs981500514 Chr.12: 193650-193650 on Build GRCh38
rs1024721906 Chr.12: 193648-193648 on Build GRCh38
rs138252563 Chr.12: 193404-193404 on Build GRCh38
rs747426899 Chr.12: 193311-193311 on Build GRCh38
rs1037908258 Chr.12: 193510-193510 on Build GRCh38
rs561661233 Chr.12: 193560-193560 on Build GRCh38
rs561457444 Chr.12: 193572-193572 on Build GRCh38
rs200299625 Chr.12: 193392-193392 on Build GRCh38
rs769582586 Chr.12: 193325-193325 on Build GRCh38
rs781719176 Chr.12: 193324-193324 on Build GRCh38
rs377675131 Chr.12: 193389-193389 on Build GRCh38
rs772013395 Chr.12: 193416-193416 on Build GRCh38
rs114757719 Chr.12: 193567-193567 on Build GRCh38
rs1022037755 Chr.12: 193559-193559 on Build GRCh38
rs756505662 Chr.12: 193383-193383 on Build GRCh38
rs370106509 Chr.12: 193373-193373 on Build GRCh38
rs116264295 Chr.12: 193599-193599 on Build GRCh38
rs769540976 Chr.12: 193226-193226 on Build GRCh38
rs773887396 Chr.12: 193331-193331 on Build GRCh38
rs114829305 Chr.12: 193470-193470 on Build GRCh38
rs959094510 Chr.12: 193368-193368 on Build GRCh38
rs199744288 Chr.12: 193276-193276 on Build GRCh38
rs933370869 Chr.12: 193476-193476 on Build GRCh38
rs138663159 Chr.12: 193369-193369 on Build GRCh38
rs773797327 Chr.12: 193399-193399 on Build GRCh38
rs1033323608 Chr.12: 193586-193586 on Build GRCh38
rs572885121 Chr.12: 193509-193509 on Build GRCh38
rs780645855 Chr.12: 193298-193298 on Build GRCh38
rs772925314 Chr.12: 193236-193236 on Build GRCh38
rs767347103 Chr.12: 193256-193256 on Build GRCh38
rs540268697 Chr.12: 193520-193520 on Build GRCh38
rs936618839 Chr.12: 193432-193432 on Build GRCh38
rs750575623 Chr.12: 193357-193357 on Build GRCh38
rs915995626 Chr.12: 193496-193496 on Build GRCh38
rs765436008 Chr.12: 193274-193274 on Build GRCh38
rs142742144 Chr.12: 193498-193498 on Build GRCh38
rs766419258 Chr.12: 193365-193365 on Build GRCh38
rs982750331 Chr.12: 193588-193588 on Build GRCh38
rs774727179 Chr.12: 193235-193236 on Build GRCh38
rs762312153 Chr.12: 193242-193242 on Build GRCh38
rs760274126 Chr.12: 193632-193632 on Build GRCh38
rs75572113 Chr.12: 193320-193320 on Build GRCh38
rs150581150 Chr.12: 193362-193362 on Build GRCh38
rs759393815 Chr.12: 193255-193255 on Build GRCh38
rs1051863134 Chr.12: 193480-193480 on Build GRCh38
rs766344624 Chr.12: 193243-193243 on Build GRCh38
rs748526957 Chr.12: 193378-193378 on Build GRCh38
rs371844842 Chr.12: 193625-193625 on Build GRCh38
rs771550500 Chr.12: 193592-193592 on Build GRCh38
rs941347981 Chr.12: 193468-193468 on Build GRCh38
rs962782232 Chr.12: 193598-193598 on Build GRCh38
rs755610765 Chr.12: 193372-193372 on Build GRCh38
rs770583274 Chr.12: 193393-193393 on Build GRCh38
rs1010978854 Chr.12: 193550-193550 on Build GRCh38
rs983083155 Chr.12: 193434-193434 on Build GRCh38
rs915881093 Chr.12: 193604-193606 on Build GRCh38
rs892159168 Chr.12: 193521-193521 on Build GRCh38
rs941474910 Chr.12: 193265-193265 on Build GRCh38
rs757538971 Chr.12: 193261-193261 on Build GRCh38
rs1015920636 Chr.12: 193591-193591 on Build GRCh38
rs544161209 Chr.12: 193584-193584 on Build GRCh38
rs974696858 Chr.12: 193433-193433 on Build GRCh38
rs770296734 Chr.12: 193330-193330 on Build GRCh38
rs1025201483 Chr.12: 193391-193391 on Build GRCh38

View Full Product Details