Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 1786278-1786787 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADIPOR2
Entrez Gene ID
79602
Gene Name
adiponectin receptor 2
Gene Aliases
ACDCR2, PAQR2
UniGene
Hs.371642
Gene Chromosome Location
Chr.12: 1690981-1788679 on Build GRCh38
Transcripts
NM_024551.2 XM_005253789.1 XM_006719018.2 XM_011521024.2

SNP Detail

SNP ID Chromosome Location
rs768640748 Chr.12: 1786707-1786707 on Build GRCh38
rs533077487 Chr.12: 1786447-1786447 on Build GRCh38
rs1046075113 Chr.12: 1786742-1786742 on Build GRCh38
rs936305835 Chr.12: 1786636-1786636 on Build GRCh38
rs1049447074 Chr.12: 1786656-1786656 on Build GRCh38
rs1057001164 Chr.12: 1786486-1786486 on Build GRCh38
rs1028827271 Chr.12: 1786489-1786489 on Build GRCh38
rs187521934 Chr.12: 1786398-1786398 on Build GRCh38
rs1026337775 Chr.12: 1786755-1786755 on Build GRCh38
rs1013219475 Chr.12: 1786346-1786346 on Build GRCh38
rs920059074 Chr.12: 1786660-1786660 on Build GRCh38
rs1028233061 Chr.12: 1786306-1786306 on Build GRCh38
rs774999225 Chr.12: 1786438-1786441 on Build GRCh38
rs1055306181 Chr.12: 1786705-1786705 on Build GRCh38
rs368981439 Chr.12: 1786421-1786421 on Build GRCh38
rs1055378806 Chr.12: 1786312-1786312 on Build GRCh38
rs551213793 Chr.12: 1786449-1786449 on Build GRCh38
rs562864824 Chr.12: 1786443-1786443 on Build GRCh38
rs974641907 Chr.12: 1786474-1786474 on Build GRCh38
rs756977827 Chr.12: 1786455-1786455 on Build GRCh38
rs940058475 Chr.12: 1786597-1786597 on Build GRCh38
rs762405880 Chr.12: 1786598-1786599 on Build GRCh38
rs1008468368 Chr.12: 1786358-1786358 on Build GRCh38
rs931472925 Chr.12: 1786673-1786673 on Build GRCh38
rs566339084 Chr.12: 1786466-1786466 on Build GRCh38
rs903635954 Chr.12: 1786635-1786635 on Build GRCh38
rs555902485 Chr.12: 1786634-1786634 on Build GRCh38
rs371544429 Chr.12: 1786571-1786571 on Build GRCh38
rs889665445 Chr.12: 1786659-1786660 on Build GRCh38
rs912579333 Chr.12: 1786589-1786589 on Build GRCh38
rs763649829 Chr.12: 1786365-1786365 on Build GRCh38
rs188309120 Chr.12: 1786568-1786568 on Build GRCh38
rs112653490 Chr.12: 1786490-1786490 on Build GRCh38
rs751480856 Chr.12: 1786693-1786693 on Build GRCh38
rs1008146829 Chr.12: 1786672-1786672 on Build GRCh38
rs549156055 Chr.12: 1786496-1786496 on Build GRCh38
rs550730419 Chr.12: 1786402-1786402 on Build GRCh38
rs192753878 Chr.12: 1786425-1786425 on Build GRCh38
rs949630058 Chr.12: 1786735-1786735 on Build GRCh38
rs984779341 Chr.12: 1786430-1786430 on Build GRCh38
rs771232113 Chr.12: 1786391-1786391 on Build GRCh38
rs896998344 Chr.12: 1786718-1786718 on Build GRCh38
rs567278877 Chr.12: 1786535-1786535 on Build GRCh38
rs1019449252 Chr.12: 1786369-1786369 on Build GRCh38
rs895366485 Chr.12: 1786706-1786706 on Build GRCh38
rs1024654395 Chr.12: 1786353-1786353 on Build GRCh38
rs895441535 Chr.12: 1786345-1786345 on Build GRCh38
rs986732637 Chr.12: 1786550-1786550 on Build GRCh38
rs575453647 Chr.12: 1786403-1786403 on Build GRCh38

View Full Product Details