Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 10386805-10387305 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLRC4-KLRK1
Entrez Gene ID
100528032
Gene Name
KLRC4-KLRK1 readthrough
Gene Aliases
-
UniGene
Hs.387787
Gene Chromosome Location
Chr.12: 10372353-10410146 on Build GRCh38
Transcripts
NM_001199805.1 NM_007360.3 NR_120430.1

Gene Symbol
KLRK1
Entrez Gene ID
22914
Gene Name
killer cell lectin like receptor K1
Gene Aliases
CD314, D12S2489E, KLR, NKG2-D, NKG2D
UniGene
Hs.387787
Gene Chromosome Location
Chr.12: 10372353-10390054 on Build GRCh38
Transcripts
NM_001199805.1 NM_007360.3 NR_120430.1

Gene Symbol
LOC101928100
Entrez Gene ID
101928100
Gene Name
uncharacterized LOC101928100
Gene Aliases
-
UniGene
Hs.638425
Gene Chromosome Location
Chr.12: 10363769-10398506 on Build GRCh38
Transcripts
NM_001199805.1 NM_007360.3 NR_120430.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
275780 c.88T>G p.F30V Chr.12: 10386963-10386963 on Build GRCh38
3398279 c.97C>T p.R33* Chr.12: 10386954-10386954 on Build GRCh38
2064753 c.95C>T p.T32I Chr.12: 10386956-10386956 on Build GRCh38
1161028 c.41A>G p.E14G Chr.12: 10387010-10387010 on Build GRCh38
3986582 c.83G>A p.S28N Chr.12: 10386968-10386968 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530301513 Chr.12: 10386941-10386941 on Build GRCh38
rs371740966 Chr.12: 10386859-10386859 on Build GRCh38
rs565582185 Chr.12: 10387165-10387165 on Build GRCh38
rs988743295 Chr.12: 10386844-10386845 on Build GRCh38
rs199875572 Chr.12: 10386957-10386957 on Build GRCh38
rs763200807 Chr.12: 10386931-10386931 on Build GRCh38
rs777778235 Chr.12: 10386892-10386892 on Build GRCh38
rs913121913 Chr.12: 10387220-10387220 on Build GRCh38
rs771963702 Chr.12: 10386888-10386888 on Build GRCh38
rs1008917191 Chr.12: 10387110-10387110 on Build GRCh38
rs754479520 Chr.12: 10386876-10386876 on Build GRCh38
rs865809094 Chr.12: 10387125-10387125 on Build GRCh38
rs774470337 Chr.12: 10387030-10387030 on Build GRCh38
rs762093439 Chr.12: 10386942-10386942 on Build GRCh38
rs757031790 Chr.12: 10386988-10386988 on Build GRCh38
rs376353454 Chr.12: 10387038-10387038 on Build GRCh38
rs528583023 Chr.12: 10386971-10386971 on Build GRCh38
rs376040069 Chr.12: 10386904-10386904 on Build GRCh38
rs978582761 Chr.12: 10387269-10387269 on Build GRCh38
rs138926336 Chr.12: 10387009-10387009 on Build GRCh38
rs916153326 Chr.12: 10387264-10387264 on Build GRCh38
rs373872367 Chr.12: 10387263-10387263 on Build GRCh38
rs957559004 Chr.12: 10387244-10387244 on Build GRCh38
rs768803530 Chr.12: 10387024-10387024 on Build GRCh38
rs967460848 Chr.12: 10387166-10387166 on Build GRCh38
rs754491286 Chr.12: 10386956-10386956 on Build GRCh38
rs750451773 Chr.12: 10386872-10386872 on Build GRCh38
rs532469753 Chr.12: 10387096-10387096 on Build GRCh38
rs746907937 Chr.12: 10386899-10386899 on Build GRCh38
rs770843020 Chr.12: 10386902-10386902 on Build GRCh38
rs2733845 Chr.12: 10387216-10387216 on Build GRCh38
rs779820846 Chr.12: 10386983-10386984 on Build GRCh38
rs781566516 Chr.12: 10386875-10386875 on Build GRCh38
rs758183711 Chr.12: 10386964-10386964 on Build GRCh38
rs745839125 Chr.12: 10386995-10386995 on Build GRCh38
rs752026398 Chr.12: 10386860-10386860 on Build GRCh38
rs998908082 Chr.12: 10387168-10387168 on Build GRCh38
rs764433611 Chr.12: 10386937-10386937 on Build GRCh38
rs751735555 Chr.12: 10387229-10387229 on Build GRCh38
rs552247561 Chr.12: 10387121-10387121 on Build GRCh38
rs947230028 Chr.12: 10386878-10386878 on Build GRCh38
rs747854009 Chr.12: 10386886-10386886 on Build GRCh38
rs576110591 Chr.12: 10386846-10386846 on Build GRCh38
rs750918291 Chr.12: 10386953-10386953 on Build GRCh38
rs775663961 Chr.12: 10386927-10386927 on Build GRCh38
rs1018935406 Chr.12: 10387112-10387112 on Build GRCh38
rs991748112 Chr.12: 10386841-10386841 on Build GRCh38
rs759618556 Chr.12: 10386909-10386909 on Build GRCh38
rs145126988 Chr.12: 10386975-10386975 on Build GRCh38
rs563491525 Chr.12: 10387007-10387007 on Build GRCh38
rs769830158 Chr.12: 10386996-10386996 on Build GRCh38
rs550164323 Chr.12: 10386950-10386950 on Build GRCh38
rs1043296968 Chr.12: 10386913-10386913 on Build GRCh38
rs758690645 Chr.12: 10386955-10386955 on Build GRCh38
rs947729056 Chr.12: 10386871-10386871 on Build GRCh38
rs781134649 Chr.12: 10386990-10386990 on Build GRCh38

View Full Product Details