Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 10809389-10809884 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAS2R9
Entrez Gene ID
50835
Gene Name
taste 2 receptor member 9
Gene Aliases
T2R9, TRB6
UniGene
Hs.272391
Gene Chromosome Location
Chr.12: 10809094-10810168 on Build GRCh38
Transcripts
NM_023917.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4732236 c.387C>A p.V129V Chr.12: 10809689-10809689 on Build GRCh38
1163643 c.617_620delTTAG p.V206fs*22 Chr.12: 10809456-10809459 on Build GRCh38
3455753 c.547C>T p.L183L Chr.12: 10809529-10809529 on Build GRCh38
4770142 c.560T>C p.V187A Chr.12: 10809516-10809516 on Build GRCh38
7393078 c.455T>A p.M152K Chr.12: 10809621-10809621 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753971046 Chr.12: 10809623-10809625 on Build GRCh38
rs750159262 Chr.12: 10809810-10809810 on Build GRCh38
rs751159405 Chr.12: 10809797-10809797 on Build GRCh38
rs756156449 Chr.12: 10809814-10809814 on Build GRCh38
rs780998989 Chr.12: 10809732-10809732 on Build GRCh38
rs371157182 Chr.12: 10809831-10809831 on Build GRCh38
rs115829220 Chr.12: 10809688-10809688 on Build GRCh38
rs115762530 Chr.12: 10809507-10809507 on Build GRCh38
rs780099344 Chr.12: 10809825-10809825 on Build GRCh38
rs538294164 Chr.12: 10809500-10809500 on Build GRCh38
rs1054645251 Chr.12: 10809622-10809622 on Build GRCh38
rs11054043 Chr.12: 10809568-10809568 on Build GRCh38
rs113883583 Chr.12: 10809423-10809423 on Build GRCh38
rs1032122504 Chr.12: 10809493-10809493 on Build GRCh38
rs1000750425 Chr.12: 10809761-10809761 on Build GRCh38
rs763752516 Chr.12: 10809791-10809791 on Build GRCh38
rs374671945 Chr.12: 10809744-10809744 on Build GRCh38
rs762457178 Chr.12: 10809859-10809859 on Build GRCh38
rs777068115 Chr.12: 10809853-10809853 on Build GRCh38
rs766439744 Chr.12: 10809421-10809421 on Build GRCh38
rs74954631 Chr.12: 10809626-10809626 on Build GRCh38
rs761294086 Chr.12: 10809770-10809770 on Build GRCh38
rs11054044 Chr.12: 10809765-10809765 on Build GRCh38
rs766840521 Chr.12: 10809772-10809772 on Build GRCh38
rs753145035 Chr.12: 10809437-10809437 on Build GRCh38
rs754460148 Chr.12: 10809575-10809575 on Build GRCh38
rs749347848 Chr.12: 10809756-10809756 on Build GRCh38
rs200345708 Chr.12: 10809519-10809519 on Build GRCh38
rs750882695 Chr.12: 10809680-10809680 on Build GRCh38
rs149488505 Chr.12: 10809541-10809541 on Build GRCh38
rs778818765 Chr.12: 10809801-10809802 on Build GRCh38
rs779004980 Chr.12: 10809832-10809832 on Build GRCh38
rs79661572 Chr.12: 10809695-10809695 on Build GRCh38
rs758789858 Chr.12: 10809439-10809439 on Build GRCh38
rs3741845 Chr.12: 10809516-10809516 on Build GRCh38
rs747865458 Chr.12: 10809581-10809581 on Build GRCh38
rs769840781 Chr.12: 10809734-10809734 on Build GRCh38
rs767994640 Chr.12: 10809766-10809766 on Build GRCh38
rs959236908 Chr.12: 10809808-10809808 on Build GRCh38
rs114474097 Chr.12: 10809644-10809644 on Build GRCh38
rs778435659 Chr.12: 10809460-10809460 on Build GRCh38
rs777597147 Chr.12: 10809600-10809600 on Build GRCh38
rs771455037 Chr.12: 10809852-10809852 on Build GRCh38
rs768230107 Chr.12: 10809497-10809497 on Build GRCh38
rs765468339 Chr.12: 10809435-10809435 on Build GRCh38
rs764789134 Chr.12: 10809687-10809687 on Build GRCh38
rs757858202 Chr.12: 10809787-10809787 on Build GRCh38
rs752911951 Chr.12: 10809523-10809523 on Build GRCh38
rs144196094 Chr.12: 10809640-10809640 on Build GRCh38
rs763991481 Chr.12: 10809593-10809593 on Build GRCh38
rs147406764 Chr.12: 10809511-10809511 on Build GRCh38
rs751959302 Chr.12: 10809539-10809539 on Build GRCh38
rs372814318 Chr.12: 10809464-10809464 on Build GRCh38
rs759726680 Chr.12: 10809432-10809432 on Build GRCh38
rs767929629 Chr.12: 10809678-10809678 on Build GRCh38
rs764618198 Chr.12: 10809536-10809536 on Build GRCh38
rs370599187 Chr.12: 10809860-10809860 on Build GRCh38
rs376745268 Chr.12: 10809839-10809839 on Build GRCh38
rs909092684 Chr.12: 10809508-10809508 on Build GRCh38
rs773439257 Chr.12: 10809769-10809769 on Build GRCh38
rs776614425 Chr.12: 10809517-10809517 on Build GRCh38
rs756576629 Chr.12: 10809681-10809681 on Build GRCh38
rs377101850 Chr.12: 10809743-10809743 on Build GRCh38
rs779488054 Chr.12: 10809472-10809472 on Build GRCh38
rs200091906 Chr.12: 10809619-10809619 on Build GRCh38
rs565155192 Chr.12: 10809733-10809733 on Build GRCh38
rs781699584 Chr.12: 10809542-10809542 on Build GRCh38
rs746803543 Chr.12: 10809726-10809726 on Build GRCh38
rs966596947 Chr.12: 10809473-10809473 on Build GRCh38
rs996625067 Chr.12: 10809422-10809422 on Build GRCh38
rs777364596 Chr.12: 10809719-10809719 on Build GRCh38
rs763412285 Chr.12: 10809534-10809534 on Build GRCh38
rs764389078 Chr.12: 10809641-10809641 on Build GRCh38
rs752239256 Chr.12: 10809781-10809781 on Build GRCh38
rs955072564 Chr.12: 10809429-10809429 on Build GRCh38
rs143855266 Chr.12: 10809731-10809731 on Build GRCh38
rs1051580974 Chr.12: 10809563-10809563 on Build GRCh38
rs749400892 Chr.12: 10809826-10809826 on Build GRCh38
rs762194733 Chr.12: 10809654-10809654 on Build GRCh38
rs757777760 Chr.12: 10809471-10809471 on Build GRCh38
rs756113122 Chr.12: 10809530-10809532 on Build GRCh38
rs1013157279 Chr.12: 10809625-10809625 on Build GRCh38
rs768796979 Chr.12: 10809827-10809827 on Build GRCh38
rs760751810 Chr.12: 10809411-10809411 on Build GRCh38
rs756853761 Chr.12: 10809800-10809800 on Build GRCh38
rs746788217 Chr.12: 10809609-10809609 on Build GRCh38
rs762745677 Chr.12: 10809757-10809757 on Build GRCh38
rs757384793 Chr.12: 10809686-10809686 on Build GRCh38
rs934750872 Chr.12: 10809847-10809847 on Build GRCh38
rs148917754 Chr.12: 10809486-10809486 on Build GRCh38
rs775706163 Chr.12: 10809628-10809628 on Build GRCh38
rs779263593 Chr.12: 10809833-10809833 on Build GRCh38
rs367711180 Chr.12: 10809802-10809802 on Build GRCh38
rs370192026 Chr.12: 10809565-10809565 on Build GRCh38
rs898279087 Chr.12: 10809413-10809413 on Build GRCh38
rs572211040 Chr.12: 10809838-10809838 on Build GRCh38
rs990664277 Chr.12: 10809848-10809848 on Build GRCh38
rs778493402 Chr.12: 10809499-10809499 on Build GRCh38
rs142881666 Chr.12: 10809438-10809438 on Build GRCh38
rs942248847 Chr.12: 10809621-10809621 on Build GRCh38
rs754058874 Chr.12: 10809614-10809614 on Build GRCh38
rs774580531 Chr.12: 10809763-10809763 on Build GRCh38
rs771232686 Chr.12: 10809530-10809530 on Build GRCh38
rs758035979 Chr.12: 10809594-10809594 on Build GRCh38
rs778544213 Chr.12: 10809579-10809579 on Build GRCh38
rs771790416 Chr.12: 10809501-10809501 on Build GRCh38
rs750663977 Chr.12: 10809852-10809852 on Build GRCh38

View Full Product Details