Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 12822635-12823180 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX47
Entrez Gene ID
51202
Gene Name
DEAD-box helicase 47
Gene Aliases
E4-DBP, HQ0256, MSTP162, RRP3
UniGene
Hs.719938
Gene Chromosome Location
Chr.12: 12813346-12829981 on Build GRCh38
Transcripts
NM_016355.3 NM_201224.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6348291 c.562-1G>T p.? Chr.12: 12822660-12822660 on Build GRCh38
2228073 c.590G>A p.R197Q Chr.12: 12822689-12822689 on Build GRCh38
936938 c.611T>C p.F204S Chr.12: 12822710-12822710 on Build GRCh38
936937 c.589C>T p.R197* Chr.12: 12822688-12822688 on Build GRCh38
5500532 c.633+5A>G p.? Chr.12: 12822737-12822737 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371332178 Chr.12: 12822747-12822747 on Build GRCh38
rs894920935 Chr.12: 12822906-12822906 on Build GRCh38
rs943440207 Chr.12: 12822981-12822981 on Build GRCh38
rs984822171 Chr.12: 12823085-12823085 on Build GRCh38
rs145316378 Chr.12: 12822727-12822727 on Build GRCh38
rs147868527 Chr.12: 12822695-12822695 on Build GRCh38
rs975206544 Chr.12: 12823114-12823114 on Build GRCh38
rs757844427 Chr.12: 12822779-12822779 on Build GRCh38
rs532427679 Chr.12: 12822800-12822800 on Build GRCh38
rs1001679168 Chr.12: 12823041-12823041 on Build GRCh38
rs548716952 Chr.12: 12822933-12822933 on Build GRCh38
rs1013860421 Chr.12: 12823068-12823068 on Build GRCh38
rs774098537 Chr.12: 12822694-12822694 on Build GRCh38
rs544125905 Chr.12: 12822823-12822823 on Build GRCh38
rs961019789 Chr.12: 12822839-12822839 on Build GRCh38
rs752834943 Chr.12: 12822736-12822736 on Build GRCh38
rs773264856 Chr.12: 12822699-12822699 on Build GRCh38
rs112715839 Chr.12: 12822935-12822935 on Build GRCh38
rs1026870061 Chr.12: 12822977-12822977 on Build GRCh38
rs35296634 Chr.12: 12822797-12822797 on Build GRCh38
rs141452017 Chr.12: 12822708-12822708 on Build GRCh38
rs552506031 Chr.12: 12823056-12823056 on Build GRCh38
rs950700026 Chr.12: 12823074-12823074 on Build GRCh38
rs888266721 Chr.12: 12822998-12822998 on Build GRCh38
rs527689954 Chr.12: 12823008-12823008 on Build GRCh38
rs1046030510 Chr.12: 12822925-12822925 on Build GRCh38
rs992801738 Chr.12: 12822863-12822863 on Build GRCh38
rs189935414 Chr.12: 12822871-12822871 on Build GRCh38
rs998686157 Chr.12: 12823061-12823061 on Build GRCh38
rs759463536 Chr.12: 12822827-12822827 on Build GRCh38
rs773230731 Chr.12: 12822826-12822826 on Build GRCh38
rs372689788 Chr.12: 12822688-12822688 on Build GRCh38
rs1033876603 Chr.12: 12823043-12823043 on Build GRCh38
rs767594526 Chr.12: 12822934-12822934 on Build GRCh38
rs905647335 Chr.12: 12823036-12823036 on Build GRCh38
rs994020012 Chr.12: 12822965-12822965 on Build GRCh38
rs1038896255 Chr.12: 12822987-12822987 on Build GRCh38
rs776606946 Chr.12: 12822668-12822668 on Build GRCh38
rs7313699 Chr.12: 12822908-12822908 on Build GRCh38
rs933268182 Chr.12: 12823015-12823015 on Build GRCh38
rs779747453 Chr.12: 12823112-12823112 on Build GRCh38
rs199670577 Chr.12: 12822744-12822744 on Build GRCh38
rs1026536279 Chr.12: 12823073-12823073 on Build GRCh38
rs758868299 Chr.12: 12822745-12822745 on Build GRCh38
rs779726921 Chr.12: 12822670-12822670 on Build GRCh38
rs201398006 Chr.12: 12822731-12822731 on Build GRCh38
rs1054870307 Chr.12: 12822883-12822883 on Build GRCh38
rs374347218 Chr.12: 12822765-12822765 on Build GRCh38
rs748094560 Chr.12: 12822723-12822723 on Build GRCh38
rs879782637 Chr.12: 12822866-12822866 on Build GRCh38
rs1011156778 Chr.12: 12823032-12823032 on Build GRCh38
rs766243168 Chr.12: 12822721-12822721 on Build GRCh38
rs566944356 Chr.12: 12822983-12822983 on Build GRCh38
rs1042702948 Chr.12: 12823033-12823033 on Build GRCh38
rs369431237 Chr.12: 12822661-12822661 on Build GRCh38
rs937508890 Chr.12: 12822882-12822882 on Build GRCh38
rs541188537 Chr.12: 12823045-12823045 on Build GRCh38
rs565041433 Chr.12: 12822748-12822748 on Build GRCh38
rs891469833 Chr.12: 12823022-12823022 on Build GRCh38
rs778387970 Chr.12: 12822662-12822662 on Build GRCh38
rs761143937 Chr.12: 12823155-12823158 on Build GRCh38
rs769565762 Chr.12: 12822793-12822793 on Build GRCh38
rs967390622 Chr.12: 12823086-12823086 on Build GRCh38
rs1052936577 Chr.12: 12823019-12823019 on Build GRCh38
rs909197971 Chr.12: 12823092-12823092 on Build GRCh38
rs540178927 Chr.12: 12822692-12822692 on Build GRCh38
rs978982784 Chr.12: 12823105-12823105 on Build GRCh38

View Full Product Details