Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 30639323-30639819 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IPO8
Entrez Gene ID
10526
Gene Name
importin 8
Gene Aliases
RANBP8
UniGene
Hs.505136
Gene Chromosome Location
Chr.12: 30628981-30695995 on Build GRCh38
Transcripts
NM_001190995.1 NM_006390.3 XM_017018691.1 XM_017018692.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
938628 c.2422C>T p.H808Y Chr.12: 30639582-30639582 on Build GRCh38
1210830 c.2311C>T p.R771* Chr.12: 30639693-30639693 on Build GRCh38
4413446 c.2472T>G p.D824E Chr.12: 30639532-30639532 on Build GRCh38
1705349 c.2312G>A p.R771Q Chr.12: 30639692-30639692 on Build GRCh38
938630 c.2314G>T p.G772W Chr.12: 30639690-30639690 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764351621 Chr.12: 30639544-30639544 on Build GRCh38
rs919021606 Chr.12: 30639372-30639372 on Build GRCh38
rs893694946 Chr.12: 30639653-30639653 on Build GRCh38
rs1056730820 Chr.12: 30639606-30639606 on Build GRCh38
rs371183250 Chr.12: 30639628-30639628 on Build GRCh38
rs775413184 Chr.12: 30639693-30639693 on Build GRCh38
rs777207997 Chr.12: 30639492-30639492 on Build GRCh38
rs554370953 Chr.12: 30639479-30639479 on Build GRCh38
rs760106437 Chr.12: 30639493-30639493 on Build GRCh38
rs959568840 Chr.12: 30639793-30639793 on Build GRCh38
rs759367696 Chr.12: 30639746-30639746 on Build GRCh38
rs777431327 Chr.12: 30639774-30639774 on Build GRCh38
rs997460109 Chr.12: 30639419-30639419 on Build GRCh38
rs747292279 Chr.12: 30639511-30639511 on Build GRCh38
rs770376784 Chr.12: 30639692-30639692 on Build GRCh38
rs757911609 Chr.12: 30639768-30639768 on Build GRCh38
rs753670161 Chr.12: 30639587-30639587 on Build GRCh38
rs200975958 Chr.12: 30639524-30639524 on Build GRCh38
rs778254539 Chr.12: 30639596-30639596 on Build GRCh38
rs1010935294 Chr.12: 30639668-30639668 on Build GRCh38
rs781710025 Chr.12: 30639631-30639631 on Build GRCh38
rs766121963 Chr.12: 30639725-30639725 on Build GRCh38
rs775674328 Chr.12: 30639523-30639523 on Build GRCh38
rs1029212454 Chr.12: 30639428-30639429 on Build GRCh38
rs973468247 Chr.12: 30639353-30639353 on Build GRCh38
rs556566621 Chr.12: 30639763-30639763 on Build GRCh38
rs775937664 Chr.12: 30639725-30639725 on Build GRCh38
rs753706211 Chr.12: 30639744-30639744 on Build GRCh38
rs939575856 Chr.12: 30639572-30639572 on Build GRCh38
rs752299664 Chr.12: 30639597-30639597 on Build GRCh38
rs141553474 Chr.12: 30639619-30639619 on Build GRCh38
rs754973897 Chr.12: 30639594-30639594 on Build GRCh38
rs1051547042 Chr.12: 30639495-30639495 on Build GRCh38
rs373797969 Chr.12: 30639561-30639561 on Build GRCh38
rs759043684 Chr.12: 30639789-30639789 on Build GRCh38
rs750206949 Chr.12: 30639550-30639550 on Build GRCh38
rs771358885 Chr.12: 30639626-30639626 on Build GRCh38
rs538132238 Chr.12: 30639518-30639518 on Build GRCh38
rs768862935 Chr.12: 30639717-30639717 on Build GRCh38
rs757971515 Chr.12: 30639608-30639608 on Build GRCh38
rs765275572 Chr.12: 30639747-30639747 on Build GRCh38
rs967029401 Chr.12: 30639538-30639538 on Build GRCh38
rs916701937 Chr.12: 30639609-30639609 on Build GRCh38
rs12316608 Chr.12: 30639736-30639736 on Build GRCh38
rs201728857 Chr.12: 30639637-30639637 on Build GRCh38
rs887890019 Chr.12: 30639687-30639687 on Build GRCh38
rs372979657 Chr.12: 30639778-30639778 on Build GRCh38
rs752141940 Chr.12: 30639760-30639760 on Build GRCh38
rs755971731 Chr.12: 30639564-30639564 on Build GRCh38
rs766167102 Chr.12: 30639571-30639571 on Build GRCh38
rs781382537 Chr.12: 30639783-30639783 on Build GRCh38
rs772180787 Chr.12: 30639665-30639666 on Build GRCh38
rs763153347 Chr.12: 30639711-30639711 on Build GRCh38
rs376752588 Chr.12: 30639610-30639610 on Build GRCh38
rs757004704 Chr.12: 30639780-30639780 on Build GRCh38
rs765998115 Chr.12: 30639664-30639664 on Build GRCh38
rs774490198 Chr.12: 30639720-30639720 on Build GRCh38
rs762035678 Chr.12: 30639721-30639721 on Build GRCh38
rs1042388757 Chr.12: 30639669-30639669 on Build GRCh38

View Full Product Details