Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 47734420-47734929 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC105369748
Entrez Gene ID
105369748
Gene Name
uncharacterized LOC105369748
Gene Aliases
-
UniGene
Hs.722382
Gene Chromosome Location
Chr.12: 47706059-47740134 on Build GRCh38
Transcripts
NM_001098531.2 NM_001098532.2 NM_006105.5

Gene Symbol
RAPGEF3
Entrez Gene ID
10411
Gene Name
Rap guanine nucleotide exchange factor 3
Gene Aliases
CAMP-GEFI, EPAC, EPAC1, HSU79275, bcm910
UniGene
Hs.8578
Gene Chromosome Location
Chr.12: 47734670-47759106 on Build GRCh38
Transcripts
NM_001098531.2 NM_001098532.2 NM_006105.5

SNP Detail

SNP ID Chromosome Location
rs555332927 Chr.12: 47734674-47734674 on Build GRCh38
rs983680664 Chr.12: 47734719-47734719 on Build GRCh38
rs1008418928 Chr.12: 47734709-47734709 on Build GRCh38
rs923598548 Chr.12: 47734824-47734824 on Build GRCh38
rs140433170 Chr.12: 47734540-47734540 on Build GRCh38
rs916259249 Chr.12: 47734462-47734462 on Build GRCh38
rs960830855 Chr.12: 47734728-47734728 on Build GRCh38
rs7133585 Chr.12: 47734627-47734627 on Build GRCh38
rs992680762 Chr.12: 47734898-47734898 on Build GRCh38
rs540314957 Chr.12: 47734712-47734712 on Build GRCh38
rs900579753 Chr.12: 47734886-47734886 on Build GRCh38
rs968298907 Chr.12: 47734838-47734838 on Build GRCh38
rs10875683 Chr.12: 47734500-47734500 on Build GRCh38
rs144281586 Chr.12: 47734905-47734905 on Build GRCh38
rs1017673401 Chr.12: 47734590-47734590 on Build GRCh38
rs569542425 Chr.12: 47734543-47734543 on Build GRCh38
rs2240080 Chr.12: 47734721-47734721 on Build GRCh38
rs577274257 Chr.12: 47734723-47734723 on Build GRCh38
rs1009003861 Chr.12: 47734557-47734557 on Build GRCh38
rs757283 Chr.12: 47734761-47734761 on Build GRCh38
rs946739668 Chr.12: 47734453-47734453 on Build GRCh38
rs554712601 Chr.12: 47734550-47734550 on Build GRCh38
rs189313028 Chr.12: 47734901-47734901 on Build GRCh38
rs938006956 Chr.12: 47734867-47734867 on Build GRCh38
rs1039175169 Chr.12: 47734878-47734879 on Build GRCh38
rs1056527954 Chr.12: 47734522-47734522 on Build GRCh38
rs926627953 Chr.12: 47734858-47734858 on Build GRCh38
rs184556482 Chr.12: 47734726-47734726 on Build GRCh38
rs796740743 Chr.12: 47734472-47734472 on Build GRCh38
rs1054776049 Chr.12: 47734454-47734454 on Build GRCh38
rs755913698 Chr.12: 47734631-47734631 on Build GRCh38
rs1021094989 Chr.12: 47734773-47734773 on Build GRCh38
rs369357072 Chr.12: 47734862-47734862 on Build GRCh38
rs558921953 Chr.12: 47734632-47734632 on Build GRCh38
rs73302662 Chr.12: 47734906-47734906 on Build GRCh38
rs574767399 Chr.12: 47734697-47734697 on Build GRCh38
rs889428312 Chr.12: 47734682-47734682 on Build GRCh38
rs979652143 Chr.12: 47734848-47734848 on Build GRCh38
rs1051000377 Chr.12: 47734840-47734840 on Build GRCh38
rs964729389 Chr.12: 47734615-47734615 on Build GRCh38
rs1043749780 Chr.12: 47734498-47734498 on Build GRCh38
rs979502153 Chr.12: 47734796-47734796 on Build GRCh38
rs988324507 Chr.12: 47734763-47734763 on Build GRCh38
rs150874763 Chr.12: 47734521-47734521 on Build GRCh38
rs999401501 Chr.12: 47734523-47734523 on Build GRCh38
rs890977902 Chr.12: 47734847-47734847 on Build GRCh38
rs564717233 Chr.12: 47734732-47734732 on Build GRCh38

View Full Product Details