Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49633340-49633855 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRPF40B
Entrez Gene ID
25766
Gene Name
pre-mRNA processing factor 40 homolog B
Gene Aliases
HYPC
UniGene
Hs.706827
Gene Chromosome Location
Chr.12: 49622717-49644669 on Build GRCh38
Transcripts
NM_001031698.2 NM_012272.2 XM_006719324.3 XM_006719325.3 XM_011538135.2 XM_011538136.2 XM_011538137.2 XM_011538138.2 XM_011538139.2 XM_011538140.2 XM_011538141.1 XM_011538143.2 XM_011538144.2 XM_017019135.1 XM_017019136.1 XM_017019137.1 XM_017019138.1 XM_017019139.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
940183 c.410G>A p.C137Y Chr.12: 49633443-49633443 on Build GRCh38
5770851 c.511G>C p.E171Q Chr.12: 49633544-49633544 on Build GRCh38
51832 c.502G>A p.D168N Chr.12: 49633535-49633535 on Build GRCh38
1362140 c.503A>C p.D168A Chr.12: 49633536-49633536 on Build GRCh38
3931644 c.406C>T p.Q136* Chr.12: 49633439-49633439 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377383072 Chr.12: 49633699-49633699 on Build GRCh38
rs116571114 Chr.12: 49633460-49633460 on Build GRCh38
rs753773941 Chr.12: 49633704-49633704 on Build GRCh38
rs369667514 Chr.12: 49633566-49633566 on Build GRCh38
rs772769626 Chr.12: 49633599-49633599 on Build GRCh38
rs528349627 Chr.12: 49633525-49633525 on Build GRCh38
rs1042870051 Chr.12: 49633384-49633384 on Build GRCh38
rs549784548 Chr.12: 49633583-49633583 on Build GRCh38
rs755877223 Chr.12: 49633448-49633448 on Build GRCh38
rs774623847 Chr.12: 49633588-49633588 on Build GRCh38
rs115525092 Chr.12: 49633519-49633519 on Build GRCh38
rs932337154 Chr.12: 49633396-49633396 on Build GRCh38
rs751621195 Chr.12: 49633554-49633554 on Build GRCh38
rs760274324 Chr.12: 49633639-49633639 on Build GRCh38
rs996470979 Chr.12: 49633789-49633789 on Build GRCh38
rs753372496 Chr.12: 49633531-49633531 on Build GRCh38
rs774118549 Chr.12: 49633615-49633616 on Build GRCh38
rs754823653 Chr.12: 49633534-49633534 on Build GRCh38
rs752547654 Chr.12: 49633692-49633692 on Build GRCh38
rs779736493 Chr.12: 49633580-49633580 on Build GRCh38
rs766114078 Chr.12: 49633450-49633450 on Build GRCh38
rs972518623 Chr.12: 49633702-49633702 on Build GRCh38
rs115693590 Chr.12: 49633488-49633488 on Build GRCh38
rs570815870 Chr.12: 49633720-49633721 on Build GRCh38
rs780594636 Chr.12: 49633511-49633511 on Build GRCh38
rs762733994 Chr.12: 49633695-49633695 on Build GRCh38
rs765172164 Chr.12: 49633620-49633622 on Build GRCh38
rs774988682 Chr.12: 49633456-49633456 on Build GRCh38
rs1049424624 Chr.12: 49633809-49633809 on Build GRCh38
rs764921485 Chr.12: 49633678-49633678 on Build GRCh38
rs769010308 Chr.12: 49633810-49633810 on Build GRCh38
rs776861441 Chr.12: 49633383-49633383 on Build GRCh38
rs1042986234 Chr.12: 49633589-49633589 on Build GRCh38
rs774656758 Chr.12: 49633435-49633435 on Build GRCh38
rs759715413 Chr.12: 49633385-49633385 on Build GRCh38
rs757358739 Chr.12: 49633466-49633466 on Build GRCh38
rs753730788 Chr.12: 49633455-49633455 on Build GRCh38
rs776245938 Chr.12: 49633599-49633600 on Build GRCh38
rs776377039 Chr.12: 49633651-49633651 on Build GRCh38
rs111724065 Chr.12: 49633550-49633550 on Build GRCh38
rs781289819 Chr.12: 49633485-49633485 on Build GRCh38
rs1048662532 Chr.12: 49633775-49633775 on Build GRCh38
rs758338828 Chr.12: 49633367-49633367 on Build GRCh38
rs146358365 Chr.12: 49633464-49633464 on Build GRCh38
rs770939994 Chr.12: 49633381-49633381 on Build GRCh38
rs368976299 Chr.12: 49633521-49633521 on Build GRCh38
rs544075321 Chr.12: 49633585-49633585 on Build GRCh38
rs111889469 Chr.12: 49633548-49633548 on Build GRCh38
rs773786374 Chr.12: 49633617-49633617 on Build GRCh38
rs760469292 Chr.12: 49633481-49633486 on Build GRCh38
rs563323303 Chr.12: 49633407-49633407 on Build GRCh38
rs912309460 Chr.12: 49633673-49633673 on Build GRCh38
rs759152170 Chr.12: 49633664-49633664 on Build GRCh38
rs139010275 Chr.12: 49633432-49633432 on Build GRCh38
rs924920487 Chr.12: 49633371-49633371 on Build GRCh38
rs979734520 Chr.12: 49633577-49633577 on Build GRCh38
rs7980832 Chr.12: 49633562-49633562 on Build GRCh38
rs766132488 Chr.12: 49633526-49633526 on Build GRCh38
rs771569615 Chr.12: 49633409-49633409 on Build GRCh38
rs770061278 Chr.12: 49633492-49633492 on Build GRCh38
rs767092050 Chr.12: 49633442-49633442 on Build GRCh38
rs201260222 Chr.12: 49633498-49633498 on Build GRCh38
rs934491985 Chr.12: 49633727-49633727 on Build GRCh38
rs947209572 Chr.12: 49633373-49633373 on Build GRCh38
rs769100446 Chr.12: 49633587-49633587 on Build GRCh38
rs199781280 Chr.12: 49633596-49633596 on Build GRCh38
rs1041206603 Chr.12: 49633732-49633732 on Build GRCh38
rs888107584 Chr.12: 49633826-49633826 on Build GRCh38
rs187093840 Chr.12: 49633763-49633763 on Build GRCh38
rs966989325 Chr.12: 49633366-49633366 on Build GRCh38
rs903013215 Chr.12: 49633400-49633400 on Build GRCh38
rs1005790236 Chr.12: 49633828-49633828 on Build GRCh38
rs762445793 Chr.12: 49633520-49633520 on Build GRCh38
rs778921618 Chr.12: 49633465-49633465 on Build GRCh38
rs953700615 Chr.12: 49633619-49633620 on Build GRCh38
rs775973102 Chr.12: 49633416-49633416 on Build GRCh38
rs774948565 Chr.12: 49633513-49633513 on Build GRCh38
rs763215765 Chr.12: 49633423-49633423 on Build GRCh38
rs749917262 Chr.12: 49633447-49633447 on Build GRCh38
rs559532750 Chr.12: 49633512-49633512 on Build GRCh38
rs768657231 Chr.12: 49633534-49633535 on Build GRCh38
rs770296776 Chr.12: 49633643-49633643 on Build GRCh38
rs181229887 Chr.12: 49633561-49633561 on Build GRCh38
rs142195498 Chr.12: 49633441-49633441 on Build GRCh38
rs535981605 Chr.12: 49633706-49633706 on Build GRCh38

View Full Product Details