Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 102148093-102148611 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PARPBP
Entrez Gene ID
55010
Gene Name
PARP1 binding protein
Gene Aliases
AROM, C12orf48, PARI
UniGene
Hs.330663
Gene Chromosome Location
Chr.12: 102120171-102197520 on Build GRCh38
Transcripts
NM_001319988.1 NM_001319993.1 NM_001319994.1 NM_001319995.1 NM_001319996.1 NM_017915.4 NR_135123.1 XM_011538509.1 XM_011538514.2 XM_017019534.1 XM_017019535.1 XM_017019536.1 XM_017019537.1 XM_017019538.1 XM_017019539.1 XM_017019540.1 XM_017019541.1 XM_017019542.1 XM_017019543.1 XM_017019544.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6015132 c.323T>C p.I108T Chr.12: 102148399-102148399 on Build GRCh38
1586129 c.275G>A p.R92K Chr.12: 102148351-102148351 on Build GRCh38
6737987 c.91C>T p.Q31* Chr.12: 102148410-102148410 on Build GRCh38
1586128 c.294C>A p.F98L Chr.12: 102148370-102148370 on Build GRCh38
274204 c.72A>C p.L24F Chr.12: 102148391-102148391 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs902237653 Chr.12: 102148451-102148451 on Build GRCh38
rs146858211 Chr.12: 102148365-102148365 on Build GRCh38
rs770237141 Chr.12: 102148327-102148327 on Build GRCh38
rs759565924 Chr.12: 102148464-102148464 on Build GRCh38
rs766545153 Chr.12: 102148379-102148379 on Build GRCh38
rs755278471 Chr.12: 102148387-102148387 on Build GRCh38
rs531319438 Chr.12: 102148120-102148120 on Build GRCh38
rs189309284 Chr.12: 102148360-102148360 on Build GRCh38
rs954196042 Chr.12: 102148318-102148318 on Build GRCh38
rs201931080 Chr.12: 102148450-102148451 on Build GRCh38
rs535837377 Chr.12: 102148529-102148529 on Build GRCh38
rs750485473 Chr.12: 102148302-102148302 on Build GRCh38
rs143799695 Chr.12: 102148403-102148403 on Build GRCh38
rs745389902 Chr.12: 102148409-102148409 on Build GRCh38
rs751609649 Chr.12: 102148455-102148455 on Build GRCh38
rs80182547 Chr.12: 102148212-102148212 on Build GRCh38
rs780349930 Chr.12: 102148225-102148225 on Build GRCh38
rs570617394 Chr.12: 102148388-102148388 on Build GRCh38
rs767291778 Chr.12: 102148202-102148202 on Build GRCh38
rs1028752998 Chr.12: 102148538-102148538 on Build GRCh38
rs568013016 Chr.12: 102148183-102148183 on Build GRCh38
rs776988657 Chr.12: 102148236-102148236 on Build GRCh38
rs767664122 Chr.12: 102148480-102148480 on Build GRCh38
rs773864586 Chr.12: 102148337-102148337 on Build GRCh38
rs370016162 Chr.12: 102148574-102148574 on Build GRCh38
rs762477349 Chr.12: 102148271-102148271 on Build GRCh38
rs759195789 Chr.12: 102148275-102148275 on Build GRCh38
rs569976229 Chr.12: 102148432-102148432 on Build GRCh38
rs751621123 Chr.12: 102148471-102148471 on Build GRCh38
rs776588096 Chr.12: 102148199-102148201 on Build GRCh38
rs755519767 Chr.12: 102148477-102148478 on Build GRCh38
rs538702845 Chr.12: 102148227-102148229 on Build GRCh38
rs1027803785 Chr.12: 102148189-102148189 on Build GRCh38
rs781578179 Chr.12: 102148325-102148325 on Build GRCh38
rs756478975 Chr.12: 102148395-102148395 on Build GRCh38
rs778531249 Chr.12: 102148407-102148407 on Build GRCh38
rs758505900 Chr.12: 102148306-102148306 on Build GRCh38
rs771072574 Chr.12: 102148544-102148544 on Build GRCh38
rs779173180 Chr.12: 102148216-102148216 on Build GRCh38
rs555825142 Chr.12: 102148539-102148539 on Build GRCh38
rs765908375 Chr.12: 102148274-102148274 on Build GRCh38
rs1019163041 Chr.12: 102148171-102148171 on Build GRCh38
rs753839083 Chr.12: 102148352-102148352 on Build GRCh38
rs764149732 Chr.12: 102148290-102148290 on Build GRCh38
rs773744775 Chr.12: 102148329-102148329 on Build GRCh38
rs80135324 Chr.12: 102148381-102148381 on Build GRCh38
rs140043679 Chr.12: 102148422-102148422 on Build GRCh38
rs778198735 Chr.12: 102148397-102148397 on Build GRCh38
rs869203462 Chr.12: 102148211-102148212 on Build GRCh38
rs937191091 Chr.12: 102148411-102148411 on Build GRCh38
rs375425230 Chr.12: 102148436-102148436 on Build GRCh38
rs748632563 Chr.12: 102148326-102148326 on Build GRCh38
rs748732746 Chr.12: 102148335-102148335 on Build GRCh38
rs756168613 Chr.12: 102148289-102148289 on Build GRCh38
rs548422032 Chr.12: 102148357-102148357 on Build GRCh38
rs1032549293 Chr.12: 102148305-102148305 on Build GRCh38
rs760487298 Chr.12: 102148417-102148417 on Build GRCh38
rs752933973 Chr.12: 102148487-102148487 on Build GRCh38
rs752648521 Chr.12: 102148287-102148287 on Build GRCh38
rs185647149 Chr.12: 102148198-102148198 on Build GRCh38
rs757406308 Chr.12: 102148207-102148207 on Build GRCh38
rs527525020 Chr.12: 102148205-102148205 on Build GRCh38
rs747267431 Chr.12: 102148229-102148229 on Build GRCh38
rs61936658 Chr.12: 102148214-102148214 on Build GRCh38
rs750612420 Chr.12: 102148249-102148250 on Build GRCh38
rs975354964 Chr.12: 102148188-102148188 on Build GRCh38
rs769265709 Chr.12: 102148252-102148252 on Build GRCh38
rs746955886 Chr.12: 102148198-102148199 on Build GRCh38
rs756460503 Chr.12: 102148496-102148496 on Build GRCh38
rs758644913 Chr.12: 102148221-102148221 on Build GRCh38
rs200640822 Chr.12: 102148419-102148419 on Build GRCh38
rs556815499 Chr.12: 102148119-102148119 on Build GRCh38
rs772762236 Chr.12: 102148262-102148262 on Build GRCh38
rs747312058 Chr.12: 102148320-102148320 on Build GRCh38
rs757730862 Chr.12: 102148514-102148514 on Build GRCh38
rs767405684 Chr.12: 102148286-102148286 on Build GRCh38
rs748438568 Chr.12: 102148166-102148166 on Build GRCh38
rs769075882 Chr.12: 102148232-102148232 on Build GRCh38
rs1009889507 Chr.12: 102148475-102148475 on Build GRCh38
rs748724190 Chr.12: 102148250-102148250 on Build GRCh38
rs374160614 Chr.12: 102148346-102148346 on Build GRCh38
rs377556453 Chr.12: 102148347-102148347 on Build GRCh38
rs1018562106 Chr.12: 102148536-102148536 on Build GRCh38
rs753020014 Chr.12: 102148393-102148393 on Build GRCh38
rs895743436 Chr.12: 102148438-102148438 on Build GRCh38
rs751263978 Chr.12: 102148201-102148201 on Build GRCh38
rs771739645 Chr.12: 102148343-102148343 on Build GRCh38
rs952255822 Chr.12: 102148194-102148194 on Build GRCh38
rs370115747 Chr.12: 102148510-102148510 on Build GRCh38
rs746065106 Chr.12: 102148219-102148219 on Build GRCh38
rs749868298 Chr.12: 102148399-102148399 on Build GRCh38
rs779315973 Chr.12: 102148524-102148524 on Build GRCh38
rs996428065 Chr.12: 102148566-102148566 on Build GRCh38
rs761852108 Chr.12: 102148430-102148430 on Build GRCh38
rs900773398 Chr.12: 102148548-102148548 on Build GRCh38
rs760742796 Chr.12: 102148199-102148200 on Build GRCh38
rs1001369296 Chr.12: 102148200-102148200 on Build GRCh38
rs780078295 Chr.12: 102148310-102148310 on Build GRCh38
rs778180341 Chr.12: 102148503-102148503 on Build GRCh38
rs76358859 Chr.12: 102148199-102148199 on Build GRCh38
rs548217913 Chr.12: 102148128-102148128 on Build GRCh38
rs918690882 Chr.12: 102148161-102148161 on Build GRCh38
rs148169413 Chr.12: 102148410-102148410 on Build GRCh38
rs759270887 Chr.12: 102148338-102148338 on Build GRCh38

View Full Product Details