Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 114366072-114366623 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBX5
Entrez Gene ID
6910
Gene Name
T-box 5
Gene Aliases
HOS
UniGene
Hs.381715
Gene Chromosome Location
Chr.12: 114353911-114408708 on Build GRCh38
Transcripts
NM_000192.3 NM_080717.2 NM_181486.2 XM_017019912.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5748926 c.1025T>C p.L342P Chr.12: 114366122-114366122 on Build GRCh38
3456502 c.977G>A p.R326K Chr.12: 114366170-114366170 on Build GRCh38
2172915 c.982+5G>C p.? Chr.12: 114366160-114366160 on Build GRCh38
5600630 c.982G>A p.E328K Chr.12: 114366165-114366165 on Build GRCh38
4562851 c.946G>A p.E316K Chr.12: 114366201-114366201 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs571667858 Chr.12: 114366414-114366414 on Build GRCh38
rs753912503 Chr.12: 114366431-114366431 on Build GRCh38
rs759009256 Chr.12: 114366182-114366182 on Build GRCh38
rs929557347 Chr.12: 114366571-114366571 on Build GRCh38
rs760334883 Chr.12: 114366225-114366225 on Build GRCh38
rs973621936 Chr.12: 114366245-114366245 on Build GRCh38
rs764726865 Chr.12: 114366405-114366405 on Build GRCh38
rs377223201 Chr.12: 114366151-114366151 on Build GRCh38
rs1039442231 Chr.12: 114366342-114366342 on Build GRCh38
rs201071418 Chr.12: 114366356-114366356 on Build GRCh38
rs370805715 Chr.12: 114366379-114366379 on Build GRCh38
rs911794484 Chr.12: 114366568-114366568 on Build GRCh38
rs953210556 Chr.12: 114366557-114366557 on Build GRCh38
rs1024789848 Chr.12: 114366444-114366444 on Build GRCh38
rs752101162 Chr.12: 114366203-114366203 on Build GRCh38
rs139154253 Chr.12: 114366113-114366113 on Build GRCh38
rs758752836 Chr.12: 114366233-114366233 on Build GRCh38
rs147405081 Chr.12: 114366360-114366360 on Build GRCh38
rs3741694 Chr.12: 114366122-114366122 on Build GRCh38
rs757373962 Chr.12: 114366348-114366348 on Build GRCh38
rs963386803 Chr.12: 114366512-114366512 on Build GRCh38
rs750627729 Chr.12: 114366545-114366545 on Build GRCh38
rs770206057 Chr.12: 114366253-114366253 on Build GRCh38
rs973268253 Chr.12: 114366537-114366537 on Build GRCh38
rs1029274901 Chr.12: 114366549-114366549 on Build GRCh38
rs201164281 Chr.12: 114366220-114366220 on Build GRCh38
rs200078178 Chr.12: 114366141-114366141 on Build GRCh38
rs763672657 Chr.12: 114366228-114366228 on Build GRCh38
rs750855891 Chr.12: 114366229-114366229 on Build GRCh38
rs748400295 Chr.12: 114366147-114366147 on Build GRCh38
rs372915117 Chr.12: 114366412-114366412 on Build GRCh38
rs181098485 Chr.12: 114366533-114366533 on Build GRCh38
rs531799406 Chr.12: 114366397-114366397 on Build GRCh38
rs762836528 Chr.12: 114366152-114366152 on Build GRCh38
rs863223788 Chr.12: 114366312-114366312 on Build GRCh38
rs867051300 Chr.12: 114366224-114366224 on Build GRCh38
rs767123996 Chr.12: 114366100-114366100 on Build GRCh38
rs919531597 Chr.12: 114366552-114366552 on Build GRCh38
rs987987717 Chr.12: 114366103-114366103 on Build GRCh38
rs752365122 Chr.12: 114366111-114366111 on Build GRCh38
rs767086503 Chr.12: 114366199-114366199 on Build GRCh38
rs755387654 Chr.12: 114366247-114366247 on Build GRCh38
rs35110399 Chr.12: 114366343-114366343 on Build GRCh38
rs780609256 Chr.12: 114366123-114366123 on Build GRCh38
rs201637366 Chr.12: 114366252-114366252 on Build GRCh38
rs773978838 Chr.12: 114366288-114366288 on Build GRCh38
rs926640743 Chr.12: 114366150-114366150 on Build GRCh38
rs970507942 Chr.12: 114366458-114366458 on Build GRCh38
rs774986848 Chr.12: 114366305-114366305 on Build GRCh38
rs550240868 Chr.12: 114366408-114366408 on Build GRCh38
rs147077037 Chr.12: 114366299-114366299 on Build GRCh38
rs1035940533 Chr.12: 114366407-114366407 on Build GRCh38
rs146908258 Chr.12: 114366319-114366319 on Build GRCh38
rs760177359 Chr.12: 114366306-114366306 on Build GRCh38
rs375873875 Chr.12: 114366140-114366140 on Build GRCh38
rs878853750 Chr.12: 114366189-114366189 on Build GRCh38
rs745840026 Chr.12: 114366292-114366292 on Build GRCh38
rs778126616 Chr.12: 114366280-114366280 on Build GRCh38
rs756238293 Chr.12: 114366330-114366330 on Build GRCh38
rs752134526 Chr.12: 114366237-114366237 on Build GRCh38
rs781342709 Chr.12: 114366251-114366251 on Build GRCh38
rs778784399 Chr.12: 114366352-114366352 on Build GRCh38
rs369697961 Chr.12: 114366318-114366318 on Build GRCh38
rs963270797 Chr.12: 114366244-114366244 on Build GRCh38
rs777287804 Chr.12: 114366349-114366349 on Build GRCh38
rs886048997 Chr.12: 114366301-114366301 on Build GRCh38
rs867641094 Chr.12: 114366267-114366267 on Build GRCh38
rs564499369 Chr.12: 114366310-114366310 on Build GRCh38
rs200382742 Chr.12: 114366378-114366378 on Build GRCh38
rs764291197 Chr.12: 114366418-114366418 on Build GRCh38
rs28730762 Chr.12: 114366149-114366149 on Build GRCh38
rs778180226 Chr.12: 114366334-114366334 on Build GRCh38
rs535994221 Chr.12: 114366143-114366143 on Build GRCh38
rs980831875 Chr.12: 114366472-114366472 on Build GRCh38
rs142609815 Chr.12: 114366313-114366313 on Build GRCh38
rs763797226 Chr.12: 114366307-114366307 on Build GRCh38
rs537547818 Chr.12: 114366293-114366293 on Build GRCh38
rs987370667 Chr.12: 114366565-114366565 on Build GRCh38
rs779274382 Chr.12: 114366120-114366120 on Build GRCh38
rs375955080 Chr.12: 114366361-114366361 on Build GRCh38
rs78344365 Chr.12: 114366417-114366417 on Build GRCh38
rs147977741 Chr.12: 114366320-114366320 on Build GRCh38
rs115178276 Chr.12: 114366311-114366311 on Build GRCh38
rs761537129 Chr.12: 114366400-114366400 on Build GRCh38
rs774321607 Chr.12: 114366173-114366173 on Build GRCh38
rs575701831 Chr.12: 114366153-114366153 on Build GRCh38
rs770728362 Chr.12: 114366283-114366283 on Build GRCh38
rs377625550 Chr.12: 114366366-114366366 on Build GRCh38
rs973631007 Chr.12: 114366210-114366210 on Build GRCh38
rs543015692 Chr.12: 114366282-114366282 on Build GRCh38
rs752879569 Chr.12: 114366329-114366329 on Build GRCh38
rs1033688004 Chr.12: 114366509-114366509 on Build GRCh38
rs757507401 Chr.12: 114366438-114366438 on Build GRCh38
rs766716477 Chr.12: 114366363-114366364 on Build GRCh38
rs963419850 Chr.12: 114366498-114366499 on Build GRCh38

View Full Product Details