Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 132772151-132772656 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOLGA3
Entrez Gene ID
2802
Gene Name
golgin A3
Gene Aliases
GCP170, MEA-2
UniGene
Hs.507333
Gene Chromosome Location
Chr.12: 132768909-132829131 on Build GRCh38
Transcripts
NM_005895.3 XM_005266164.4 XM_005266167.4 XM_006719736.3 XM_006719737.3 XM_011534793.2 XM_011534794.2

SNP Detail

SNP ID Chromosome Location
rs964002651 Chr.12: 132772179-132772181 on Build GRCh38
rs1033376795 Chr.12: 132772488-132772488 on Build GRCh38
rs186095958 Chr.12: 132772459-132772459 on Build GRCh38
rs927068566 Chr.12: 132772557-132772557 on Build GRCh38
rs150250212 Chr.12: 132772270-132772270 on Build GRCh38
rs3182579 Chr.12: 132772442-132772442 on Build GRCh38
rs1009786243 Chr.12: 132772458-132772458 on Build GRCh38
rs10747082 Chr.12: 132772589-132772589 on Build GRCh38
rs528481150 Chr.12: 132772286-132772286 on Build GRCh38
rs956974193 Chr.12: 132772225-132772225 on Build GRCh38
rs546267582 Chr.12: 132772468-132772468 on Build GRCh38
rs896046176 Chr.12: 132772445-132772445 on Build GRCh38
rs918254078 Chr.12: 132772552-132772552 on Build GRCh38
rs985316514 Chr.12: 132772516-132772516 on Build GRCh38
rs1048118186 Chr.12: 132772554-132772554 on Build GRCh38
rs766408381 Chr.12: 132772231-132772231 on Build GRCh38
rs1029117966 Chr.12: 132772490-132772490 on Build GRCh38
rs370699037 Chr.12: 132772566-132772566 on Build GRCh38
rs769496180 Chr.12: 132772546-132772547 on Build GRCh38
rs1038917286 Chr.12: 132772563-132772563 on Build GRCh38
rs1001736108 Chr.12: 132772199-132772199 on Build GRCh38
rs768001834 Chr.12: 132772241-132772243 on Build GRCh38
rs530785276 Chr.12: 132772413-132772413 on Build GRCh38
rs117217517 Chr.12: 132772265-132772265 on Build GRCh38
rs546346746 Chr.12: 132772531-132772531 on Build GRCh38
rs63403663 Chr.12: 132772390-132772390 on Build GRCh38
rs988851260 Chr.12: 132772211-132772211 on Build GRCh38
rs538236880 Chr.12: 132772530-132772530 on Build GRCh38
rs3182583 Chr.12: 132772370-132772370 on Build GRCh38
rs775236056 Chr.12: 132772344-132772344 on Build GRCh38
rs1032589956 Chr.12: 132772214-132772214 on Build GRCh38
rs987368135 Chr.12: 132772556-132772556 on Build GRCh38
rs1042400914 Chr.12: 132772565-132772565 on Build GRCh38
rs180812771 Chr.12: 132772207-132772207 on Build GRCh38
rs912405206 Chr.12: 132772526-132772526 on Build GRCh38
rs1001685081 Chr.12: 132772437-132772437 on Build GRCh38
rs973145661 Chr.12: 132772492-132772492 on Build GRCh38
rs1000227337 Chr.12: 132772584-132772584 on Build GRCh38
rs1009000670 Chr.12: 132772619-132772619 on Build GRCh38
rs867170454 Chr.12: 132772544-132772544 on Build GRCh38
rs146733732 Chr.12: 132772566-132772567 on Build GRCh38
rs1039809449 Chr.12: 132772407-132772407 on Build GRCh38
rs906041890 Chr.12: 132772187-132772187 on Build GRCh38
rs1035935905 Chr.12: 132772203-132772203 on Build GRCh38
rs565173140 Chr.12: 132772361-132772361 on Build GRCh38
rs867437457 Chr.12: 132772564-132772564 on Build GRCh38
rs906031836 Chr.12: 132772416-132772416 on Build GRCh38
rs968802438 Chr.12: 132772251-132772251 on Build GRCh38
rs931036226 Chr.12: 132772405-132772405 on Build GRCh38
rs1010918190 Chr.12: 132772454-132772454 on Build GRCh38
rs919157242 Chr.12: 132772342-132772342 on Build GRCh38
rs200631103 Chr.12: 132772386-132772386 on Build GRCh38
rs879865801 Chr.12: 132772338-132772338 on Build GRCh38
rs980939764 Chr.12: 132772473-132772473 on Build GRCh38
rs536785424 Chr.12: 132772544-132772546 on Build GRCh38
rs776077686 Chr.12: 132772543-132772544 on Build GRCh38
rs538265053 Chr.12: 132772496-132772496 on Build GRCh38
rs370885309 Chr.12: 132772463-132772463 on Build GRCh38
rs1127070 Chr.12: 132772461-132772461 on Build GRCh38
rs563865451 Chr.12: 132772216-132772216 on Build GRCh38
rs953420389 Chr.12: 132772596-132772596 on Build GRCh38
rs943210921 Chr.12: 132772562-132772562 on Build GRCh38
rs777675512 Chr.12: 132772476-132772476 on Build GRCh38
rs936707900 Chr.12: 132772567-132772567 on Build GRCh38
rs142291653 Chr.12: 132772491-132772491 on Build GRCh38
rs752749782 Chr.12: 132772296-132772296 on Build GRCh38
rs773391216 Chr.12: 132772196-132772196 on Build GRCh38
rs1000857823 Chr.12: 132772419-132772419 on Build GRCh38
rs139008204 Chr.12: 132772287-132772287 on Build GRCh38
rs1020722350 Chr.12: 132772213-132772213 on Build GRCh38
rs3182581 Chr.12: 132772399-132772399 on Build GRCh38
rs1057200266 Chr.12: 132772559-132772559 on Build GRCh38
rs2046289 Chr.12: 132772377-132772377 on Build GRCh38
rs1025216620 Chr.12: 132772594-132772594 on Build GRCh38
rs3182580 Chr.12: 132772436-132772436 on Build GRCh38
rs559817080 Chr.12: 132772281-132772281 on Build GRCh38
rs911839178 Chr.12: 132772557-132772558 on Build GRCh38
rs34381645 Chr.12: 132772544-132772545 on Build GRCh38
rs3182578 Chr.12: 132772443-132772443 on Build GRCh38
rs12309204 Chr.12: 132772467-132772467 on Build GRCh38
rs1025649007 Chr.12: 132772465-132772465 on Build GRCh38
rs58406410 Chr.12: 132772563-132772565 on Build GRCh38
rs980505620 Chr.12: 132772284-132772284 on Build GRCh38
rs937109353 Chr.12: 132772558-132772558 on Build GRCh38
rs928973251 Chr.12: 132772553-132772554 on Build GRCh38
rs895980279 Chr.12: 132772561-132772561 on Build GRCh38
rs931013595 Chr.12: 132772553-132772553 on Build GRCh38
rs3182582 Chr.12: 132772393-132772393 on Build GRCh38
rs1053878894 Chr.12: 132772578-132772578 on Build GRCh38
rs896829684 Chr.12: 132772382-132772382 on Build GRCh38

View Full Product Details