Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 25251561-25252075 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTMR6
Entrez Gene ID
9107
Gene Name
myotubularin related protein 6
Gene Aliases
-
UniGene
Hs.643702
Gene Chromosome Location
Chr.13: 25246201-25287566 on Build GRCh38
Transcripts
NM_004685.3 XM_011535306.1 XM_011535307.1 XM_017020845.1 XM_017020846.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2069569 c.1496A>G p.Y499C Chr.13: 25251758-25251758 on Build GRCh38
1734043 c.1471A>G p.N491D Chr.13: 25251860-25251860 on Build GRCh38
1289885 c.1563G>A p.E521E Chr.13: 25251691-25251691 on Build GRCh38
5970946 c.1386G>A p.L462L Chr.13: 25251945-25251945 on Build GRCh38
5952001 c.1526G>A p.R509K Chr.13: 25251728-25251728 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777565178 Chr.13: 25251779-25251779 on Build GRCh38
rs146057731 Chr.13: 25251906-25251906 on Build GRCh38
rs772413591 Chr.13: 25251917-25251917 on Build GRCh38
rs1050317428 Chr.13: 25251857-25251857 on Build GRCh38
rs778454169 Chr.13: 25252012-25252012 on Build GRCh38
rs767753877 Chr.13: 25251739-25251739 on Build GRCh38
rs923591431 Chr.13: 25251743-25251743 on Build GRCh38
rs76317728 Chr.13: 25251945-25251945 on Build GRCh38
rs199892242 Chr.13: 25251827-25251827 on Build GRCh38
rs769331411 Chr.13: 25251810-25251810 on Build GRCh38
rs754865055 Chr.13: 25251605-25251605 on Build GRCh38
rs141853933 Chr.13: 25251852-25251852 on Build GRCh38
rs771366436 Chr.13: 25252029-25252029 on Build GRCh38
rs572366130 Chr.13: 25251876-25251876 on Build GRCh38
rs766414006 Chr.13: 25251863-25251863 on Build GRCh38
rs758190885 Chr.13: 25251625-25251625 on Build GRCh38
rs750462481 Chr.13: 25251744-25251744 on Build GRCh38
rs371094946 Chr.13: 25251814-25251814 on Build GRCh38
rs376021333 Chr.13: 25251689-25251689 on Build GRCh38
rs761969841 Chr.13: 25251722-25251722 on Build GRCh38
rs748650975 Chr.13: 25252014-25252014 on Build GRCh38
rs746591372 Chr.13: 25251641-25251641 on Build GRCh38
rs760864949 Chr.13: 25251855-25251855 on Build GRCh38
rs771448432 Chr.13: 25252003-25252004 on Build GRCh38
rs987274291 Chr.13: 25251589-25251589 on Build GRCh38
rs974517307 Chr.13: 25251648-25251648 on Build GRCh38
rs576112454 Chr.13: 25251806-25251806 on Build GRCh38
rs772466411 Chr.13: 25251842-25251842 on Build GRCh38
rs755729166 Chr.13: 25251902-25251902 on Build GRCh38
rs4312169 Chr.13: 25251751-25251751 on Build GRCh38
rs769060070 Chr.13: 25251826-25251826 on Build GRCh38
rs748076922 Chr.13: 25251622-25251622 on Build GRCh38
rs964113142 Chr.13: 25251615-25251615 on Build GRCh38
rs750202009 Chr.13: 25251995-25251995 on Build GRCh38
rs144582137 Chr.13: 25251679-25251679 on Build GRCh38
rs370493335 Chr.13: 25251905-25251905 on Build GRCh38
rs898703950 Chr.13: 25251894-25251894 on Build GRCh38
rs778859254 Chr.13: 25251845-25251845 on Build GRCh38
rs770720770 Chr.13: 25251678-25251678 on Build GRCh38
rs1000771043 Chr.13: 25251938-25251938 on Build GRCh38
rs758305289 Chr.13: 25252046-25252046 on Build GRCh38
rs138289796 Chr.13: 25251950-25251950 on Build GRCh38
rs200817508 Chr.13: 25251799-25251799 on Build GRCh38
rs533554917 Chr.13: 25251599-25251599 on Build GRCh38
rs201709535 Chr.13: 25251895-25251895 on Build GRCh38
rs765983201 Chr.13: 25252000-25252000 on Build GRCh38
rs761906117 Chr.13: 25251981-25251981 on Build GRCh38
rs564101021 Chr.13: 25251784-25251785 on Build GRCh38
rs776691722 Chr.13: 25252036-25252036 on Build GRCh38
rs745562094 Chr.13: 25251807-25251807 on Build GRCh38
rs755746839 Chr.13: 25251996-25251996 on Build GRCh38
rs750093054 Chr.13: 25251897-25251897 on Build GRCh38
rs774706545 Chr.13: 25251717-25251717 on Build GRCh38
rs747363693 Chr.13: 25252025-25252025 on Build GRCh38
rs762898535 Chr.13: 25251878-25251878 on Build GRCh38
rs774413920 Chr.13: 25251980-25251980 on Build GRCh38
rs765453389 Chr.13: 25251972-25251972 on Build GRCh38
rs372446234 Chr.13: 25252020-25252020 on Build GRCh38
rs749722751 Chr.13: 25251946-25251952 on Build GRCh38
rs753572087 Chr.13: 25252004-25252004 on Build GRCh38
rs758271738 Chr.13: 25251776-25251776 on Build GRCh38
rs536417177 Chr.13: 25251762-25251762 on Build GRCh38
rs777495032 Chr.13: 25251628-25251628 on Build GRCh38
rs779561417 Chr.13: 25251903-25251903 on Build GRCh38
rs776731789 Chr.13: 25251953-25251953 on Build GRCh38
rs140082792 Chr.13: 25251966-25251966 on Build GRCh38
rs766419466 Chr.13: 25251753-25251753 on Build GRCh38
rs748703971 Chr.13: 25251911-25251911 on Build GRCh38
rs769223877 Chr.13: 25251682-25251682 on Build GRCh38
rs187816844 Chr.13: 25251888-25251888 on Build GRCh38
rs113373326 Chr.13: 25251877-25251877 on Build GRCh38
rs778061798 Chr.13: 25252019-25252019 on Build GRCh38
rs911331513 Chr.13: 25251597-25251597 on Build GRCh38
rs112378600 Chr.13: 25251600-25251600 on Build GRCh38
rs558630526 Chr.13: 25251839-25251839 on Build GRCh38
rs751421897 Chr.13: 25251781-25251781 on Build GRCh38
rs770216169 Chr.13: 25251976-25251976 on Build GRCh38
rs779494309 Chr.13: 25251812-25251812 on Build GRCh38
rs764158169 Chr.13: 25251880-25251880 on Build GRCh38
rs756160061 Chr.13: 25251833-25251833 on Build GRCh38
rs757092189 Chr.13: 25251891-25251891 on Build GRCh38
rs767351906 Chr.13: 25251991-25251991 on Build GRCh38
rs767779503 Chr.13: 25251711-25251711 on Build GRCh38
rs754462109 Chr.13: 25252005-25252005 on Build GRCh38
rs758912334 Chr.13: 25252016-25252016 on Build GRCh38
rs763425454 Chr.13: 25251707-25251707 on Build GRCh38
rs760892112 Chr.13: 25251929-25251929 on Build GRCh38
rs771079522 Chr.13: 25251944-25251944 on Build GRCh38

View Full Product Details