Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 32316289-32316759 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRCA2
Entrez Gene ID
675
Gene Name
BRCA2, DNA repair associated
Gene Aliases
BRCC2, BROVCA2, FACD, FAD, FAD1, FANCD, FANCD1, GLM3, PNCA2, XRCC11
UniGene
Hs.34012
Gene Chromosome Location
Chr.13: 32315480-32399672 on Build GRCh38
Transcripts
NM_000059.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3981996 c.3G>A p.M1I Chr.13: 32316463-32316463 on Build GRCh38
6966217 c.67+1G>T p.? Chr.13: 32316528-32316528 on Build GRCh38
2071226 c.47A>G p.K16R Chr.13: 32316507-32316507 on Build GRCh38
6343313 c.53G>T p.R18L Chr.13: 32316513-32316513 on Build GRCh38
6506579 c.67+1G>T p.? Chr.13: 32316528-32316528 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756079749 Chr.13: 32316581-32316581 on Build GRCh38
rs80358836 Chr.13: 32316465-32316465 on Build GRCh38
rs76874770 Chr.13: 32316450-32316450 on Build GRCh38
rs397507687 Chr.13: 32316497-32316504 on Build GRCh38
rs397507772 Chr.13: 32316511-32316512 on Build GRCh38
rs997987465 Chr.13: 32316615-32316615 on Build GRCh38
rs529148674 Chr.13: 32316543-32316543 on Build GRCh38
rs763296677 Chr.13: 32316433-32316433 on Build GRCh38
rs398122544 Chr.13: 32316519-32316519 on Build GRCh38
rs896998548 Chr.13: 32316321-32316321 on Build GRCh38
rs878853592 Chr.13: 32316517-32316517 on Build GRCh38
rs886040552 Chr.13: 32316508-32316509 on Build GRCh38
rs189026060 Chr.13: 32316609-32316609 on Build GRCh38
rs397507566 Chr.13: 32316428-32316428 on Build GRCh38
rs764618190 Chr.13: 32316434-32316434 on Build GRCh38
rs276174799 Chr.13: 32316406-32316406 on Build GRCh38
rs276174802 Chr.13: 32316452-32316452 on Build GRCh38
rs749389200 Chr.13: 32316606-32316606 on Build GRCh38
rs765644109 Chr.13: 32316418-32316418 on Build GRCh38
rs367697028 Chr.13: 32316564-32316564 on Build GRCh38
rs879915769 Chr.13: 32316514-32316514 on Build GRCh38
rs80358762 Chr.13: 32316513-32316513 on Build GRCh38
rs754976009 Chr.13: 32316559-32316559 on Build GRCh38
rs950489130 Chr.13: 32316344-32316345 on Build GRCh38
rs1057519494 Chr.13: 32316489-32316489 on Build GRCh38
rs863224464 Chr.13: 32316461-32316461 on Build GRCh38
rs80358527 Chr.13: 32316486-32316486 on Build GRCh38
rs276174800 Chr.13: 32316417-32316417 on Build GRCh38
rs994689164 Chr.13: 32316635-32316635 on Build GRCh38
rs80359399 Chr.13: 32316496-32316496 on Build GRCh38
rs754814638 Chr.13: 32316451-32316451 on Build GRCh38
rs983136114 Chr.13: 32316701-32316704 on Build GRCh38
rs371493518 Chr.13: 32316567-32316567 on Build GRCh38
rs183997506 Chr.13: 32316386-32316386 on Build GRCh38
rs11571574 Chr.13: 32316589-32316589 on Build GRCh38
rs751297272 Chr.13: 32316362-32316362 on Build GRCh38
rs545355025 Chr.13: 32316419-32316419 on Build GRCh38
rs786201560 Chr.13: 32316512-32316512 on Build GRCh38
rs397507367 Chr.13: 32316522-32316522 on Build GRCh38
rs759888980 Chr.13: 32316416-32316416 on Build GRCh38
rs587781738 Chr.13: 32316532-32316532 on Build GRCh38
rs551309156 Chr.13: 32316612-32316612 on Build GRCh38
rs757349122 Chr.13: 32316438-32316438 on Build GRCh38
rs276174797 Chr.13: 32316427-32316427 on Build GRCh38
rs886038198 Chr.13: 32316500-32316500 on Build GRCh38
rs935412934 Chr.13: 32316554-32316554 on Build GRCh38
rs587782872 Chr.13: 32316495-32316495 on Build GRCh38
rs431825277 Chr.13: 32316458-32316458 on Build GRCh38
rs80359418 Chr.13: 32316463-32316463 on Build GRCh38
rs886040574 Chr.13: 32316512-32316521 on Build GRCh38
rs587782137 Chr.13: 32316471-32316471 on Build GRCh38
rs1027279204 Chr.13: 32316656-32316656 on Build GRCh38
rs979662084 Chr.13: 32316694-32316694 on Build GRCh38
rs761283611 Chr.13: 32316457-32316457 on Build GRCh38
rs876659884 Chr.13: 32316460-32316460 on Build GRCh38
rs80359298 Chr.13: 32316477-32316478 on Build GRCh38
rs80359582 Chr.13: 32316523-32316523 on Build GRCh38
rs398122721 Chr.13: 32316408-32316408 on Build GRCh38
rs276174798 Chr.13: 32316410-32316412 on Build GRCh38
rs920231956 Chr.13: 32316311-32316311 on Build GRCh38
rs80358622 Chr.13: 32316497-32316497 on Build GRCh38
rs967271763 Chr.13: 32316659-32316659 on Build GRCh38
rs1026907148 Chr.13: 32316388-32316388 on Build GRCh38
rs397507623 Chr.13: 32316482-32316483 on Build GRCh38
rs758732038 Chr.13: 32316420-32316421 on Build GRCh38
rs896941748 Chr.13: 32316582-32316582 on Build GRCh38
rs80359393 Chr.13: 32316496-32316497 on Build GRCh38
rs81002796 Chr.13: 32316528-32316528 on Build GRCh38
rs397507881 Chr.13: 32316527-32316527 on Build GRCh38
rs770479195 Chr.13: 32316467-32316467 on Build GRCh38
rs786203080 Chr.13: 32316488-32316488 on Build GRCh38
rs138705202 Chr.13: 32316446-32316446 on Build GRCh38
rs587780865 Chr.13: 32316546-32316546 on Build GRCh38
rs876660440 Chr.13: 32316507-32316507 on Build GRCh38
rs747900115 Chr.13: 32316457-32316457 on Build GRCh38
rs746566500 Chr.13: 32316466-32316466 on Build GRCh38
rs373546450 Chr.13: 32316531-32316531 on Build GRCh38
rs80358650 Chr.13: 32316463-32316463 on Build GRCh38
rs80359445 Chr.13: 32316504-32316505 on Build GRCh38
rs538947930 Chr.13: 32316585-32316585 on Build GRCh38
rs750422809 Chr.13: 32316549-32316549 on Build GRCh38
rs1021043789 Chr.13: 32316643-32316643 on Build GRCh38
rs863224439 Chr.13: 32316529-32316529 on Build GRCh38
rs120074212 Chr.13: 32316676-32316676 on Build GRCh38
rs983123559 Chr.13: 32316387-32316387 on Build GRCh38
rs775945279 Chr.13: 32316431-32316431 on Build GRCh38
rs786201444 Chr.13: 32316487-32316487 on Build GRCh38
rs483353081 Chr.13: 32316508-32316510 on Build GRCh38
rs1032535406 Chr.13: 32316719-32316719 on Build GRCh38
rs397507820 Chr.13: 32316465-32316465 on Build GRCh38
rs276174796 Chr.13: 32316447-32316447 on Build GRCh38
rs886040931 Chr.13: 32316528-32316528 on Build GRCh38
rs889746951 Chr.13: 32316602-32316602 on Build GRCh38
rs765135557 Chr.13: 32316561-32316561 on Build GRCh38
rs276174795 Chr.13: 32316449-32316449 on Build GRCh38
rs276174801 Chr.13: 32316415-32316415 on Build GRCh38
rs864622207 Chr.13: 32316490-32316490 on Build GRCh38
rs758183252 Chr.13: 32316565-32316565 on Build GRCh38
rs774714454 Chr.13: 32316535-32316535 on Build GRCh38
rs80359343 Chr.13: 32316486-32316486 on Build GRCh38
rs767648965 Chr.13: 32316544-32316544 on Build GRCh38
rs80358547 Chr.13: 32316462-32316462 on Build GRCh38
rs1799943 Chr.13: 32316435-32316435 on Build GRCh38
rs386833396 Chr.13: 32316510-32316510 on Build GRCh38
rs80359483 Chr.13: 32316511-32316512 on Build GRCh38
rs950353511 Chr.13: 32316669-32316669 on Build GRCh38
rs80359400 Chr.13: 32316497-32316498 on Build GRCh38
rs397507571 Chr.13: 32316470-32316470 on Build GRCh38
rs751741252 Chr.13: 32316437-32316437 on Build GRCh38
rs794727232 Chr.13: 32316476-32316476 on Build GRCh38
rs80358597 Chr.13: 32316494-32316494 on Build GRCh38
rs81002885 Chr.13: 32316529-32316529 on Build GRCh38
rs397507260 Chr.13: 32316402-32316402 on Build GRCh38
rs777190345 Chr.13: 32316412-32316412 on Build GRCh38

View Full Product Details