Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 38349912-38350419 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UFM1
Entrez Gene ID
51569
Gene Name
ubiquitin fold modifier 1
Gene Aliases
BM-002, C13orf20
UniGene
Hs.643655
Gene Chromosome Location
Chr.13: 38349771-38363007 on Build GRCh38
Transcripts
NM_001286703.1 NM_001286704.1 NM_001286705.1 NM_001286706.1 NM_016617.3 NR_104584.1 NR_104585.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3468781 c.14C>T p.S5F Chr.13: 38350010-38350010 on Build GRCh38
6331741 c.58A>G p.T20A Chr.13: 38350172-38350172 on Build GRCh38
3468780 c.9G>T p.K3N Chr.13: 38350005-38350005 on Build GRCh38
4447420 c.3-5C>T p.? Chr.13: 38349994-38349994 on Build GRCh38
6199819 c.79C>G p.P27A Chr.13: 38350193-38350193 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1027317083 Chr.13: 38350068-38350068 on Build GRCh38
rs1005123648 Chr.13: 38350093-38350093 on Build GRCh38
rs780692347 Chr.13: 38350099-38350099 on Build GRCh38
rs759711461 Chr.13: 38350185-38350185 on Build GRCh38
rs766682188 Chr.13: 38350130-38350130 on Build GRCh38
rs369545378 Chr.13: 38350164-38350164 on Build GRCh38
rs779839332 Chr.13: 38350223-38350223 on Build GRCh38
rs2231330 Chr.13: 38349946-38349946 on Build GRCh38
rs779447610 Chr.13: 38350067-38350067 on Build GRCh38
rs747984923 Chr.13: 38350230-38350230 on Build GRCh38
rs905600072 Chr.13: 38350355-38350355 on Build GRCh38
rs779389046 Chr.13: 38349985-38349985 on Build GRCh38
rs887412547 Chr.13: 38350297-38350297 on Build GRCh38
rs763307752 Chr.13: 38350190-38350190 on Build GRCh38
rs765549058 Chr.13: 38350142-38350142 on Build GRCh38
rs751751774 Chr.13: 38349966-38349966 on Build GRCh38
rs537884186 Chr.13: 38350137-38350137 on Build GRCh38
rs774130782 Chr.13: 38350186-38350186 on Build GRCh38
rs766657101 Chr.13: 38350050-38350050 on Build GRCh38
rs756816236 Chr.13: 38350209-38350209 on Build GRCh38
rs747804187 Chr.13: 38349942-38349942 on Build GRCh38
rs549597630 Chr.13: 38350365-38350365 on Build GRCh38
rs369977032 Chr.13: 38350029-38350029 on Build GRCh38
rs769318522 Chr.13: 38350262-38350262 on Build GRCh38
rs755770689 Chr.13: 38350220-38350220 on Build GRCh38
rs1049980285 Chr.13: 38350207-38350221 on Build GRCh38
rs540254964 Chr.13: 38350266-38350266 on Build GRCh38
rs548239028 Chr.13: 38350371-38350371 on Build GRCh38
rs373348072 Chr.13: 38350234-38350234 on Build GRCh38
rs767662803 Chr.13: 38350045-38350045 on Build GRCh38
rs372075200 Chr.13: 38349978-38349978 on Build GRCh38
rs764320402 Chr.13: 38350261-38350261 on Build GRCh38
rs770679594 Chr.13: 38350090-38350090 on Build GRCh38
rs375915793 Chr.13: 38349996-38349996 on Build GRCh38
rs190964856 Chr.13: 38350395-38350395 on Build GRCh38
rs763360840 Chr.13: 38350248-38350248 on Build GRCh38
rs947503960 Chr.13: 38350104-38350104 on Build GRCh38
rs376995470 Chr.13: 38350245-38350245 on Build GRCh38
rs186157173 Chr.13: 38349972-38349972 on Build GRCh38
rs759473206 Chr.13: 38349957-38349957 on Build GRCh38
rs758346172 Chr.13: 38350173-38350174 on Build GRCh38
rs762022567 Chr.13: 38350038-38350038 on Build GRCh38
rs776505241 Chr.13: 38350241-38350241 on Build GRCh38
rs776417282 Chr.13: 38350001-38350001 on Build GRCh38
rs1004569067 Chr.13: 38350301-38350301 on Build GRCh38
rs765400008 Chr.13: 38350292-38350292 on Build GRCh38
rs923900190 Chr.13: 38350268-38350268 on Build GRCh38
rs981635087 Chr.13: 38350125-38350125 on Build GRCh38
rs368319157 Chr.13: 38349976-38349976 on Build GRCh38
rs2231331 Chr.13: 38349980-38349980 on Build GRCh38
rs771780280 Chr.13: 38349943-38349943 on Build GRCh38
rs149897266 Chr.13: 38350200-38350200 on Build GRCh38
rs914385696 Chr.13: 38350083-38350083 on Build GRCh38
rs750631146 Chr.13: 38350046-38350046 on Build GRCh38
rs532339006 Chr.13: 38350276-38350276 on Build GRCh38
rs146365514 Chr.13: 38350277-38350277 on Build GRCh38
rs764270321 Chr.13: 38349965-38349965 on Build GRCh38
rs755323915 Chr.13: 38350066-38350066 on Build GRCh38
rs753173366 Chr.13: 38350073-38350073 on Build GRCh38
rs746636859 Chr.13: 38349994-38349994 on Build GRCh38
rs971947242 Chr.13: 38350117-38350117 on Build GRCh38
rs371532681 Chr.13: 38349939-38349939 on Build GRCh38
rs971679990 Chr.13: 38350310-38350310 on Build GRCh38
rs955993278 Chr.13: 38350393-38350393 on Build GRCh38
rs772894975 Chr.13: 38349990-38349990 on Build GRCh38
rs759243065 Chr.13: 38350005-38350005 on Build GRCh38
rs1003567962 Chr.13: 38350357-38350357 on Build GRCh38
rs753905959 Chr.13: 38350274-38350274 on Build GRCh38
rs747152372 Chr.13: 38350180-38350180 on Build GRCh38
rs921948474 Chr.13: 38350141-38350141 on Build GRCh38
rs753228455 Chr.13: 38350145-38350145 on Build GRCh38
rs769906555 Chr.13: 38350014-38350014 on Build GRCh38
rs1015550046 Chr.13: 38350305-38350305 on Build GRCh38
rs761798482 Chr.13: 38350191-38350191 on Build GRCh38
rs776932458 Chr.13: 38350184-38350184 on Build GRCh38
rs754296728 Chr.13: 38350061-38350061 on Build GRCh38
rs766251135 Chr.13: 38350251-38350258 on Build GRCh38
rs753987963 Chr.13: 38350132-38350132 on Build GRCh38
rs749251369 Chr.13: 38350175-38350175 on Build GRCh38
rs371109857 Chr.13: 38350051-38350051 on Build GRCh38
rs370347538 Chr.13: 38350270-38350270 on Build GRCh38
rs537711805 Chr.13: 38350278-38350278 on Build GRCh38
rs200825211 Chr.13: 38350079-38350079 on Build GRCh38
rs747716159 Chr.13: 38349986-38349986 on Build GRCh38
rs770891635 Chr.13: 38350181-38350181 on Build GRCh38
rs775542915 Chr.13: 38350033-38350033 on Build GRCh38
rs143474592 Chr.13: 38350047-38350047 on Build GRCh38
rs772840051 Chr.13: 38349945-38349945 on Build GRCh38
rs746658555 Chr.13: 38350084-38350084 on Build GRCh38
rs750255338 Chr.13: 38350004-38350005 on Build GRCh38
rs2231332 Chr.13: 38350126-38350126 on Build GRCh38
rs568857824 Chr.13: 38350112-38350112 on Build GRCh38
rs766169319 Chr.13: 38349950-38349950 on Build GRCh38
rs201225534 Chr.13: 38350080-38350080 on Build GRCh38
rs776467711 Chr.13: 38349953-38349953 on Build GRCh38
rs774150714 Chr.13: 38350178-38350178 on Build GRCh38
rs771245954 Chr.13: 38350278-38350278 on Build GRCh38
rs766174458 Chr.13: 38350271-38350271 on Build GRCh38
rs758848845 Chr.13: 38350147-38350147 on Build GRCh38
rs768772362 Chr.13: 38350177-38350177 on Build GRCh38
rs780936086 Chr.13: 38350157-38350157 on Build GRCh38
rs755446233 Chr.13: 38349982-38349982 on Build GRCh38
rs373667763 Chr.13: 38350106-38350106 on Build GRCh38
rs756696496 Chr.13: 38350151-38350151 on Build GRCh38
rs778347398 Chr.13: 38350379-38350379 on Build GRCh38
rs762161291 Chr.13: 38350263-38350263 on Build GRCh38
rs367841465 Chr.13: 38350156-38350156 on Build GRCh38
rs754697828 Chr.13: 38350228-38350228 on Build GRCh38
rs372350077 Chr.13: 38350063-38350063 on Build GRCh38
rs1022637018 Chr.13: 38349960-38349960 on Build GRCh38
rs745721387 Chr.13: 38350105-38350105 on Build GRCh38
rs560460635 Chr.13: 38350275-38350275 on Build GRCh38
rs763813597 Chr.13: 38350172-38350172 on Build GRCh38
rs200144982 Chr.13: 38350229-38350229 on Build GRCh38
rs746099519 Chr.13: 38350243-38350243 on Build GRCh38
rs766899186 Chr.13: 38350215-38350215 on Build GRCh38
rs751134835 Chr.13: 38350149-38350149 on Build GRCh38
rs770658525 Chr.13: 38349999-38349999 on Build GRCh38
rs771551080 Chr.13: 38349987-38349987 on Build GRCh38
rs767068320 Chr.13: 38350265-38350265 on Build GRCh38
rs1048870044 Chr.13: 38350279-38350279 on Build GRCh38
rs569005658 Chr.13: 38350304-38350304 on Build GRCh38
rs751990771 Chr.13: 38350203-38350203 on Build GRCh38
rs762934137 Chr.13: 38350109-38350109 on Build GRCh38
rs745468378 Chr.13: 38350162-38350162 on Build GRCh38
rs770073293 Chr.13: 38350244-38350244 on Build GRCh38
rs750105365 Chr.13: 38350218-38350218 on Build GRCh38

View Full Product Details