Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 38354151-38354661 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UFM1
Entrez Gene ID
51569
Gene Name
ubiquitin fold modifier 1
Gene Aliases
BM-002, C13orf20
UniGene
Hs.643655
Gene Chromosome Location
Chr.13: 38349771-38363007 on Build GRCh38
Transcripts
NM_001286703.1 NM_001286704.1 NM_001286705.1 NM_001286706.1 NM_016617.3 NR_104584.1 NR_104585.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
111740 c.114_117+8delAGAAGTAAGTAC p.? Chr.13: 38354293-38354304 on Build GRCh38
3711059 c.83C>T p.P28L Chr.13: 38354262-38354262 on Build GRCh38
3711060 c.137C>T p.P46L Chr.13: 38354262-38354262 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs182883932 Chr.13: 38354369-38354369 on Build GRCh38
rs753573990 Chr.13: 38354324-38354324 on Build GRCh38
rs949127075 Chr.13: 38354486-38354486 on Build GRCh38
rs372162320 Chr.13: 38354520-38354520 on Build GRCh38
rs369434541 Chr.13: 38354236-38354236 on Build GRCh38
rs765937431 Chr.13: 38354323-38354323 on Build GRCh38
rs111800762 Chr.13: 38354528-38354528 on Build GRCh38
rs1000894375 Chr.13: 38354365-38354365 on Build GRCh38
rs201843668 Chr.13: 38354274-38354274 on Build GRCh38
rs1053727536 Chr.13: 38354531-38354531 on Build GRCh38
rs545292419 Chr.13: 38354543-38354544 on Build GRCh38
rs917631945 Chr.13: 38354622-38354622 on Build GRCh38
rs199968544 Chr.13: 38354252-38354252 on Build GRCh38
rs773823425 Chr.13: 38354312-38354312 on Build GRCh38
rs750744162 Chr.13: 38354214-38354214 on Build GRCh38
rs866212800 Chr.13: 38354254-38354255 on Build GRCh38
rs578161373 Chr.13: 38354537-38354537 on Build GRCh38
rs190302933 Chr.13: 38354333-38354333 on Build GRCh38
rs747779315 Chr.13: 38354311-38354312 on Build GRCh38
rs937531608 Chr.13: 38354450-38354450 on Build GRCh38
rs780280545 Chr.13: 38354243-38354243 on Build GRCh38
rs762225542 Chr.13: 38354295-38354295 on Build GRCh38
rs556904312 Chr.13: 38354352-38354352 on Build GRCh38
rs1047160891 Chr.13: 38354610-38354610 on Build GRCh38
rs550892896 Chr.13: 38354491-38354491 on Build GRCh38
rs778550463 Chr.13: 38354529-38354529 on Build GRCh38
rs902433514 Chr.13: 38354208-38354208 on Build GRCh38
rs919672246 Chr.13: 38354577-38354577 on Build GRCh38
rs755786615 Chr.13: 38354314-38354315 on Build GRCh38
rs114712162 Chr.13: 38354519-38354519 on Build GRCh38
rs998245291 Chr.13: 38354617-38354617 on Build GRCh38
rs759041867 Chr.13: 38354332-38354332 on Build GRCh38
rs955467037 Chr.13: 38354443-38354443 on Build GRCh38
rs780703900 Chr.13: 38354277-38354278 on Build GRCh38
rs901255568 Chr.13: 38354594-38354594 on Build GRCh38
rs769071773 Chr.13: 38354284-38354284 on Build GRCh38
rs772521766 Chr.13: 38354301-38354301 on Build GRCh38
rs898132448 Chr.13: 38354200-38354200 on Build GRCh38
rs1018399932 Chr.13: 38354341-38354341 on Build GRCh38
rs561098451 Chr.13: 38354241-38354241 on Build GRCh38
rs754445570 Chr.13: 38354255-38354256 on Build GRCh38
rs749474900 Chr.13: 38354254-38354254 on Build GRCh38
rs376938294 Chr.13: 38354305-38354305 on Build GRCh38
rs571531309 Chr.13: 38354434-38354434 on Build GRCh38
rs902150220 Chr.13: 38354518-38354518 on Build GRCh38
rs879695092 Chr.13: 38354503-38354503 on Build GRCh38
rs929647612 Chr.13: 38354209-38354209 on Build GRCh38
rs766842456 Chr.13: 38354334-38354334 on Build GRCh38
rs754437231 Chr.13: 38354521-38354521 on Build GRCh38
rs745991196 Chr.13: 38354217-38354217 on Build GRCh38
rs774523551 Chr.13: 38354292-38354292 on Build GRCh38
rs2231333 Chr.13: 38354370-38354370 on Build GRCh38
rs762215838 Chr.13: 38354346-38354346 on Build GRCh38
rs916364013 Chr.13: 38354452-38354452 on Build GRCh38
rs984359680 Chr.13: 38354372-38354372 on Build GRCh38
rs187520854 Chr.13: 38354422-38354422 on Build GRCh38
rs991603575 Chr.13: 38354451-38354451 on Build GRCh38
rs376449299 Chr.13: 38354342-38354342 on Build GRCh38
rs929622836 Chr.13: 38354589-38354589 on Build GRCh38
rs760940461 Chr.13: 38354260-38354260 on Build GRCh38
rs536436247 Chr.13: 38354535-38354535 on Build GRCh38
rs748118754 Chr.13: 38354188-38354188 on Build GRCh38
rs911043384 Chr.13: 38354487-38354487 on Build GRCh38
rs1009986041 Chr.13: 38354386-38354386 on Build GRCh38
rs537298900 Chr.13: 38354479-38354479 on Build GRCh38
rs760309470 Chr.13: 38354317-38354317 on Build GRCh38

View Full Product Details