Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 43879549-43880055 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC122
Entrez Gene ID
160857
Gene Name
coiled-coil domain containing 122
Gene Aliases
-
UniGene
Hs.170849
Gene Chromosome Location
Chr.13: 43821959-43879690 on Build GRCh38
Transcripts
NM_144974.3 XM_017020397.1 XM_017020398.1 XM_017020399.1 NM_001128303.1 NM_153218.2 XM_005266261.3 XM_006719766.3 XM_006719767.3 XM_011534933.2 XM_011534934.2 XM_011534935.2 XM_017020394.1

Gene Symbol
LACC1
Entrez Gene ID
144811
Gene Name
laccase domain containing 1
Gene Aliases
C13orf31
UniGene
Hs.210586
Gene Chromosome Location
Chr.13: 43878834-43893932 on Build GRCh38
Transcripts
NM_144974.3 XM_017020397.1 XM_017020398.1 XM_017020399.1 NM_001128303.1 NM_153218.2 XM_005266261.3 XM_006719766.3 XM_006719767.3 XM_011534933.2 XM_011534934.2 XM_011534935.2 XM_017020394.1

SNP Detail

SNP ID Chromosome Location
rs917816149 Chr.13: 43879691-43879691 on Build GRCh38
rs61959963 Chr.13: 43879793-43879793 on Build GRCh38
rs991416541 Chr.13: 43879848-43879848 on Build GRCh38
rs950134022 Chr.13: 43879652-43879652 on Build GRCh38
rs902073681 Chr.13: 43879966-43879966 on Build GRCh38
rs920447757 Chr.13: 43879829-43879829 on Build GRCh38
rs61959964 Chr.13: 43879803-43879803 on Build GRCh38
rs3816313 Chr.13: 43879763-43879763 on Build GRCh38
rs71099859 Chr.13: 43879820-43879837 on Build GRCh38
rs908339801 Chr.13: 43879937-43879937 on Build GRCh38
rs879437366 Chr.13: 43879791-43879795 on Build GRCh38
rs201087095 Chr.13: 43879798-43879802 on Build GRCh38
rs891418186 Chr.13: 43879970-43879970 on Build GRCh38
rs61959965 Chr.13: 43879805-43879805 on Build GRCh38
rs191723484 Chr.13: 43879998-43879998 on Build GRCh38
rs76474346 Chr.13: 43880022-43880022 on Build GRCh38
rs997074490 Chr.13: 43879700-43879700 on Build GRCh38
rs868293223 Chr.13: 43879795-43879795 on Build GRCh38
rs11147930 Chr.13: 43879769-43879769 on Build GRCh38
rs3816311 Chr.13: 43879647-43879647 on Build GRCh38
rs3816312 Chr.13: 43879737-43879737 on Build GRCh38
rs997754237 Chr.13: 43879987-43879987 on Build GRCh38
rs766851780 Chr.13: 43879795-43879827 on Build GRCh38
rs560678652 Chr.13: 43879689-43879689 on Build GRCh38
rs796409441 Chr.13: 43879810-43879818 on Build GRCh38
rs878998840 Chr.13: 43879817-43879818 on Build GRCh38
rs143579839 Chr.13: 43879949-43879949 on Build GRCh38
rs368722237 Chr.13: 43879800-43879800 on Build GRCh38
rs999425691 Chr.13: 43879672-43879672 on Build GRCh38
rs971824758 Chr.13: 43880030-43880030 on Build GRCh38
rs930521657 Chr.13: 43879846-43879846 on Build GRCh38
rs966599638 Chr.13: 43879778-43879778 on Build GRCh38
rs61959967 Chr.13: 43879813-43879813 on Build GRCh38
rs902125794 Chr.13: 43879579-43879579 on Build GRCh38
rs761099422 Chr.13: 43879819-43879827 on Build GRCh38
rs1045769451 Chr.13: 43879875-43879875 on Build GRCh38
rs1054530095 Chr.13: 43879911-43879911 on Build GRCh38
rs61959969 Chr.13: 43879827-43879827 on Build GRCh38
rs981803128 Chr.13: 43879794-43879794 on Build GRCh38
rs146153619 Chr.13: 43879796-43879800 on Build GRCh38
rs981115940 Chr.13: 43879842-43879842 on Build GRCh38
rs931156154 Chr.13: 43879893-43879893 on Build GRCh38
rs186929012 Chr.13: 43879901-43879901 on Build GRCh38
rs762617600 Chr.13: 43879626-43879626 on Build GRCh38
rs1028536154 Chr.13: 43879955-43879955 on Build GRCh38
rs920500490 Chr.13: 43879590-43879590 on Build GRCh38
rs543094170 Chr.13: 43879768-43879768 on Build GRCh38
rs1008974052 Chr.13: 43879996-43879996 on Build GRCh38
rs893319840 Chr.13: 43879934-43879934 on Build GRCh38
rs61959968 Chr.13: 43879818-43879818 on Build GRCh38
rs199946183 Chr.13: 43879761-43879769 on Build GRCh38
rs866755152 Chr.13: 43879836-43879836 on Build GRCh38
rs950096886 Chr.13: 43879867-43879867 on Build GRCh38
rs903253020 Chr.13: 43879891-43879891 on Build GRCh38
rs1046238975 Chr.13: 43879659-43879659 on Build GRCh38
rs564991602 Chr.13: 43879820-43879820 on Build GRCh38
rs1033170020 Chr.13: 43879674-43879674 on Build GRCh38
rs998079228 Chr.13: 43879580-43879580 on Build GRCh38
rs989320324 Chr.13: 43879764-43879764 on Build GRCh38
rs1028588579 Chr.13: 43879722-43879722 on Build GRCh38
rs903311407 Chr.13: 43879668-43879668 on Build GRCh38
rs915879298 Chr.13: 43879858-43879858 on Build GRCh38
rs61959966 Chr.13: 43879810-43879810 on Build GRCh38
rs930573920 Chr.13: 43879597-43879597 on Build GRCh38
rs893372258 Chr.13: 43879695-43879695 on Build GRCh38

View Full Product Details