Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 76880373-76880902 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD12
Entrez Gene ID
115207
Gene Name
potassium channel tetramerization domain containing 12
Gene Aliases
C13orf2, PFET1, PFETIN
UniGene
Hs.644125
Gene Chromosome Location
Chr.13: 76880169-76886405 on Build GRCh38
Transcripts
NM_138444.3

SNP Detail

SNP ID Chromosome Location
rs148084749 Chr.13: 76880412-76880412 on Build GRCh38
rs895934024 Chr.13: 76880773-76880773 on Build GRCh38
rs951673788 Chr.13: 76880627-76880627 on Build GRCh38
rs904183397 Chr.13: 76880728-76880728 on Build GRCh38
rs183958876 Chr.13: 76880779-76880779 on Build GRCh38
rs116539990 Chr.13: 76880654-76880654 on Build GRCh38
rs903261804 Chr.13: 76880505-76880505 on Build GRCh38
rs748262551 Chr.13: 76880540-76880540 on Build GRCh38
rs947328540 Chr.13: 76880720-76880720 on Build GRCh38
rs1026290799 Chr.13: 76880591-76880591 on Build GRCh38
rs1012918772 Chr.13: 76880774-76880774 on Build GRCh38
rs909865010 Chr.13: 76880641-76880641 on Build GRCh38
rs924403737 Chr.13: 76880737-76880737 on Build GRCh38
rs995007782 Chr.13: 76880634-76880634 on Build GRCh38
rs1023210539 Chr.13: 76880608-76880608 on Build GRCh38
rs531257467 Chr.13: 76880534-76880534 on Build GRCh38
rs9320 Chr.13: 76880414-76880414 on Build GRCh38
rs192504133 Chr.13: 76880746-76880746 on Build GRCh38
rs1026582757 Chr.13: 76880856-76880856 on Build GRCh38
rs964004184 Chr.13: 76880617-76880617 on Build GRCh38
rs992957405 Chr.13: 76880581-76880581 on Build GRCh38
rs771182232 Chr.13: 76880833-76880833 on Build GRCh38
rs200829866 Chr.13: 76880659-76880660 on Build GRCh38
rs553806228 Chr.13: 76880409-76880410 on Build GRCh38
rs1010290071 Chr.13: 76880456-76880456 on Build GRCh38
rs1051642432 Chr.13: 76880442-76880442 on Build GRCh38
rs141844727 Chr.13: 76880672-76880672 on Build GRCh38
rs975071057 Chr.13: 76880692-76880692 on Build GRCh38
rs895797093 Chr.13: 76880565-76880565 on Build GRCh38
rs551764973 Chr.13: 76880655-76880655 on Build GRCh38
rs1014346173 Chr.13: 76880760-76880760 on Build GRCh38
rs951654318 Chr.13: 76880808-76880808 on Build GRCh38
rs1012936603 Chr.13: 76880567-76880567 on Build GRCh38
rs551201078 Chr.13: 76880628-76880628 on Build GRCh38
rs949132832 Chr.13: 76880731-76880731 on Build GRCh38
rs934499562 Chr.13: 76880745-76880745 on Build GRCh38
rs921189652 Chr.13: 76880697-76880697 on Build GRCh38
rs984807700 Chr.13: 76880640-76880640 on Build GRCh38
rs187819077 Chr.13: 76880687-76880687 on Build GRCh38
rs868002121 Chr.13: 76880547-76880547 on Build GRCh38
rs895585999 Chr.13: 76880747-76880747 on Build GRCh38
rs1042988619 Chr.13: 76880727-76880727 on Build GRCh38
rs1043323033 Chr.13: 76880454-76880454 on Build GRCh38
rs964999832 Chr.13: 76880673-76880673 on Build GRCh38
rs567765832 Chr.13: 76880830-76880830 on Build GRCh38
rs565295690 Chr.13: 76880769-76880770 on Build GRCh38
rs982471014 Chr.13: 76880662-76880662 on Build GRCh38
rs1034098488 Chr.13: 76880570-76880570 on Build GRCh38
rs950916583 Chr.13: 76880870-76880870 on Build GRCh38
rs569341069 Chr.13: 76880630-76880630 on Build GRCh38

View Full Product Details