Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 95560042-95560545 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN10
Entrez Gene ID
9071
Gene Name
claudin 10
Gene Aliases
CPETRL3, OSP-L
UniGene
Hs.534377
Gene Chromosome Location
Chr.13: 95433588-95579756 on Build GRCh38
Transcripts
NM_001160100.1 NM_006984.4 NM_182848.3 XM_011521134.2 XM_017020843.1 XM_017020844.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3700428 c.282T>C p.F94F Chr.13: 95560199-95560199 on Build GRCh38
6629949 c.442G>T p.D148Y Chr.13: 95560441-95560441 on Build GRCh38
6554058 c.382G>T p.G128W Chr.13: 95560293-95560293 on Build GRCh38
1201301 c.364A>C p.I122L Chr.13: 95560275-95560275 on Build GRCh38
5815508 c.440C>T p.P147L Chr.13: 95560445-95560445 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201430734 Chr.13: 95560479-95560479 on Build GRCh38
rs758369755 Chr.13: 95560476-95560476 on Build GRCh38
rs771988001 Chr.13: 95560269-95560269 on Build GRCh38
rs374187735 Chr.13: 95560430-95560430 on Build GRCh38
rs778696030 Chr.13: 95560264-95560264 on Build GRCh38
rs772986808 Chr.13: 95560174-95560174 on Build GRCh38
rs11537907 Chr.13: 95560226-95560226 on Build GRCh38
rs766827012 Chr.13: 95560453-95560453 on Build GRCh38
rs756346152 Chr.13: 95560235-95560235 on Build GRCh38
rs887862325 Chr.13: 95560116-95560116 on Build GRCh38
rs768046859 Chr.13: 95560441-95560441 on Build GRCh38
rs769511383 Chr.13: 95560511-95560511 on Build GRCh38
rs576678124 Chr.13: 95560316-95560316 on Build GRCh38
rs928489675 Chr.13: 95560145-95560145 on Build GRCh38
rs374567220 Chr.13: 95560229-95560229 on Build GRCh38
rs752846602 Chr.13: 95560474-95560474 on Build GRCh38
rs34748844 Chr.13: 95560184-95560185 on Build GRCh38
rs774703793 Chr.13: 95560321-95560321 on Build GRCh38
rs370623600 Chr.13: 95560320-95560320 on Build GRCh38
rs760241596 Chr.13: 95560090-95560090 on Build GRCh38
rs200161653 Chr.13: 95560108-95560108 on Build GRCh38
rs747316537 Chr.13: 95560223-95560223 on Build GRCh38
rs773143926 Chr.13: 95560274-95560274 on Build GRCh38
rs760346005 Chr.13: 95560282-95560282 on Build GRCh38
rs781105813 Chr.13: 95560501-95560501 on Build GRCh38
rs758548545 Chr.13: 95560170-95560170 on Build GRCh38
rs776163122 Chr.13: 95560294-95560294 on Build GRCh38
rs373345015 Chr.13: 95560356-95560356 on Build GRCh38
rs745770511 Chr.13: 95560460-95560460 on Build GRCh38
rs1049064936 Chr.13: 95560414-95560414 on Build GRCh38
rs368714235 Chr.13: 95560447-95560447 on Build GRCh38
rs1044483008 Chr.13: 95560490-95560490 on Build GRCh38
rs372666058 Chr.13: 95560273-95560273 on Build GRCh38
rs369800452 Chr.13: 95560261-95560261 on Build GRCh38
rs202098827 Chr.13: 95560224-95560224 on Build GRCh38
rs749359616 Chr.13: 95560241-95560241 on Build GRCh38
rs755231416 Chr.13: 95560464-95560464 on Build GRCh38
rs985851504 Chr.13: 95560304-95560304 on Build GRCh38
rs767311119 Chr.13: 95560087-95560087 on Build GRCh38
rs113721014 Chr.13: 95560239-95560239 on Build GRCh38
rs768504036 Chr.13: 95560159-95560159 on Build GRCh38
rs770729064 Chr.13: 95560289-95560289 on Build GRCh38
rs142536284 Chr.13: 95560201-95560201 on Build GRCh38
rs781359177 Chr.13: 95560137-95560137 on Build GRCh38
rs770886300 Chr.13: 95560421-95560421 on Build GRCh38
rs387907413 Chr.13: 95560419-95560419 on Build GRCh38
rs747968404 Chr.13: 95560377-95560377 on Build GRCh38
rs375779020 Chr.13: 95560120-95560120 on Build GRCh38
rs766479345 Chr.13: 95560342-95560342 on Build GRCh38
rs765345833 Chr.13: 95560163-95560163 on Build GRCh38
rs149697610 Chr.13: 95560461-95560461 on Build GRCh38
rs371976897 Chr.13: 95560167-95560167 on Build GRCh38
rs374491325 Chr.13: 95560088-95560088 on Build GRCh38
rs370703602 Chr.13: 95560393-95560393 on Build GRCh38
rs761005313 Chr.13: 95560451-95560451 on Build GRCh38
rs750580303 Chr.13: 95560231-95560231 on Build GRCh38
rs761482439 Chr.13: 95560166-95560166 on Build GRCh38
rs113065528 Chr.13: 95560328-95560328 on Build GRCh38
rs756292784 Chr.13: 95560335-95560335 on Build GRCh38
rs745456011 Chr.13: 95560509-95560509 on Build GRCh38
rs375663820 Chr.13: 95560182-95560182 on Build GRCh38
rs769862960 Chr.13: 95560140-95560140 on Build GRCh38
rs140177098 Chr.13: 95560180-95560180 on Build GRCh38
rs115147677 Chr.13: 95560202-95560202 on Build GRCh38
rs993544858 Chr.13: 95560083-95560083 on Build GRCh38
rs542466700 Chr.13: 95560358-95560358 on Build GRCh38
rs766044595 Chr.13: 95560092-95560092 on Build GRCh38
rs116119435 Chr.13: 95560431-95560431 on Build GRCh38
rs762262906 Chr.13: 95560434-95560434 on Build GRCh38
rs754139363 Chr.13: 95560346-95560346 on Build GRCh38
rs376283811 Chr.13: 95560359-95560359 on Build GRCh38
rs746988094 Chr.13: 95560491-95560491 on Build GRCh38
rs774156411 Chr.13: 95560164-95560164 on Build GRCh38
rs781569954 Chr.13: 95560230-95560230 on Build GRCh38
rs991291165 Chr.13: 95560506-95560506 on Build GRCh38
rs748587625 Chr.13: 95560238-95560238 on Build GRCh38
rs34449921 Chr.13: 95560139-95560139 on Build GRCh38
rs897720515 Chr.13: 95560071-95560071 on Build GRCh38
rs376861377 Chr.13: 95560303-95560303 on Build GRCh38
rs540947662 Chr.13: 95560107-95560107 on Build GRCh38
rs776122725 Chr.13: 95560429-95560429 on Build GRCh38
rs749196318 Chr.13: 95560158-95560158 on Build GRCh38
rs765170859 Chr.13: 95560466-95560466 on Build GRCh38
rs368080206 Chr.13: 95560146-95560146 on Build GRCh38
rs79906565 Chr.13: 95560401-95560401 on Build GRCh38
rs982641603 Chr.13: 95560470-95560470 on Build GRCh38
rs767815108 Chr.13: 95560332-95560332 on Build GRCh38
rs753249422 Chr.13: 95560094-95560094 on Build GRCh38

View Full Product Details