Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 101720771-101721280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF14
Entrez Gene ID
2259
Gene Name
fibroblast growth factor 14
Gene Aliases
FGF-14, FHF-4, FHF4, SCA27
UniGene
Hs.508616
Gene Chromosome Location
Chr.13: 101720855-102402428 on Build GRCh38
Transcripts
NM_001321931.1 NM_001321932.1 NM_001321933.1 NM_001321934.1 NM_001321935.1 NM_001321936.1 NM_001321938.1 NM_001321939.1 NM_001321940.1 NM_001321941.1 NM_001321942.1 NM_001321943.1 NM_001321944.1 NM_001321945.1 NM_001321946.1 NM_001321947.1 NM_001321948.1 NM_001321949.1 NM_004115.3 NM_175929.2 NM_001271754.1 XM_005254100.4

Gene Symbol
ITGBL1
Entrez Gene ID
9358
Gene Name
integrin subunit beta like 1
Gene Aliases
OSCP, TIED
UniGene
Hs.696554
Gene Chromosome Location
Chr.13: 101452593-101720857 on Build GRCh38
Transcripts
NM_001321931.1 NM_001321932.1 NM_001321933.1 NM_001321934.1 NM_001321935.1 NM_001321936.1 NM_001321938.1 NM_001321939.1 NM_001321940.1 NM_001321941.1 NM_001321942.1 NM_001321943.1 NM_001321944.1 NM_001321945.1 NM_001321946.1 NM_001321947.1 NM_001321948.1 NM_001321949.1 NM_004115.3 NM_175929.2 NM_001271754.1 XM_005254100.4

SNP Detail

SNP ID Chromosome Location
rs955955746 Chr.13: 101721246-101721246 on Build GRCh38
rs1016317978 Chr.13: 101721073-101721073 on Build GRCh38
rs915601760 Chr.13: 101720832-101720832 on Build GRCh38
rs15608 Chr.13: 101721213-101721213 on Build GRCh38
rs1044559459 Chr.13: 101720980-101720980 on Build GRCh38
rs1026513793 Chr.13: 101721148-101721148 on Build GRCh38
rs533531542 Chr.13: 101720971-101720971 on Build GRCh38
rs948387320 Chr.13: 101720837-101720837 on Build GRCh38
rs994083744 Chr.13: 101721120-101721120 on Build GRCh38
rs141424596 Chr.13: 101721165-101721167 on Build GRCh38
rs962503853 Chr.13: 101720884-101720884 on Build GRCh38
rs973905191 Chr.13: 101720893-101720893 on Build GRCh38
rs767840771 Chr.13: 101720910-101720911 on Build GRCh38
rs909335523 Chr.13: 101720868-101720868 on Build GRCh38
rs1022662054 Chr.13: 101721169-101721169 on Build GRCh38
rs566495170 Chr.13: 101720905-101720905 on Build GRCh38
rs952294065 Chr.13: 101721151-101721151 on Build GRCh38
rs1043828363 Chr.13: 101721038-101721038 on Build GRCh38
rs761322568 Chr.13: 101721059-101721059 on Build GRCh38
rs984935410 Chr.13: 101720827-101720827 on Build GRCh38
rs529644559 Chr.13: 101721090-101721090 on Build GRCh38
rs762640115 Chr.13: 101721174-101721175 on Build GRCh38
rs57922134 Chr.13: 101720920-101720920 on Build GRCh38
rs555313981 Chr.13: 101721008-101721008 on Build GRCh38
rs983923846 Chr.13: 101720809-101720809 on Build GRCh38
rs934493797 Chr.13: 101720953-101720953 on Build GRCh38
rs946843853 Chr.13: 101721023-101721023 on Build GRCh38
rs886696133 Chr.13: 101721223-101721223 on Build GRCh38
rs558584910 Chr.13: 101721080-101721083 on Build GRCh38
rs772940026 Chr.13: 101721164-101721165 on Build GRCh38
rs142480530 Chr.13: 101721205-101721207 on Build GRCh38
rs771196126 Chr.13: 101721201-101721202 on Build GRCh38
rs1051051650 Chr.13: 101720911-101720914 on Build GRCh38
rs577910610 Chr.13: 101721149-101721149 on Build GRCh38
rs987122267 Chr.13: 101721153-101721153 on Build GRCh38
rs189407491 Chr.13: 101721037-101721037 on Build GRCh38
rs766695039 Chr.13: 101721195-101721196 on Build GRCh38
rs950774008 Chr.13: 101721065-101721065 on Build GRCh38
rs1019209761 Chr.13: 101721118-101721118 on Build GRCh38
rs543783809 Chr.13: 101720937-101720937 on Build GRCh38
rs1052935608 Chr.13: 101720968-101720968 on Build GRCh38
rs1046197 Chr.13: 101721219-101721219 on Build GRCh38
rs1003522096 Chr.13: 101721001-101721001 on Build GRCh38
rs892946304 Chr.13: 101720989-101720989 on Build GRCh38
rs912516724 Chr.13: 101721155-101721155 on Build GRCh38
rs951799231 Chr.13: 101720805-101720805 on Build GRCh38
rs142833116 Chr.13: 101721119-101721119 on Build GRCh38
rs963475614 Chr.13: 101721087-101721087 on Build GRCh38
rs981360720 Chr.13: 101720848-101720848 on Build GRCh38
rs984545304 Chr.13: 101721164-101721164 on Build GRCh38
rs906007712 Chr.13: 101721216-101721216 on Build GRCh38
rs775124846 Chr.13: 101721177-101721177 on Build GRCh38
rs386773958 Chr.13: 101721213-101721219 on Build GRCh38
rs764687981 Chr.13: 101721088-101721088 on Build GRCh38
rs912197341 Chr.13: 101720918-101720918 on Build GRCh38
rs559888067 Chr.13: 101720941-101720942 on Build GRCh38
rs886049931 Chr.13: 101721070-101721070 on Build GRCh38
rs914298159 Chr.13: 101721181-101721181 on Build GRCh38
rs759620996 Chr.13: 101721219-101721222 on Build GRCh38
rs747506157 Chr.13: 101721222-101721223 on Build GRCh38

View Full Product Details