Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 110430321-110430825 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL4A2
Entrez Gene ID
1284
Gene Name
collagen type IV alpha 2
Gene Aliases
ICH, POREN2
UniGene
Hs.508716
Gene Chromosome Location
Chr.13: 110307284-110513027 on Build GRCh38
Transcripts

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6633157 c.600C>T p.R200R Chr.13: 110430559-110430559 on Build GRCh38
4501251 c.583C>T p.Q195* Chr.13: 110430434-110430434 on Build GRCh38
6015406 c.587G>A p.G196E Chr.13: 110430546-110430546 on Build GRCh38
7207944 c.592C>T p.P198S Chr.13: 110430551-110430551 on Build GRCh38
5519231 c.595G>A p.G199S Chr.13: 110430554-110430554 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752167352 Chr.13: 110430475-110430475 on Build GRCh38
rs940255297 Chr.13: 110430652-110430652 on Build GRCh38
rs74124319 Chr.13: 110430553-110430553 on Build GRCh38
rs552174744 Chr.13: 110430588-110430588 on Build GRCh38
rs760161017 Chr.13: 110430605-110430605 on Build GRCh38
rs763421457 Chr.13: 110430466-110430466 on Build GRCh38
rs775479003 Chr.13: 110430428-110430428 on Build GRCh38
rs777328139 Chr.13: 110430444-110430444 on Build GRCh38
rs763674789 Chr.13: 110430494-110430494 on Build GRCh38
rs375316442 Chr.13: 110430620-110430620 on Build GRCh38
rs751522074 Chr.13: 110430770-110430770 on Build GRCh38
rs766319507 Chr.13: 110430764-110430764 on Build GRCh38
rs367972578 Chr.13: 110430458-110430458 on Build GRCh38
rs150748464 Chr.13: 110430695-110430695 on Build GRCh38
rs199883212 Chr.13: 110430773-110430773 on Build GRCh38
rs200660165 Chr.13: 110430496-110430496 on Build GRCh38
rs768958595 Chr.13: 110430440-110430440 on Build GRCh38
rs537477351 Chr.13: 110430649-110430649 on Build GRCh38
rs764728927 Chr.13: 110430741-110430741 on Build GRCh38
rs562071212 Chr.13: 110430430-110430430 on Build GRCh38
rs755717707 Chr.13: 110430396-110430396 on Build GRCh38
rs1035460729 Chr.13: 110430594-110430594 on Build GRCh38
rs764962591 Chr.13: 110430645-110430645 on Build GRCh38
rs781472691 Chr.13: 110430659-110430659 on Build GRCh38
rs749012559 Chr.13: 110430408-110430408 on Build GRCh38
rs758220603 Chr.13: 110430545-110430545 on Build GRCh38
rs778610185 Chr.13: 110430522-110430522 on Build GRCh38
rs761817376 Chr.13: 110430541-110430541 on Build GRCh38
rs990636960 Chr.13: 110430619-110430619 on Build GRCh38
rs752656990 Chr.13: 110430790-110430790 on Build GRCh38
rs760073315 Chr.13: 110430722-110430722 on Build GRCh38
rs1049951254 Chr.13: 110430703-110430703 on Build GRCh38
rs888417743 Chr.13: 110430792-110430792 on Build GRCh38
rs369621560 Chr.13: 110430556-110430556 on Build GRCh38
rs750148617 Chr.13: 110430753-110430753 on Build GRCh38
rs770721562 Chr.13: 110430413-110430413 on Build GRCh38
rs889587710 Chr.13: 110430377-110430377 on Build GRCh38
rs988508564 Chr.13: 110430638-110430638 on Build GRCh38
rs750949469 Chr.13: 110430367-110430367 on Build GRCh38
rs766732733 Chr.13: 110430592-110430592 on Build GRCh38
rs202050913 Chr.13: 110430651-110430651 on Build GRCh38
rs748263528 Chr.13: 110430661-110430661 on Build GRCh38
rs754183214 Chr.13: 110430650-110430650 on Build GRCh38
rs762255585 Chr.13: 110430443-110430443 on Build GRCh38
rs749561811 Chr.13: 110430699-110430699 on Build GRCh38
rs776068639 Chr.13: 110430724-110430724 on Build GRCh38
rs367748098 Chr.13: 110430386-110430386 on Build GRCh38
rs755628265 Chr.13: 110430488-110430488 on Build GRCh38
rs62621885 Chr.13: 110430425-110430425 on Build GRCh38
rs748639011 Chr.13: 110430561-110430562 on Build GRCh38
rs201716258 Chr.13: 110430567-110430567 on Build GRCh38
rs754507343 Chr.13: 110430370-110430370 on Build GRCh38
rs190117538 Chr.13: 110430427-110430427 on Build GRCh38
rs959464417 Chr.13: 110430569-110430569 on Build GRCh38
rs763569120 Chr.13: 110430723-110430723 on Build GRCh38
rs376087853 Chr.13: 110430680-110430680 on Build GRCh38
rs116714390 Chr.13: 110430729-110430729 on Build GRCh38
rs757038559 Chr.13: 110430517-110430517 on Build GRCh38
rs376017335 Chr.13: 110430364-110430364 on Build GRCh38
rs922722829 Chr.13: 110430714-110430714 on Build GRCh38
rs572328853 Chr.13: 110430388-110430388 on Build GRCh38
rs748422794 Chr.13: 110430558-110430558 on Build GRCh38
rs756907222 Chr.13: 110430639-110430639 on Build GRCh38
rs766933562 Chr.13: 110430469-110430469 on Build GRCh38
rs774831276 Chr.13: 110430706-110430706 on Build GRCh38
rs754575068 Chr.13: 110430771-110430771 on Build GRCh38
rs762651337 Chr.13: 110430754-110430754 on Build GRCh38
rs774849640 Chr.13: 110430600-110430600 on Build GRCh38
rs765589060 Chr.13: 110430457-110430457 on Build GRCh38
rs778894700 Chr.13: 110430422-110430422 on Build GRCh38
rs749736969 Chr.13: 110430535-110430538 on Build GRCh38
rs777437302 Chr.13: 110430399-110430399 on Build GRCh38
rs373229609 Chr.13: 110430702-110430702 on Build GRCh38
rs74124320 Chr.13: 110430635-110430635 on Build GRCh38
rs372514987 Chr.13: 110430646-110430646 on Build GRCh38
rs750358950 Chr.13: 110430554-110430554 on Build GRCh38

View Full Product Details