Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 21499228-21499737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
METTL3
Entrez Gene ID
56339
Gene Name
methyltransferase like 3
Gene Aliases
IME4, M6A, MT-A70, Spo8
UniGene
Hs.168799
Gene Chromosome Location
Chr.14: 21498131-21511375 on Build GRCh38
Transcripts
NM_019852.4 XM_006720206.3 XM_011536968.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4701833 c.1433A>G p.H478R Chr.14: 21499511-21499511 on Build GRCh38
469795 c.1441G>A p.E481K Chr.14: 21499503-21499503 on Build GRCh38
7163317 c.1516G>A p.E506K Chr.14: 21499308-21499308 on Build GRCh38
1607490 c.1452+1_1452+11delGTGAGCAGCAG p.? Chr.14: 21499481-21499491 on Build GRCh38
3494961 c.1472C>T p.P491L Chr.14: 21499352-21499352 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755054613 Chr.14: 21499327-21499327 on Build GRCh38
rs768514747 Chr.14: 21499396-21499396 on Build GRCh38
rs772781305 Chr.14: 21499627-21499627 on Build GRCh38
rs778683350 Chr.14: 21499372-21499372 on Build GRCh38
rs140146410 Chr.14: 21499546-21499546 on Build GRCh38
rs761585539 Chr.14: 21499273-21499274 on Build GRCh38
rs3168879 Chr.14: 21499443-21499443 on Build GRCh38
rs761801841 Chr.14: 21499591-21499591 on Build GRCh38
rs760040385 Chr.14: 21499634-21499634 on Build GRCh38
rs746954678 Chr.14: 21499602-21499602 on Build GRCh38
rs372480303 Chr.14: 21499470-21499470 on Build GRCh38
rs746493972 Chr.14: 21499444-21499444 on Build GRCh38
rs540836022 Chr.14: 21499635-21499635 on Build GRCh38
rs1012499313 Chr.14: 21499645-21499645 on Build GRCh38
rs939768765 Chr.14: 21499279-21499279 on Build GRCh38
rs774768803 Chr.14: 21499540-21499540 on Build GRCh38
rs985707468 Chr.14: 21499431-21499432 on Build GRCh38
rs770760253 Chr.14: 21499255-21499255 on Build GRCh38
rs746890631 Chr.14: 21499420-21499420 on Build GRCh38
rs771412995 Chr.14: 21499274-21499274 on Build GRCh38
rs752936315 Chr.14: 21499348-21499348 on Build GRCh38
rs779995154 Chr.14: 21499592-21499592 on Build GRCh38
rs146603540 Chr.14: 21499522-21499522 on Build GRCh38
rs772993260 Chr.14: 21499574-21499575 on Build GRCh38
rs775075020 Chr.14: 21499278-21499278 on Build GRCh38
rs201104563 Chr.14: 21499606-21499606 on Build GRCh38
rs771220600 Chr.14: 21499631-21499631 on Build GRCh38
rs201068854 Chr.14: 21499293-21499293 on Build GRCh38
rs768415429 Chr.14: 21499285-21499285 on Build GRCh38
rs766854082 Chr.14: 21499301-21499301 on Build GRCh38
rs760807140 Chr.14: 21499646-21499646 on Build GRCh38
rs977266253 Chr.14: 21499315-21499315 on Build GRCh38
rs751656594 Chr.14: 21499306-21499306 on Build GRCh38
rs149041981 Chr.14: 21499560-21499560 on Build GRCh38
rs760158490 Chr.14: 21499284-21499284 on Build GRCh38
rs569532008 Chr.14: 21499552-21499552 on Build GRCh38
rs557657730 Chr.14: 21499642-21499642 on Build GRCh38
rs376791809 Chr.14: 21499409-21499409 on Build GRCh38
rs745848069 Chr.14: 21499272-21499272 on Build GRCh38
rs767615017 Chr.14: 21499548-21499548 on Build GRCh38
rs946965341 Chr.14: 21499520-21499520 on Build GRCh38
rs749643805 Chr.14: 21499629-21499629 on Build GRCh38
rs774309591 Chr.14: 21499577-21499577 on Build GRCh38
rs768307011 Chr.14: 21499452-21499452 on Build GRCh38
rs1015449570 Chr.14: 21499657-21499657 on Build GRCh38
rs917007264 Chr.14: 21499688-21499688 on Build GRCh38
rs750896377 Chr.14: 21499295-21499295 on Build GRCh38
rs992617811 Chr.14: 21499667-21499667 on Build GRCh38
rs77798186 Chr.14: 21499471-21499471 on Build GRCh38
rs750226006 Chr.14: 21499576-21499576 on Build GRCh38
rs774880730 Chr.14: 21499633-21499633 on Build GRCh38
rs781369441 Chr.14: 21499333-21499333 on Build GRCh38
rs181527844 Chr.14: 21499411-21499411 on Build GRCh38
rs190732474 Chr.14: 21499298-21499298 on Build GRCh38
rs984419689 Chr.14: 21499655-21499655 on Build GRCh38
rs1053310517 Chr.14: 21499440-21499440 on Build GRCh38
rs1008963979 Chr.14: 21499486-21499486 on Build GRCh38
rs765287812 Chr.14: 21499385-21499386 on Build GRCh38
rs571760146 Chr.14: 21499624-21499624 on Build GRCh38
rs952670013 Chr.14: 21499639-21499639 on Build GRCh38
rs934416154 Chr.14: 21499549-21499549 on Build GRCh38
rs3168880 Chr.14: 21499294-21499294 on Build GRCh38
rs779920049 Chr.14: 21499416-21499416 on Build GRCh38
rs752850121 Chr.14: 21499551-21499551 on Build GRCh38
rs576831643 Chr.14: 21499692-21499692 on Build GRCh38
rs763295671 Chr.14: 21499513-21499513 on Build GRCh38
rs961194038 Chr.14: 21499663-21499663 on Build GRCh38
rs747044799 Chr.14: 21499258-21499258 on Build GRCh38
rs776386256 Chr.14: 21499463-21499463 on Build GRCh38
rs577897694 Chr.14: 21499673-21499673 on Build GRCh38
rs776474375 Chr.14: 21499292-21499292 on Build GRCh38
rs757005746 Chr.14: 21499358-21499358 on Build GRCh38
rs373137461 Chr.14: 21499622-21499622 on Build GRCh38

View Full Product Details