Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24268427-24268932 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RABGGTA
Entrez Gene ID
5875
Gene Name
Rab geranylgeranyltransferase alpha subunit
Gene Aliases
PTAR3
UniGene
Hs.377992
Gene Chromosome Location
Chr.14: 24265538-24271627 on Build GRCh38
Transcripts
NM_004581.5 NM_182836.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5576989 c.861G>A p.W287* Chr.14: 24268764-24268764 on Build GRCh38
4050244 c.956G>A p.R319H Chr.14: 24268564-24268564 on Build GRCh38
7281103 c.992G>A p.C331Y Chr.14: 24268528-24268528 on Build GRCh38
469858 c.919G>C p.A307P Chr.14: 24268601-24268601 on Build GRCh38
6032765 c.993C>T p.C331C Chr.14: 24268527-24268527 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759771319 Chr.14: 24268708-24268708 on Build GRCh38
rs758675574 Chr.14: 24268496-24268496 on Build GRCh38
rs757517248 Chr.14: 24268457-24268457 on Build GRCh38
rs754962568 Chr.14: 24268599-24268599 on Build GRCh38
rs758761401 Chr.14: 24268589-24268589 on Build GRCh38
rs779795973 Chr.14: 24268740-24268740 on Build GRCh38
rs369745480 Chr.14: 24268565-24268565 on Build GRCh38
rs750403065 Chr.14: 24268877-24268877 on Build GRCh38
rs757196611 Chr.14: 24268609-24268609 on Build GRCh38
rs541650456 Chr.14: 24268505-24268505 on Build GRCh38
rs762289778 Chr.14: 24268681-24268681 on Build GRCh38
rs777796239 Chr.14: 24268606-24268606 on Build GRCh38
rs750718192 Chr.14: 24268489-24268489 on Build GRCh38
rs751790243 Chr.14: 24268707-24268707 on Build GRCh38
rs767771354 Chr.14: 24268831-24268831 on Build GRCh38
rs758152257 Chr.14: 24268731-24268731 on Build GRCh38
rs921497285 Chr.14: 24268793-24268793 on Build GRCh38
rs563239776 Chr.14: 24268627-24268627 on Build GRCh38
rs766731749 Chr.14: 24268698-24268698 on Build GRCh38
rs888540606 Chr.14: 24268892-24268892 on Build GRCh38
rs779885726 Chr.14: 24268885-24268885 on Build GRCh38
rs779621614 Chr.14: 24268645-24268645 on Build GRCh38
rs372667304 Chr.14: 24268563-24268563 on Build GRCh38
rs564621166 Chr.14: 24268527-24268527 on Build GRCh38
rs55836004 Chr.14: 24268735-24268735 on Build GRCh38
rs751369800 Chr.14: 24268886-24268886 on Build GRCh38
rs780633413 Chr.14: 24268746-24268746 on Build GRCh38
rs373544140 Chr.14: 24268562-24268562 on Build GRCh38
rs528274570 Chr.14: 24268775-24268775 on Build GRCh38
rs565810680 Chr.14: 24268743-24268743 on Build GRCh38
rs1015483361 Chr.14: 24268566-24268566 on Build GRCh38
rs757205231 Chr.14: 24268715-24268715 on Build GRCh38
rs780338638 Chr.14: 24268504-24268504 on Build GRCh38
rs749223411 Chr.14: 24268608-24268608 on Build GRCh38
rs188124827 Chr.14: 24268728-24268728 on Build GRCh38
rs767542837 Chr.14: 24268710-24268710 on Build GRCh38
rs369776993 Chr.14: 24268526-24268526 on Build GRCh38
rs770420524 Chr.14: 24268684-24268684 on Build GRCh38
rs183337992 Chr.14: 24268580-24268580 on Build GRCh38
rs758313582 Chr.14: 24268883-24268883 on Build GRCh38
rs373655263 Chr.14: 24268573-24268573 on Build GRCh38
rs767929445 Chr.14: 24268759-24268759 on Build GRCh38
rs1013066914 Chr.14: 24268485-24268485 on Build GRCh38
rs549174526 Chr.14: 24268899-24268899 on Build GRCh38
rs751704583 Chr.14: 24268591-24268591 on Build GRCh38
rs915451075 Chr.14: 24268624-24268624 on Build GRCh38
rs760686607 Chr.14: 24268869-24268869 on Build GRCh38
rs776966441 Chr.14: 24268570-24268570 on Build GRCh38
rs768346085 Chr.14: 24268742-24268742 on Build GRCh38
rs528916642 Chr.14: 24268693-24268693 on Build GRCh38
rs765178760 Chr.14: 24268872-24268872 on Build GRCh38
rs764686950 Chr.14: 24268564-24268564 on Build GRCh38
rs774754408 Chr.14: 24268816-24268816 on Build GRCh38
rs778752690 Chr.14: 24268720-24268720 on Build GRCh38
rs754231558 Chr.14: 24268455-24268455 on Build GRCh38
rs773808816 Chr.14: 24268781-24268781 on Build GRCh38
rs778734234 Chr.14: 24268622-24268622 on Build GRCh38
rs770590924 Chr.14: 24268533-24268533 on Build GRCh38
rs774100217 Chr.14: 24268546-24268546 on Build GRCh38
rs752808767 Chr.14: 24268711-24268711 on Build GRCh38
rs988300913 Chr.14: 24268676-24268676 on Build GRCh38
rs768095277 Chr.14: 24268664-24268664 on Build GRCh38
rs371226710 Chr.14: 24268795-24268795 on Build GRCh38
rs1013918845 Chr.14: 24268483-24268483 on Build GRCh38
rs747628262 Chr.14: 24268748-24268748 on Build GRCh38
rs771714707 Chr.14: 24268638-24268638 on Build GRCh38
rs773717077 Chr.14: 24268686-24268686 on Build GRCh38
rs771694585 Chr.14: 24268550-24268550 on Build GRCh38
rs202052623 Chr.14: 24268463-24268463 on Build GRCh38
rs780840337 Chr.14: 24268905-24268905 on Build GRCh38
rs754591709 Chr.14: 24268897-24268897 on Build GRCh38
rs373123048 Chr.14: 24268688-24268688 on Build GRCh38
rs868382861 Chr.14: 24268639-24268639 on Build GRCh38
rs944324847 Chr.14: 24268724-24268724 on Build GRCh38
rs770489351 Chr.14: 24268638-24268638 on Build GRCh38
rs374750633 Chr.14: 24268814-24268814 on Build GRCh38
rs759734789 Chr.14: 24268823-24268823 on Build GRCh38
rs746563340 Chr.14: 24268663-24268663 on Build GRCh38
rs368854695 Chr.14: 24268819-24268819 on Build GRCh38
rs377451545 Chr.14: 24268741-24268741 on Build GRCh38
rs370715682 Chr.14: 24268590-24268590 on Build GRCh38
rs752720389 Chr.14: 24268836-24268836 on Build GRCh38
rs773686718 Chr.14: 24268676-24268686 on Build GRCh38
rs776185796 Chr.14: 24268668-24268668 on Build GRCh38
rs771460926 Chr.14: 24268809-24268809 on Build GRCh38
rs533618121 Chr.14: 24268572-24268572 on Build GRCh38
rs745568957 Chr.14: 24268549-24268549 on Build GRCh38

View Full Product Details