Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24370186-24370691 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NFATC4
Entrez Gene ID
4776
Gene Name
nuclear factor of activated T-cells 4
Gene Aliases
NF-AT3, NF-ATC4, NFAT3
UniGene
Hs.77810
Gene Chromosome Location
Chr.14: 24366911-24379604 on Build GRCh38
Transcripts
NM_001136022.2 NM_001198965.1 NM_001198966.2 NM_001198967.2 NM_001288802.1 NM_001320043.1 NM_004554.4 XM_011536797.2 XM_011536798.1 XM_011536799.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6725512 c.1017G>T p.R339R Chr.14: 24370319-24370319 on Build GRCh38
3885956 c.1287A>G p.G429G Chr.14: 24370496-24370496 on Build GRCh38
5566049 c.1069C>T p.R357W Chr.14: 24370503-24370503 on Build GRCh38
4564611 c.1110_1111GG>AA p.E371K Chr.14: 24370508-24370509 on Build GRCh38
6075672 c.1310G>A p.G437D Chr.14: 24370519-24370519 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs955445198 Chr.14: 24370255-24370255 on Build GRCh38
rs1007569041 Chr.14: 24370462-24370462 on Build GRCh38
rs2229310 Chr.14: 24370550-24370550 on Build GRCh38
rs981442583 Chr.14: 24370317-24370317 on Build GRCh38
rs769441467 Chr.14: 24370552-24370552 on Build GRCh38
rs145735892 Chr.14: 24370424-24370424 on Build GRCh38
rs770039458 Chr.14: 24370421-24370421 on Build GRCh38
rs752444035 Chr.14: 24370251-24370251 on Build GRCh38
rs267603968 Chr.14: 24370466-24370466 on Build GRCh38
rs763820472 Chr.14: 24370362-24370362 on Build GRCh38
rs751915801 Chr.14: 24370249-24370249 on Build GRCh38
rs768667669 Chr.14: 24370220-24370220 on Build GRCh38
rs748276093 Chr.14: 24370320-24370320 on Build GRCh38
rs760576477 Chr.14: 24370441-24370441 on Build GRCh38
rs550028535 Chr.14: 24370612-24370612 on Build GRCh38
rs778422832 Chr.14: 24370606-24370606 on Build GRCh38
rs747953815 Chr.14: 24370618-24370618 on Build GRCh38
rs1043475370 Chr.14: 24370343-24370343 on Build GRCh38
rs898643666 Chr.14: 24370503-24370503 on Build GRCh38
rs756661952 Chr.14: 24370281-24370281 on Build GRCh38
rs372629167 Chr.14: 24370583-24370583 on Build GRCh38
rs141855863 Chr.14: 24370567-24370567 on Build GRCh38
rs56305813 Chr.14: 24370605-24370605 on Build GRCh38
rs760857991 Chr.14: 24370216-24370216 on Build GRCh38
rs28365132 Chr.14: 24370638-24370638 on Build GRCh38
rs749390935 Chr.14: 24370518-24370518 on Build GRCh38
rs201320818 Chr.14: 24370339-24370339 on Build GRCh38
rs563480110 Chr.14: 24370617-24370617 on Build GRCh38
rs1050753964 Chr.14: 24370429-24370429 on Build GRCh38
rs1025709814 Chr.14: 24370229-24370229 on Build GRCh38
rs267603969 Chr.14: 24370467-24370467 on Build GRCh38
rs200091677 Chr.14: 24370610-24370610 on Build GRCh38
rs751998990 Chr.14: 24370496-24370496 on Build GRCh38
rs995729372 Chr.14: 24370546-24370546 on Build GRCh38
rs28365129 Chr.14: 24370446-24370446 on Build GRCh38
rs990645469 Chr.14: 24370325-24370325 on Build GRCh38
rs576702065 Chr.14: 24370318-24370318 on Build GRCh38
rs367852293 Chr.14: 24370226-24370226 on Build GRCh38
rs145126892 Chr.14: 24370236-24370236 on Build GRCh38
rs775232914 Chr.14: 24370436-24370436 on Build GRCh38
rs368939796 Chr.14: 24370564-24370564 on Build GRCh38
rs780374843 Chr.14: 24370302-24370302 on Build GRCh38
rs372295120 Chr.14: 24370614-24370614 on Build GRCh38
rs879139888 Chr.14: 24370422-24370422 on Build GRCh38
rs780104322 Chr.14: 24370545-24370545 on Build GRCh38
rs775453007 Chr.14: 24370354-24370354 on Build GRCh38
rs747288091 Chr.14: 24370309-24370309 on Build GRCh38
rs758927641 Chr.14: 24370388-24370388 on Build GRCh38
rs779923071 Chr.14: 24370654-24370654 on Build GRCh38
rs749336968 Chr.14: 24370426-24370426 on Build GRCh38
rs758804078 Chr.14: 24370301-24370301 on Build GRCh38
rs924191397 Chr.14: 24370358-24370358 on Build GRCh38
rs753004524 Chr.14: 24370600-24370600 on Build GRCh38
rs781536985 Chr.14: 24370403-24370403 on Build GRCh38
rs764821739 Chr.14: 24370477-24370477 on Build GRCh38
rs755244684 Chr.14: 24370401-24370401 on Build GRCh38
rs905702316 Chr.14: 24370483-24370483 on Build GRCh38
rs779453735 Chr.14: 24370291-24370291 on Build GRCh38
rs768330456 Chr.14: 24370444-24370444 on Build GRCh38
rs773760506 Chr.14: 24370626-24370626 on Build GRCh38
rs28365131 Chr.14: 24370611-24370611 on Build GRCh38
rs146803212 Chr.14: 24370363-24370363 on Build GRCh38
rs774375150 Chr.14: 24370237-24370237 on Build GRCh38
rs764696807 Chr.14: 24370372-24370372 on Build GRCh38
rs755440036 Chr.14: 24370501-24370501 on Build GRCh38
rs770565017 Chr.14: 24370625-24370625 on Build GRCh38
rs775757742 Chr.14: 24370208-24370208 on Build GRCh38
rs763833570 Chr.14: 24370583-24370583 on Build GRCh38
rs200059234 Chr.14: 24370293-24370293 on Build GRCh38
rs768957495 Chr.14: 24370315-24370315 on Build GRCh38
rs200197248 Chr.14: 24370433-24370433 on Build GRCh38
rs1018558602 Chr.14: 24370497-24370497 on Build GRCh38
rs781384710 Chr.14: 24370502-24370502 on Build GRCh38
rs753054199 Chr.14: 24370504-24370504 on Build GRCh38
rs111575647 Chr.14: 24370601-24370601 on Build GRCh38
rs533502621 Chr.14: 24370273-24370273 on Build GRCh38
rs529186981 Chr.14: 24370624-24370624 on Build GRCh38
rs766889405 Chr.14: 24370495-24370495 on Build GRCh38
rs780499534 Chr.14: 24370392-24370392 on Build GRCh38
rs916412659 Chr.14: 24370328-24370328 on Build GRCh38
rs774467153 Chr.14: 24370335-24370335 on Build GRCh38
rs750979890 Chr.14: 24370376-24370376 on Build GRCh38
rs753391587 Chr.14: 24370268-24370268 on Build GRCh38
rs755808600 Chr.14: 24370257-24370257 on Build GRCh38
rs759464719 Chr.14: 24370240-24370240 on Build GRCh38
rs372551807 Chr.14: 24370622-24370622 on Build GRCh38
rs759980664 Chr.14: 24370571-24370571 on Build GRCh38
rs765805909 Chr.14: 24370569-24370569 on Build GRCh38
rs762387275 Chr.14: 24370490-24370490 on Build GRCh38
rs756856999 Chr.14: 24370367-24370367 on Build GRCh38
rs747408188 Chr.14: 24370397-24370397 on Build GRCh38
rs988513502 Chr.14: 24370660-24370660 on Build GRCh38
rs568364142 Chr.14: 24370662-24370662 on Build GRCh38
rs766646108 Chr.14: 24370242-24370242 on Build GRCh38
rs140616415 Chr.14: 24370406-24370406 on Build GRCh38
rs761351694 Chr.14: 24370474-24370474 on Build GRCh38
rs368967896 Chr.14: 24370619-24370619 on Build GRCh38
rs200325730 Chr.14: 24370551-24370551 on Build GRCh38
rs752150894 Chr.14: 24370570-24370570 on Build GRCh38
rs767509332 Chr.14: 24370346-24370346 on Build GRCh38
rs949780426 Chr.14: 24370427-24370427 on Build GRCh38
rs753201262 Chr.14: 24370482-24370482 on Build GRCh38
rs760319993 Chr.14: 24370355-24370355 on Build GRCh38
rs749967304 Chr.14: 24370485-24370485 on Build GRCh38
rs778071519 Chr.14: 24370515-24370515 on Build GRCh38
rs771121427 Chr.14: 24370520-24370520 on Build GRCh38
rs540621074 Chr.14: 24370289-24370289 on Build GRCh38
rs769984138 Chr.14: 24370324-24370324 on Build GRCh38

View Full Product Details