Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24505424-24505933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMA1
Entrez Gene ID
1215
Gene Name
chymase 1
Gene Aliases
CYH, MCT1, chymase
UniGene
Hs.135626
Gene Chromosome Location
Chr.14: 24505355-24508265 on Build GRCh38
Transcripts
NM_001308083.1 NM_001836.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6075654 c.677C>A p.P226H Chr.14: 24505583-24505583 on Build GRCh38
6630896 c.666G>A p.S222S Chr.14: 24505594-24505594 on Build GRCh38
5171115 c.661C>T p.R221W Chr.14: 24505599-24505599 on Build GRCh38
6630895 c.695G>A p.R232Q Chr.14: 24505565-24505565 on Build GRCh38
6037699 c.710G>T p.R237L Chr.14: 24505550-24505550 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376938676 Chr.14: 24505565-24505565 on Build GRCh38
rs753178749 Chr.14: 24505671-24505672 on Build GRCh38
rs1024717800 Chr.14: 24505906-24505906 on Build GRCh38
rs140295781 Chr.14: 24505550-24505550 on Build GRCh38
rs781755382 Chr.14: 24505676-24505676 on Build GRCh38
rs754810177 Chr.14: 24505468-24505468 on Build GRCh38
rs767899284 Chr.14: 24505646-24505646 on Build GRCh38
rs200774278 Chr.14: 24505566-24505566 on Build GRCh38
rs376803877 Chr.14: 24505558-24505558 on Build GRCh38
rs994137637 Chr.14: 24505904-24505904 on Build GRCh38
rs773873620 Chr.14: 24505559-24505559 on Build GRCh38
rs371301206 Chr.14: 24505694-24505694 on Build GRCh38
rs757481917 Chr.14: 24505530-24505530 on Build GRCh38
rs747478110 Chr.14: 24505612-24505612 on Build GRCh38
rs982597430 Chr.14: 24505778-24505778 on Build GRCh38
rs760627071 Chr.14: 24505647-24505647 on Build GRCh38
rs1043420558 Chr.14: 24505800-24505800 on Build GRCh38
rs750416753 Chr.14: 24505532-24505532 on Build GRCh38
rs914451934 Chr.14: 24505668-24505668 on Build GRCh38
rs760911999 Chr.14: 24505523-24505523 on Build GRCh38
rs758379105 Chr.14: 24505597-24505597 on Build GRCh38
rs769517576 Chr.14: 24505487-24505487 on Build GRCh38
rs775950520 Chr.14: 24505514-24505514 on Build GRCh38
rs746942808 Chr.14: 24505467-24505467 on Build GRCh38
rs970944165 Chr.14: 24505758-24505758 on Build GRCh38
rs556094098 Chr.14: 24505745-24505745 on Build GRCh38
rs753673742 Chr.14: 24505658-24505658 on Build GRCh38
rs143164371 Chr.14: 24505611-24505611 on Build GRCh38
rs574096992 Chr.14: 24505649-24505649 on Build GRCh38
rs764456752 Chr.14: 24505525-24505525 on Build GRCh38
rs905215870 Chr.14: 24505462-24505462 on Build GRCh38
rs142397893 Chr.14: 24505594-24505594 on Build GRCh38
rs769649841 Chr.14: 24505754-24505754 on Build GRCh38
rs145606317 Chr.14: 24505497-24505497 on Build GRCh38
rs760102150 Chr.14: 24505510-24505510 on Build GRCh38
rs374224314 Chr.14: 24505659-24505659 on Build GRCh38
rs963587828 Chr.14: 24505750-24505750 on Build GRCh38
rs539126881 Chr.14: 24505464-24505464 on Build GRCh38
rs904540389 Chr.14: 24505824-24505824 on Build GRCh38
rs373394059 Chr.14: 24505474-24505474 on Build GRCh38
rs750037025 Chr.14: 24505667-24505667 on Build GRCh38
rs200452938 Chr.14: 24505598-24505598 on Build GRCh38
rs778682089 Chr.14: 24505666-24505666 on Build GRCh38
rs774324609 Chr.14: 24505700-24505700 on Build GRCh38
rs772671767 Chr.14: 24505568-24505568 on Build GRCh38
rs766885748 Chr.14: 24505475-24505475 on Build GRCh38
rs553154016 Chr.14: 24505724-24505724 on Build GRCh38
rs959451307 Chr.14: 24505820-24505820 on Build GRCh38
rs1037343171 Chr.14: 24505899-24505899 on Build GRCh38
rs746471143 Chr.14: 24505672-24505672 on Build GRCh38
rs763151685 Chr.14: 24505641-24505641 on Build GRCh38
rs888774143 Chr.14: 24505737-24505737 on Build GRCh38
rs991927118 Chr.14: 24505488-24505488 on Build GRCh38
rs926748563 Chr.14: 24505787-24505787 on Build GRCh38
rs968910061 Chr.14: 24505460-24505460 on Build GRCh38
rs770181227 Chr.14: 24505625-24505625 on Build GRCh38
rs755977857 Chr.14: 24505599-24505599 on Build GRCh38
rs573099074 Chr.14: 24505749-24505749 on Build GRCh38
rs571589495 Chr.14: 24505606-24505606 on Build GRCh38
rs747849591 Chr.14: 24505484-24505484 on Build GRCh38
rs777556273 Chr.14: 24505603-24505603 on Build GRCh38
rs1951133 Chr.14: 24505826-24505826 on Build GRCh38
rs1007325607 Chr.14: 24505895-24505895 on Build GRCh38
rs917712638 Chr.14: 24505643-24505643 on Build GRCh38
rs924425016 Chr.14: 24505862-24505862 on Build GRCh38
rs773284314 Chr.14: 24505626-24505626 on Build GRCh38
rs753580823 Chr.14: 24505605-24505605 on Build GRCh38
rs771135098 Chr.14: 24505695-24505695 on Build GRCh38
rs763979088 Chr.14: 24505654-24505654 on Build GRCh38
rs137883267 Chr.14: 24505584-24505584 on Build GRCh38
rs933204461 Chr.14: 24505863-24505863 on Build GRCh38
rs933186811 Chr.14: 24505746-24505746 on Build GRCh38
rs145379274 Chr.14: 24505556-24505556 on Build GRCh38
rs765509983 Chr.14: 24505585-24505585 on Build GRCh38
rs888831279 Chr.14: 24505873-24505873 on Build GRCh38
rs769322139 Chr.14: 24505674-24505674 on Build GRCh38
rs758000352 Chr.14: 24505671-24505671 on Build GRCh38
rs73596976 Chr.14: 24505498-24505498 on Build GRCh38
rs1046793605 Chr.14: 24505450-24505450 on Build GRCh38
rs1020120691 Chr.14: 24505529-24505529 on Build GRCh38
rs10140250 Chr.14: 24505757-24505757 on Build GRCh38
rs777161235 Chr.14: 24505619-24505619 on Build GRCh38
rs372242977 Chr.14: 24505511-24505511 on Build GRCh38
rs773781864 Chr.14: 24505504-24505504 on Build GRCh38
rs75089419 Chr.14: 24505850-24505850 on Build GRCh38
rs12100565 Chr.14: 24505714-24505714 on Build GRCh38
rs145496377 Chr.14: 24505547-24505547 on Build GRCh38
rs781057151 Chr.14: 24505473-24505473 on Build GRCh38
rs773552345 Chr.14: 24505686-24505686 on Build GRCh38
rs748942989 Chr.14: 24505551-24505551 on Build GRCh38
rs573747911 Chr.14: 24505520-24505520 on Build GRCh38
rs749183970 Chr.14: 24505792-24505792 on Build GRCh38
rs770271239 Chr.14: 24505682-24505682 on Build GRCh38
rs748016473 Chr.14: 24505543-24505543 on Build GRCh38
rs778329149 Chr.14: 24505782-24505782 on Build GRCh38
rs746542097 Chr.14: 24505538-24505538 on Build GRCh38
rs575158336 Chr.14: 24505821-24505821 on Build GRCh38
rs546717814 Chr.14: 24505909-24505909 on Build GRCh38
rs751597406 Chr.14: 24505646-24505646 on Build GRCh38
rs761157051 Chr.14: 24505580-24505580 on Build GRCh38
rs769230485 Chr.14: 24505618-24505618 on Build GRCh38
rs947727665 Chr.14: 24505645-24505645 on Build GRCh38
rs748573929 Chr.14: 24505680-24505680 on Build GRCh38
rs764546466 Chr.14: 24505583-24505583 on Build GRCh38
rs762233096 Chr.14: 24505621-24505621 on Build GRCh38
rs557045901 Chr.14: 24505644-24505644 on Build GRCh38
rs759580723 Chr.14: 24505708-24505708 on Build GRCh38
rs1051509585 Chr.14: 24505864-24505864 on Build GRCh38
rs880678 Chr.14: 24505838-24505838 on Build GRCh38
rs898850371 Chr.14: 24505900-24505900 on Build GRCh38
rs373795207 Chr.14: 24505539-24505539 on Build GRCh38
rs184287309 Chr.14: 24505783-24505783 on Build GRCh38
rs767862206 Chr.14: 24505513-24505513 on Build GRCh38
rs760172371 Chr.14: 24505567-24505569 on Build GRCh38
rs1007543810 Chr.14: 24505777-24505777 on Build GRCh38
rs780194957 Chr.14: 24505466-24505466 on Build GRCh38
rs775834473 Chr.14: 24505574-24505574 on Build GRCh38
rs745429348 Chr.14: 24505608-24505608 on Build GRCh38

View Full Product Details