Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 67569833-67570331 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPHN
Entrez Gene ID
10243
Gene Name
gephyrin
Gene Aliases
GEPH, GPH, GPHRYN, HKPX1, MOCODC
UniGene
Hs.208765
Gene Chromosome Location
Chr.14: 66507407-67735831 on Build GRCh38
Transcripts
XM_017020925.1 NM_020715.2 XM_011537009.1 XM_011537015.2 XM_017021499.1 XM_017021500.1 XM_017021501.1 XM_017021502.1 XM_017021503.1 XM_017021504.1

Gene Symbol
PLEKHH1
Entrez Gene ID
57475
Gene Name
pleckstrin homology, MyTH4 and FERM domain containing H1
Gene Aliases
-
UniGene
Hs.594236
Gene Chromosome Location
Chr.14: 67533155-67589766 on Build GRCh38
Transcripts
XM_017020925.1 NM_020715.2 XM_011537009.1 XM_011537015.2 XM_017021499.1 XM_017021500.1 XM_017021501.1 XM_017021502.1 XM_017021503.1 XM_017021504.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4464896 c.1475C>T p.A492V Chr.14: 67569936-67569936 on Build GRCh38
4464895 c.1358C>T p.A453V Chr.14: 67569936-67569936 on Build GRCh38
158703 c.1423G>A p.A475T Chr.14: 67570001-67570001 on Build GRCh38
6463634 c.1484C>T p.S495F Chr.14: 67569945-67569945 on Build GRCh38
6463633 c.1367C>T p.S456F Chr.14: 67569945-67569945 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs773200744 Chr.14: 67570028-67570028 on Build GRCh38
rs144631480 Chr.14: 67570309-67570309 on Build GRCh38
rs762855729 Chr.14: 67570308-67570308 on Build GRCh38
rs762795049 Chr.14: 67569913-67569913 on Build GRCh38
rs368951410 Chr.14: 67569874-67569874 on Build GRCh38
rs752176538 Chr.14: 67569961-67569961 on Build GRCh38
rs45580633 Chr.14: 67569857-67569857 on Build GRCh38
rs888756769 Chr.14: 67570094-67570094 on Build GRCh38
rs780846311 Chr.14: 67569956-67569956 on Build GRCh38
rs893986397 Chr.14: 67570135-67570135 on Build GRCh38
rs996543051 Chr.14: 67569950-67569950 on Build GRCh38
rs1039295023 Chr.14: 67570186-67570186 on Build GRCh38
rs776846601 Chr.14: 67570012-67570012 on Build GRCh38
rs59589798 Chr.14: 67569872-67569872 on Build GRCh38
rs780547017 Chr.14: 67570002-67570002 on Build GRCh38
rs1015675772 Chr.14: 67570286-67570286 on Build GRCh38
rs901456663 Chr.14: 67570101-67570101 on Build GRCh38
rs945492572 Chr.14: 67570140-67570140 on Build GRCh38
rs1030471266 Chr.14: 67570255-67570255 on Build GRCh38
rs779350639 Chr.14: 67569989-67569989 on Build GRCh38
rs748191491 Chr.14: 67570018-67570018 on Build GRCh38
rs1056542078 Chr.14: 67570110-67570110 on Build GRCh38
rs982827683 Chr.14: 67570030-67570030 on Build GRCh38
rs900664371 Chr.14: 67570220-67570220 on Build GRCh38
rs183489446 Chr.14: 67570146-67570146 on Build GRCh38
rs149341763 Chr.14: 67570147-67570147 on Build GRCh38
rs955266453 Chr.14: 67570272-67570272 on Build GRCh38
rs191264111 Chr.14: 67570100-67570100 on Build GRCh38
rs769903014 Chr.14: 67570020-67570020 on Build GRCh38
rs537917119 Chr.14: 67570230-67570231 on Build GRCh38
rs1031128866 Chr.14: 67570122-67570122 on Build GRCh38
rs2273593 Chr.14: 67570136-67570136 on Build GRCh38
rs1050143189 Chr.14: 67570055-67570055 on Build GRCh38
rs766156545 Chr.14: 67569932-67569932 on Build GRCh38
rs926437398 Chr.14: 67570087-67570087 on Build GRCh38
rs750302656 Chr.14: 67569905-67569905 on Build GRCh38
rs35091785 Chr.14: 67569976-67569977 on Build GRCh38
rs754561229 Chr.14: 67569945-67569945 on Build GRCh38
rs974058679 Chr.14: 67570083-67570083 on Build GRCh38
rs755581767 Chr.14: 67569978-67569978 on Build GRCh38
rs747329536 Chr.14: 67570004-67570004 on Build GRCh38
rs1008539958 Chr.14: 67570098-67570098 on Build GRCh38
rs955144038 Chr.14: 67569966-67569966 on Build GRCh38
rs765091371 Chr.14: 67569884-67569884 on Build GRCh38
rs751252580 Chr.14: 67569943-67569943 on Build GRCh38
rs770585859 Chr.14: 67570010-67570010 on Build GRCh38
rs745521473 Chr.14: 67570041-67570041 on Build GRCh38
rs962910684 Chr.14: 67570039-67570039 on Build GRCh38
rs1000099512 Chr.14: 67570108-67570108 on Build GRCh38
rs762844377 Chr.14: 67570035-67570035 on Build GRCh38
rs1029392417 Chr.14: 67569963-67569963 on Build GRCh38
rs962442747 Chr.14: 67570287-67570287 on Build GRCh38
rs969360576 Chr.14: 67570253-67570253 on Build GRCh38
rs541207390 Chr.14: 67569875-67569875 on Build GRCh38
rs45457800 Chr.14: 67570036-67570036 on Build GRCh38
rs964886768 Chr.14: 67569926-67569926 on Build GRCh38
rs746376967 Chr.14: 67569992-67569992 on Build GRCh38
rs750719773 Chr.14: 67569986-67569986 on Build GRCh38
rs774151262 Chr.14: 67570047-67570047 on Build GRCh38
rs772607954 Chr.14: 67569996-67569996 on Build GRCh38
rs372734884 Chr.14: 67570281-67570281 on Build GRCh38
rs549547910 Chr.14: 67570150-67570150 on Build GRCh38

View Full Product Details