Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 69572256-69572747 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC177
Entrez Gene ID
56936
Gene Name
coiled-coil domain containing 177
Gene Aliases
C14orf162, PLPL
UniGene
Hs.458319
Gene Chromosome Location
Chr.14: 69569814-69574883 on Build GRCh38
Transcripts
NM_001271507.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6863648 c.1115T>C p.L372P Chr.14: 69572460-69572460 on Build GRCh38
6220690 c.918C>T p.G306G Chr.14: 69572705-69572705 on Build GRCh38
6863647 c.1163T>C p.L388P Chr.14: 69572460-69572460 on Build GRCh38
6220691 c.870C>T p.G290G Chr.14: 69572705-69572705 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs932569889 Chr.14: 69572323-69572323 on Build GRCh38
rs1042189662 Chr.14: 69572395-69572395 on Build GRCh38
rs532991181 Chr.14: 69572529-69572529 on Build GRCh38
rs112597196 Chr.14: 69572725-69572725 on Build GRCh38
rs1053993540 Chr.14: 69572307-69572307 on Build GRCh38
rs982508571 Chr.14: 69572467-69572467 on Build GRCh38
rs982340576 Chr.14: 69572722-69572722 on Build GRCh38
rs897543948 Chr.14: 69572419-69572420 on Build GRCh38
rs569667443 Chr.14: 69572645-69572645 on Build GRCh38
rs1004252816 Chr.14: 69572628-69572628 on Build GRCh38
rs1000253961 Chr.14: 69572577-69572577 on Build GRCh38
rs989574045 Chr.14: 69572566-69572566 on Build GRCh38
rs1000285120 Chr.14: 69572419-69572419 on Build GRCh38
rs899717947 Chr.14: 69572317-69572317 on Build GRCh38
rs912432688 Chr.14: 69572705-69572705 on Build GRCh38
rs548978631 Chr.14: 69572723-69572723 on Build GRCh38
rs996046057 Chr.14: 69572326-69572326 on Build GRCh38
rs962462174 Chr.14: 69572495-69572495 on Build GRCh38
rs894259537 Chr.14: 69572663-69572663 on Build GRCh38
rs1038213392 Chr.14: 69572300-69572300 on Build GRCh38
rs954007038 Chr.14: 69572688-69572688 on Build GRCh38
rs973802036 Chr.14: 69572659-69572659 on Build GRCh38
rs1024975559 Chr.14: 69572619-69572619 on Build GRCh38
rs973874136 Chr.14: 69572497-69572497 on Build GRCh38
rs945324389 Chr.14: 69572710-69572710 on Build GRCh38
rs921044889 Chr.14: 69572668-69572668 on Build GRCh38
rs540973036 Chr.14: 69572501-69572501 on Build GRCh38
rs982054919 Chr.14: 69572527-69572527 on Build GRCh38
rs912516693 Chr.14: 69572538-69572538 on Build GRCh38
rs986651367 Chr.14: 69572699-69572699 on Build GRCh38
rs890347168 Chr.14: 69572597-69572597 on Build GRCh38
rs944663466 Chr.14: 69572548-69572548 on Build GRCh38
rs1013566575 Chr.14: 69572450-69572450 on Build GRCh38
rs931783200 Chr.14: 69572582-69572582 on Build GRCh38
rs756630341 Chr.14: 69572462-69572462 on Build GRCh38
rs890436124 Chr.14: 69572341-69572341 on Build GRCh38
rs1013118839 Chr.14: 69572598-69572598 on Build GRCh38
rs958922835 Chr.14: 69572448-69572448 on Build GRCh38
rs894632880 Chr.14: 69572593-69572593 on Build GRCh38
rs903246497 Chr.14: 69572565-69572565 on Build GRCh38
rs990972425 Chr.14: 69572496-69572496 on Build GRCh38
rs186090620 Chr.14: 69572626-69572626 on Build GRCh38
rs766909610 Chr.14: 69572636-69572636 on Build GRCh38
rs1042085407 Chr.14: 69572713-69572713 on Build GRCh38
rs529128209 Chr.14: 69572359-69572359 on Build GRCh38
rs1012341089 Chr.14: 69572687-69572687 on Build GRCh38
rs1054493472 Chr.14: 69572585-69572585 on Build GRCh38
rs1033152022 Chr.14: 69572447-69572447 on Build GRCh38
rs915352315 Chr.14: 69572569-69572569 on Build GRCh38
rs903276235 Chr.14: 69572406-69572406 on Build GRCh38
rs1041724733 Chr.14: 69572561-69572561 on Build GRCh38
rs560978456 Chr.14: 69572287-69572287 on Build GRCh38
rs1050428079 Chr.14: 69572327-69572327 on Build GRCh38
rs890399695 Chr.14: 69572336-69572336 on Build GRCh38
rs962432099 Chr.14: 69572654-69572654 on Build GRCh38
rs941172314 Chr.14: 69572279-69572279 on Build GRCh38
rs919368673 Chr.14: 69572490-69572490 on Build GRCh38
rs935382131 Chr.14: 69572558-69572558 on Build GRCh38
rs908249647 Chr.14: 69572494-69572494 on Build GRCh38
rs971462943 Chr.14: 69572464-69572464 on Build GRCh38
rs530665858 Chr.14: 69572707-69572707 on Build GRCh38
rs1050277740 Chr.14: 69572595-69572595 on Build GRCh38
rs559111245 Chr.14: 69572503-69572503 on Build GRCh38
rs76334242 Chr.14: 69572727-69572727 on Build GRCh38
rs944621990 Chr.14: 69572600-69572600 on Build GRCh38
rs1027403401 Chr.14: 69572646-69572646 on Build GRCh38
rs551409308 Chr.14: 69572612-69572612 on Build GRCh38

View Full Product Details