Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 70524614-70525115 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM20
Entrez Gene ID
8748
Gene Name
ADAM metallopeptidase domain 20
Gene Aliases
-
UniGene
Hs.177984
Gene Chromosome Location
Chr.14: 70520817-70564742 on Build GRCh38
Transcripts
NM_003814.4 XM_005268151.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6521636 c.210G>T p.G70G Chr.14: 70524698-70524698 on Build GRCh38
5722224 c.251C>T p.S84F Chr.14: 70524657-70524657 on Build GRCh38
1515849 c.270A>G p.P90P Chr.14: 70524638-70524638 on Build GRCh38
4623007 c.284C>A p.P95H Chr.14: 70524624-70524624 on Build GRCh38
3815220 c.100A>C p.T34P Chr.14: 70524808-70524808 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs935101179 Chr.14: 70524845-70524845 on Build GRCh38
rs774621988 Chr.14: 70524950-70524950 on Build GRCh38
rs989643101 Chr.14: 70525064-70525064 on Build GRCh38
rs759941193 Chr.14: 70524773-70524773 on Build GRCh38
rs766765851 Chr.14: 70524937-70524938 on Build GRCh38
rs754136240 Chr.14: 70524769-70524769 on Build GRCh38
rs771166581 Chr.14: 70524891-70524891 on Build GRCh38
rs1024394504 Chr.14: 70524651-70524651 on Build GRCh38
rs192108977 Chr.14: 70524995-70524995 on Build GRCh38
rs752081447 Chr.14: 70524955-70524955 on Build GRCh38
rs761900874 Chr.14: 70524638-70524638 on Build GRCh38
rs952524221 Chr.14: 70524689-70524689 on Build GRCh38
rs765700772 Chr.14: 70524785-70524785 on Build GRCh38
rs367747671 Chr.14: 70524848-70524848 on Build GRCh38
rs769500504 Chr.14: 70524685-70524685 on Build GRCh38
rs914758986 Chr.14: 70524864-70524864 on Build GRCh38
rs1059166 Chr.14: 70524701-70524701 on Build GRCh38
rs1025905022 Chr.14: 70525058-70525058 on Build GRCh38
rs768457876 Chr.14: 70524704-70524704 on Build GRCh38
rs780117254 Chr.14: 70524909-70524909 on Build GRCh38
rs200334934 Chr.14: 70524917-70524917 on Build GRCh38
rs749126085 Chr.14: 70524699-70524699 on Build GRCh38
rs556648367 Chr.14: 70525051-70525051 on Build GRCh38
rs777568501 Chr.14: 70524652-70524652 on Build GRCh38
rs771946427 Chr.14: 70524874-70524874 on Build GRCh38
rs756351593 Chr.14: 70524654-70524654 on Build GRCh38
rs779071246 Chr.14: 70524979-70524979 on Build GRCh38
rs779513861 Chr.14: 70524670-70524670 on Build GRCh38
rs17176236 Chr.14: 70525089-70525089 on Build GRCh38
rs950445798 Chr.14: 70524805-70524805 on Build GRCh38
rs773794388 Chr.14: 70524658-70524685 on Build GRCh38
rs768386169 Chr.14: 70524850-70524850 on Build GRCh38
rs962947756 Chr.14: 70525019-70525019 on Build GRCh38
rs1014215411 Chr.14: 70524976-70524977 on Build GRCh38
rs74062730 Chr.14: 70524933-70524933 on Build GRCh38
rs760949231 Chr.14: 70524753-70524753 on Build GRCh38
rs199645906 Chr.14: 70524678-70524678 on Build GRCh38
rs766758251 Chr.14: 70524764-70524764 on Build GRCh38
rs776925963 Chr.14: 70524893-70524893 on Build GRCh38
rs956372612 Chr.14: 70525061-70525061 on Build GRCh38
rs758885113 Chr.14: 70524820-70524820 on Build GRCh38
rs750605319 Chr.14: 70524653-70524653 on Build GRCh38
rs538264689 Chr.14: 70525040-70525040 on Build GRCh38
rs766849076 Chr.14: 70524655-70524655 on Build GRCh38
rs748114890 Chr.14: 70524868-70524868 on Build GRCh38
rs755738554 Chr.14: 70524828-70524828 on Build GRCh38
rs373576636 Chr.14: 70524742-70524742 on Build GRCh38
rs73282130 Chr.14: 70524988-70524988 on Build GRCh38
rs753189243 Chr.14: 70524812-70524812 on Build GRCh38
rs965296016 Chr.14: 70525034-70525034 on Build GRCh38
rs914955258 Chr.14: 70525068-70525068 on Build GRCh38
rs987587159 Chr.14: 70524839-70524839 on Build GRCh38
rs1017130846 Chr.14: 70525018-70525018 on Build GRCh38
rs987168287 Chr.14: 70525066-70525066 on Build GRCh38
rs965014835 Chr.14: 70524712-70524712 on Build GRCh38
rs754295950 Chr.14: 70524663-70524663 on Build GRCh38
rs778636555 Chr.14: 70524857-70524857 on Build GRCh38
rs756719590 Chr.14: 70524676-70524676 on Build GRCh38
rs142051635 Chr.14: 70524672-70524672 on Build GRCh38
rs773739401 Chr.14: 70525014-70525014 on Build GRCh38
rs866351370 Chr.14: 70524752-70524752 on Build GRCh38
rs761674464 Chr.14: 70524719-70524719 on Build GRCh38
rs755520106 Chr.14: 70524668-70524668 on Build GRCh38
rs138673978 Chr.14: 70524685-70524686 on Build GRCh38
rs531984152 Chr.14: 70524813-70524813 on Build GRCh38
rs767502311 Chr.14: 70524640-70524640 on Build GRCh38
rs749938906 Chr.14: 70524827-70524827 on Build GRCh38
rs371491610 Chr.14: 70524882-70524882 on Build GRCh38
rs202217158 Chr.14: 70524698-70524698 on Build GRCh38
rs759851332 Chr.14: 70524913-70524913 on Build GRCh38
rs753068057 Chr.14: 70524931-70524931 on Build GRCh38
rs764555626 Chr.14: 70524951-70524951 on Build GRCh38
rs774282355 Chr.14: 70524709-70524709 on Build GRCh38
rs918960136 Chr.14: 70524734-70524734 on Build GRCh38
rs61739470 Chr.14: 70524705-70524705 on Build GRCh38
rs140900259 Chr.14: 70524971-70524971 on Build GRCh38
rs1031104742 Chr.14: 70525044-70525044 on Build GRCh38
rs969124749 Chr.14: 70525079-70525079 on Build GRCh38
rs767624174 Chr.14: 70524741-70524741 on Build GRCh38
rs780748159 Chr.14: 70524823-70524823 on Build GRCh38
rs748967558 Chr.14: 70524849-70524849 on Build GRCh38
rs994461083 Chr.14: 70525031-70525031 on Build GRCh38
rs888286163 Chr.14: 70524992-70524992 on Build GRCh38
rs745519054 Chr.14: 70524682-70524682 on Build GRCh38
rs756400114 Chr.14: 70524904-70524904 on Build GRCh38
rs985353996 Chr.14: 70524697-70524697 on Build GRCh38
rs976861065 Chr.14: 70524727-70524727 on Build GRCh38

View Full Product Details