Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 73654409-73654909 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAL1
Entrez Gene ID
83544
Gene Name
dynein axonemal light chain 1
Gene Aliases
C14orf168, CILD16
UniGene
Hs.271270
Gene Chromosome Location
Chr.14: 73644875-73703728 on Build GRCh38
Transcripts
NM_001201366.1 NM_031427.3 XM_017021679.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4247615 c.4-5T>A p.? Chr.14: 73654842-73654842 on Build GRCh38
6435044 c.14C>A p.T5K Chr.14: 73654857-73654857 on Build GRCh38
6233568 c.4-4A>T p.? Chr.14: 73654843-73654843 on Build GRCh38
7264181 c.36G>A p.A12A Chr.14: 73654879-73654879 on Build GRCh38
2137275 c.35C>T p.A12V Chr.14: 73654878-73654878 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs78059622 Chr.14: 73654453-73654453 on Build GRCh38
rs267604049 Chr.14: 73654879-73654879 on Build GRCh38
rs752349290 Chr.14: 73654848-73654848 on Build GRCh38
rs185924269 Chr.14: 73654832-73654832 on Build GRCh38
rs762472204 Chr.14: 73654829-73654829 on Build GRCh38
rs548144070 Chr.14: 73654766-73654777 on Build GRCh38
rs753404083 Chr.14: 73654828-73654829 on Build GRCh38
rs868263275 Chr.14: 73654604-73654604 on Build GRCh38
rs975002339 Chr.14: 73654450-73654450 on Build GRCh38
rs138794449 Chr.14: 73654630-73654630 on Build GRCh38
rs147204878 Chr.14: 73654496-73654496 on Build GRCh38
rs999369407 Chr.14: 73654698-73654698 on Build GRCh38
rs532973156 Chr.14: 73654486-73654486 on Build GRCh38
rs973823178 Chr.14: 73654736-73654736 on Build GRCh38
rs866739576 Chr.14: 73654857-73654857 on Build GRCh38
rs779309893 Chr.14: 73654865-73654867 on Build GRCh38
rs1033975543 Chr.14: 73654611-73654611 on Build GRCh38
rs181334072 Chr.14: 73654555-73654555 on Build GRCh38
rs1053468912 Chr.14: 73654469-73654469 on Build GRCh38
rs1030894125 Chr.14: 73654705-73654705 on Build GRCh38
rs547716136 Chr.14: 73654720-73654720 on Build GRCh38
rs939327424 Chr.14: 73654624-73654624 on Build GRCh38
rs955341059 Chr.14: 73654706-73654706 on Build GRCh38
rs777043273 Chr.14: 73654459-73654459 on Build GRCh38
rs757174470 Chr.14: 73654831-73654831 on Build GRCh38
rs755674439 Chr.14: 73654821-73654824 on Build GRCh38
rs113189274 Chr.14: 73654515-73654515 on Build GRCh38
rs950786755 Chr.14: 73654653-73654653 on Build GRCh38
rs576147534 Chr.14: 73654843-73654843 on Build GRCh38
rs533854521 Chr.14: 73654703-73654703 on Build GRCh38
rs371874491 Chr.14: 73654759-73654759 on Build GRCh38
rs769148315 Chr.14: 73654803-73654803 on Build GRCh38
rs777360063 Chr.14: 73654829-73654829 on Build GRCh38
rs867119843 Chr.14: 73654796-73654796 on Build GRCh38
rs983682695 Chr.14: 73654788-73654788 on Build GRCh38
rs974887746 Chr.14: 73654738-73654738 on Build GRCh38
rs753725871 Chr.14: 73654781-73654781 on Build GRCh38
rs920729011 Chr.14: 73654765-73654765 on Build GRCh38
rs948104206 Chr.14: 73654878-73654878 on Build GRCh38
rs1027675052 Chr.14: 73654645-73654645 on Build GRCh38
rs939651787 Chr.14: 73654828-73654828 on Build GRCh38
rs757741182 Chr.14: 73654829-73654830 on Build GRCh38
rs1056495201 Chr.14: 73654628-73654628 on Build GRCh38
rs986859383 Chr.14: 73654719-73654719 on Build GRCh38
rs930814369 Chr.14: 73654780-73654780 on Build GRCh38
rs1003458585 Chr.14: 73654600-73654600 on Build GRCh38
rs568298706 Chr.14: 73654693-73654693 on Build GRCh38
rs116380199 Chr.14: 73654556-73654556 on Build GRCh38
rs775275165 Chr.14: 73654674-73654674 on Build GRCh38
rs371876457 Chr.14: 73654587-73654587 on Build GRCh38
rs983530848 Chr.14: 73654696-73654696 on Build GRCh38
rs539481036 Chr.14: 73654773-73654773 on Build GRCh38
rs989531770 Chr.14: 73654785-73654785 on Build GRCh38
rs865816165 Chr.14: 73654845-73654845 on Build GRCh38
rs540183120 Chr.14: 73654529-73654529 on Build GRCh38
rs886414868 Chr.14: 73654615-73654615 on Build GRCh38
rs531500740 Chr.14: 73654629-73654629 on Build GRCh38
rs763932147 Chr.14: 73654877-73654877 on Build GRCh38
rs746219926 Chr.14: 73654865-73654866 on Build GRCh38
rs540475697 Chr.14: 73654777-73654778 on Build GRCh38
rs1054981232 Chr.14: 73654860-73654863 on Build GRCh38
rs776047200 Chr.14: 73654837-73654837 on Build GRCh38
rs552281308 Chr.14: 73654582-73654582 on Build GRCh38
rs570735237 Chr.14: 73654731-73654731 on Build GRCh38
rs868121321 Chr.14: 73654792-73654792 on Build GRCh38
rs922147684 Chr.14: 73654766-73654769 on Build GRCh38
rs762585015 Chr.14: 73654851-73654851 on Build GRCh38
rs957660162 Chr.14: 73654781-73654792 on Build GRCh38
rs774853229 Chr.14: 73654819-73654819 on Build GRCh38

View Full Product Details