Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 73717164-73717674 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELMSAN1
Entrez Gene ID
91748
Gene Name
ELM2 and Myb/SANT domain containing 1
Gene Aliases
C14orf117, C14orf43, LSR68, MIDEAS, c14_5541
UniGene
Hs.594157
Gene Chromosome Location
Chr.14: 73715122-73790285 on Build GRCh38
Transcripts
NM_001043318.2 NM_194278.3 XM_005268204.1 XM_005268205.1 XM_005268206.1

SNP Detail

SNP ID Chromosome Location
rs1027713101 Chr.14: 73717234-73717234 on Build GRCh38
rs926131532 Chr.14: 73717274-73717274 on Build GRCh38
rs1049121524 Chr.14: 73717515-73717515 on Build GRCh38
rs7156893 Chr.14: 73717489-73717489 on Build GRCh38
rs993304930 Chr.14: 73717464-73717464 on Build GRCh38
rs1013897645 Chr.14: 73717619-73717619 on Build GRCh38
rs536965340 Chr.14: 73717548-73717548 on Build GRCh38
rs571470148 Chr.14: 73717537-73717537 on Build GRCh38
rs565565347 Chr.14: 73717240-73717240 on Build GRCh38
rs573681804 Chr.14: 73717598-73717598 on Build GRCh38
rs917407261 Chr.14: 73717590-73717590 on Build GRCh38
rs1056858252 Chr.14: 73717391-73717391 on Build GRCh38
rs926287061 Chr.14: 73717604-73717604 on Build GRCh38
rs549703692 Chr.14: 73717300-73717300 on Build GRCh38
rs534799283 Chr.14: 73717524-73717524 on Build GRCh38
rs936137743 Chr.14: 73717396-73717396 on Build GRCh38
rs531121861 Chr.14: 73717243-73717243 on Build GRCh38
rs979149556 Chr.14: 73717327-73717327 on Build GRCh38
rs913473923 Chr.14: 73717455-73717455 on Build GRCh38
rs1040630974 Chr.14: 73717473-73717473 on Build GRCh38
rs925055651 Chr.14: 73717376-73717376 on Build GRCh38
rs542587161 Chr.14: 73717286-73717286 on Build GRCh38
rs950392385 Chr.14: 73717594-73717594 on Build GRCh38
rs1026242565 Chr.14: 73717479-73717479 on Build GRCh38
rs148502951 Chr.14: 73717520-73717520 on Build GRCh38
rs969031401 Chr.14: 73717635-73717635 on Build GRCh38
rs1001985572 Chr.14: 73717367-73717367 on Build GRCh38
rs781505147 Chr.14: 73717500-73717501 on Build GRCh38
rs143023808 Chr.14: 73717191-73717192 on Build GRCh38
rs904857304 Chr.14: 73717326-73717326 on Build GRCh38
rs969062717 Chr.14: 73717317-73717317 on Build GRCh38
rs528966101 Chr.14: 73717456-73717456 on Build GRCh38
rs200530167 Chr.14: 73717196-73717196 on Build GRCh38
rs1023571679 Chr.14: 73717630-73717630 on Build GRCh38
rs1014841138 Chr.14: 73717265-73717265 on Build GRCh38
rs1004712187 Chr.14: 73717536-73717536 on Build GRCh38
rs996175999 Chr.14: 73717218-73717218 on Build GRCh38
rs548099432 Chr.14: 73717245-73717248 on Build GRCh38
rs1015137634 Chr.14: 73717552-73717552 on Build GRCh38
rs951851883 Chr.14: 73717241-73717241 on Build GRCh38
rs368524786 Chr.14: 73717302-73717302 on Build GRCh38
rs748164789 Chr.14: 73717589-73717589 on Build GRCh38
rs937618824 Chr.14: 73717617-73717617 on Build GRCh38
rs978812730 Chr.14: 73717253-73717253 on Build GRCh38
rs960942106 Chr.14: 73717282-73717282 on Build GRCh38
rs917462251 Chr.14: 73717299-73717299 on Build GRCh38
rs964647951 Chr.14: 73717502-73717502 on Build GRCh38
rs934791239 Chr.14: 73717279-73717279 on Build GRCh38
rs371049661 Chr.14: 73717441-73717441 on Build GRCh38
rs978819082 Chr.14: 73717513-73717513 on Build GRCh38
rs996145048 Chr.14: 73717510-73717510 on Build GRCh38
rs79317866 Chr.14: 73717530-73717530 on Build GRCh38
rs557288865 Chr.14: 73717563-73717563 on Build GRCh38
rs1008796568 Chr.14: 73717493-73717493 on Build GRCh38
rs944918100 Chr.14: 73717462-73717462 on Build GRCh38

View Full Product Details