Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 77675760-77676305 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALKBH1
Entrez Gene ID
8846
Gene Name
alkB homolog 1, histone H2A dioxygenase
Gene Aliases
ABH, ABH1, ALKBH, alkB, hABH
UniGene
Hs.94542
Gene Chromosome Location
Chr.14: 77672404-77708038 on Build GRCh38
Transcripts
NM_006020.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
958209 c.612C>T p.A204A Chr.14: 77675784-77675784 on Build GRCh38
3498274 c.576C>T p.F192F Chr.14: 77675820-77675820 on Build GRCh38
5633297 c.624A>C p.G208G Chr.14: 77675772-77675772 on Build GRCh38
3370076 c.587T>C p.L196P Chr.14: 77675809-77675809 on Build GRCh38
2139533 c.583G>C p.D195H Chr.14: 77675813-77675813 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs991130914 Chr.14: 77676088-77676088 on Build GRCh38
rs1053808781 Chr.14: 77675910-77675910 on Build GRCh38
rs780975155 Chr.14: 77675784-77675784 on Build GRCh38
rs977397715 Chr.14: 77676002-77676003 on Build GRCh38
rs763494299 Chr.14: 77675870-77675870 on Build GRCh38
rs67783226 Chr.14: 77676163-77676163 on Build GRCh38
rs908867037 Chr.14: 77676045-77676045 on Build GRCh38
rs898535205 Chr.14: 77675897-77675897 on Build GRCh38
rs747276077 Chr.14: 77675834-77675834 on Build GRCh38
rs398025818 Chr.14: 77676176-77676176 on Build GRCh38
rs777853110 Chr.14: 77675944-77675947 on Build GRCh38
rs559900463 Chr.14: 77675960-77675960 on Build GRCh38
rs1006961738 Chr.14: 77675947-77675947 on Build GRCh38
rs777688584 Chr.14: 77675825-77675825 on Build GRCh38
rs987278471 Chr.14: 77675973-77675973 on Build GRCh38
rs559304077 Chr.14: 77676279-77676279 on Build GRCh38
rs550673979 Chr.14: 77676064-77676064 on Build GRCh38
rs1008057751 Chr.14: 77676036-77676047 on Build GRCh38
rs889839260 Chr.14: 77675943-77675943 on Build GRCh38
rs553562105 Chr.14: 77676149-77676149 on Build GRCh38
rs1055313356 Chr.14: 77675822-77675822 on Build GRCh38
rs745716364 Chr.14: 77675982-77675982 on Build GRCh38
rs753272160 Chr.14: 77675799-77675799 on Build GRCh38
rs967685748 Chr.14: 77676001-77676002 on Build GRCh38
rs923829731 Chr.14: 77676210-77676210 on Build GRCh38
rs991786658 Chr.14: 77675998-77675998 on Build GRCh38
rs540438026 Chr.14: 77675952-77675952 on Build GRCh38
rs1028377092 Chr.14: 77675947-77675947 on Build GRCh38
rs940265703 Chr.14: 77676184-77676184 on Build GRCh38
rs567066988 Chr.14: 77676273-77676273 on Build GRCh38
rs367553871 Chr.14: 77676106-77676106 on Build GRCh38
rs1054141058 Chr.14: 77676253-77676253 on Build GRCh38
rs143778342 Chr.14: 77676112-77676112 on Build GRCh38
rs917599672 Chr.14: 77676134-77676134 on Build GRCh38
rs774690264 Chr.14: 77675901-77675901 on Build GRCh38
rs891197801 Chr.14: 77676248-77676248 on Build GRCh38
rs990580243 Chr.14: 77676111-77676111 on Build GRCh38
rs776421754 Chr.14: 77676078-77676078 on Build GRCh38
rs996853068 Chr.14: 77675926-77675926 on Build GRCh38
rs745937508 Chr.14: 77675844-77675844 on Build GRCh38
rs894238595 Chr.14: 77675794-77675794 on Build GRCh38
rs552047576 Chr.14: 77676023-77676023 on Build GRCh38
rs575303202 Chr.14: 77675784-77675784 on Build GRCh38
rs779602975 Chr.14: 77675789-77675789 on Build GRCh38
rs915640909 Chr.14: 77676102-77676102 on Build GRCh38
rs996820355 Chr.14: 77676242-77676242 on Build GRCh38
rs746327127 Chr.14: 77675998-77675999 on Build GRCh38
rs1019679913 Chr.14: 77675956-77675956 on Build GRCh38
rs776831961 Chr.14: 77675842-77675842 on Build GRCh38
rs755728200 Chr.14: 77675785-77675785 on Build GRCh38
rs1038579059 Chr.14: 77676197-77676197 on Build GRCh38
rs1025812953 Chr.14: 77675979-77675979 on Build GRCh38
rs747949486 Chr.14: 77675885-77675887 on Build GRCh38
rs949163165 Chr.14: 77676128-77676128 on Build GRCh38
rs543822365 Chr.14: 77675806-77675806 on Build GRCh38
rs72685246 Chr.14: 77676278-77676278 on Build GRCh38
rs768356990 Chr.14: 77675968-77675968 on Build GRCh38
rs139884884 Chr.14: 77676062-77676062 on Build GRCh38
rs536335349 Chr.14: 77676122-77676122 on Build GRCh38
rs994304528 Chr.14: 77675975-77675975 on Build GRCh38
rs201032174 Chr.14: 77675881-77675881 on Build GRCh38
rs939912437 Chr.14: 77676148-77676148 on Build GRCh38
rs936614530 Chr.14: 77676247-77676247 on Build GRCh38
rs898757794 Chr.14: 77676230-77676230 on Build GRCh38
rs374529582 Chr.14: 77675999-77675999 on Build GRCh38
rs977920549 Chr.14: 77676142-77676142 on Build GRCh38
rs145967498 Chr.14: 77675811-77675811 on Build GRCh38
rs775883346 Chr.14: 77675862-77675862 on Build GRCh38
rs1008342998 Chr.14: 77676249-77676249 on Build GRCh38
rs532298149 Chr.14: 77676003-77676003 on Build GRCh38
rs769876266 Chr.14: 77675861-77675861 on Build GRCh38
rs771274995 Chr.14: 77675835-77675835 on Build GRCh38
rs571345561 Chr.14: 77676030-77676030 on Build GRCh38

View Full Product Details