Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 87934667-87935172 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALC
Entrez Gene ID
2581
Gene Name
galactosylceramidase
Gene Aliases
-
UniGene
Hs.513439
Gene Chromosome Location
Chr.14: 87933014-87993665 on Build GRCh38
Transcripts
NM_000153.3 NM_001201401.1 NM_001201402.1 XM_011536618.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5971275 c.1975C>T p.R659C Chr.14: 87934737-87934737 on Build GRCh38
5614170 c.1892A>G p.N631S Chr.14: 87934820-87934820 on Build GRCh38
5614169 c.1970A>G p.N657S Chr.14: 87934820-87934820 on Build GRCh38
1371402 c.1950G>A p.L650L Chr.14: 87934840-87934840 on Build GRCh38
5971274 c.2053C>T p.R685C Chr.14: 87934737-87934737 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780593419 Chr.14: 87934856-87934856 on Build GRCh38
rs912303959 Chr.14: 87934987-87934987 on Build GRCh38
rs570254974 Chr.14: 87935035-87935035 on Build GRCh38
rs12888640 Chr.14: 87934730-87934730 on Build GRCh38
rs1047619650 Chr.14: 87935002-87935004 on Build GRCh38
rs748686038 Chr.14: 87934887-87934887 on Build GRCh38
rs143444570 Chr.14: 87934718-87934718 on Build GRCh38
rs762034337 Chr.14: 87934812-87934812 on Build GRCh38
rs746502573 Chr.14: 87934697-87934697 on Build GRCh38
rs530417759 Chr.14: 87934794-87934794 on Build GRCh38
rs434541 Chr.14: 87935099-87935099 on Build GRCh38
rs752296575 Chr.14: 87934710-87934710 on Build GRCh38
rs754392997 Chr.14: 87934927-87934927 on Build GRCh38
rs386779711 Chr.14: 87935098-87935099 on Build GRCh38
rs1037429777 Chr.14: 87934977-87934977 on Build GRCh38
rs781620553 Chr.14: 87934721-87934721 on Build GRCh38
rs756587314 Chr.14: 87934834-87934834 on Build GRCh38
rs376099163 Chr.14: 87934810-87934810 on Build GRCh38
rs924881775 Chr.14: 87934843-87934843 on Build GRCh38
rs768745262 Chr.14: 87934748-87934748 on Build GRCh38
rs79558046 Chr.14: 87935116-87935116 on Build GRCh38
rs773831789 Chr.14: 87934903-87934903 on Build GRCh38
rs756141815 Chr.14: 87934737-87934737 on Build GRCh38
rs183869279 Chr.14: 87934756-87934756 on Build GRCh38
rs770785840 Chr.14: 87934768-87934768 on Build GRCh38
rs763585644 Chr.14: 87934798-87934798 on Build GRCh38
rs959213054 Chr.14: 87935074-87935074 on Build GRCh38
rs903149710 Chr.14: 87934741-87934741 on Build GRCh38
rs535674130 Chr.14: 87934969-87934969 on Build GRCh38
rs370562686 Chr.14: 87935036-87935036 on Build GRCh38
rs1015848350 Chr.14: 87934701-87934701 on Build GRCh38
rs779202612 Chr.14: 87934755-87934755 on Build GRCh38
rs768231872 Chr.14: 87934899-87934899 on Build GRCh38
rs539313761 Chr.14: 87934910-87934910 on Build GRCh38
rs914759887 Chr.14: 87935029-87935029 on Build GRCh38
rs750822991 Chr.14: 87934823-87934823 on Build GRCh38
rs433764 Chr.14: 87935048-87935048 on Build GRCh38
rs749415816 Chr.14: 87934744-87934744 on Build GRCh38
rs372641636 Chr.14: 87934720-87934720 on Build GRCh38
rs116281628 Chr.14: 87934713-87934713 on Build GRCh38
rs1039895290 Chr.14: 87934995-87934995 on Build GRCh38
rs550351293 Chr.14: 87934813-87934813 on Build GRCh38
rs201596265 Chr.14: 87934751-87934751 on Build GRCh38
rs762593303 Chr.14: 87934797-87934797 on Build GRCh38
rs753856679 Chr.14: 87934858-87934858 on Build GRCh38
rs529469325 Chr.14: 87934736-87934736 on Build GRCh38
rs1003472586 Chr.14: 87935047-87935047 on Build GRCh38
rs754507781 Chr.14: 87934750-87934753 on Build GRCh38
rs199786487 Chr.14: 87935098-87935099 on Build GRCh38
rs759515663 Chr.14: 87934787-87934787 on Build GRCh38
rs564789658 Chr.14: 87934691-87934691 on Build GRCh38
rs761093081 Chr.14: 87934925-87934925 on Build GRCh38
rs970572144 Chr.14: 87934733-87934733 on Build GRCh38
rs776674605 Chr.14: 87934776-87934776 on Build GRCh38
rs780754563 Chr.14: 87934781-87934784 on Build GRCh38
rs386779710 Chr.14: 87934869-87934870 on Build GRCh38
rs769851272 Chr.14: 87934878-87934878 on Build GRCh38
rs879863225 Chr.14: 87935098-87935099 on Build GRCh38
rs748177483 Chr.14: 87934989-87934989 on Build GRCh38
rs575846501 Chr.14: 87935089-87935089 on Build GRCh38
rs1039926220 Chr.14: 87934711-87934711 on Build GRCh38
rs770352387 Chr.14: 87934894-87934895 on Build GRCh38
rs947588012 Chr.14: 87935031-87935031 on Build GRCh38
rs780073716 Chr.14: 87934742-87934742 on Build GRCh38
rs147161584 Chr.14: 87934926-87934926 on Build GRCh38
rs188467654 Chr.14: 87934924-87934924 on Build GRCh38
rs421262 Chr.14: 87934869-87934869 on Build GRCh38
rs982394931 Chr.14: 87934735-87934735 on Build GRCh38
rs977689069 Chr.14: 87934819-87934819 on Build GRCh38
rs942453256 Chr.14: 87934992-87934992 on Build GRCh38
rs772467584 Chr.14: 87934870-87934870 on Build GRCh38
rs748543021 Chr.14: 87934879-87934880 on Build GRCh38
rs755100827 Chr.14: 87934865-87934865 on Build GRCh38
rs767206135 Chr.14: 87934912-87934912 on Build GRCh38
rs781113050 Chr.14: 87934874-87934874 on Build GRCh38
rs769902280 Chr.14: 87934789-87934789 on Build GRCh38
rs200607029 Chr.14: 87934749-87934749 on Build GRCh38
rs376302808 Chr.14: 87934885-87934885 on Build GRCh38
rs390474 Chr.14: 87935098-87935098 on Build GRCh38
rs764875556 Chr.14: 87934702-87934702 on Build GRCh38

View Full Product Details