Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 103699011-103699513 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLC1
Entrez Gene ID
3831
Gene Name
kinesin light chain 1
Gene Aliases
KLC, KNS2, KNS2A
UniGene
Hs.20107
Gene Chromosome Location
Chr.14: 103629188-103701551 on Build GRCh38
Transcripts
NM_001130107.1 NM_182923.3 NM_001100118.1 NM_001100119.1 NM_005432.3 XM_005268046.2 XM_011537138.2

Gene Symbol
XRCC3
Entrez Gene ID
7517
Gene Name
X-ray repair cross complementing 3
Gene Aliases
CMM6
UniGene
Hs.592325
Gene Chromosome Location
Chr.14: 103697611-103715486 on Build GRCh38
Transcripts
NM_001130107.1 NM_182923.3 NM_001100118.1 NM_001100119.1 NM_005432.3 XM_005268046.2 XM_011537138.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4409450 c.768C>T p.I256I Chr.14: 103699370-103699370 on Build GRCh38
5747610 c.782_783delAG p.E261fs*29 Chr.14: 103699171-103699172 on Build GRCh38
2025713 c.685G>A p.A229T Chr.14: 103699453-103699453 on Build GRCh38
6275960 c.650C>A p.A217D Chr.14: 103699488-103699488 on Build GRCh38
1748720 c.666A>G p.E222E Chr.14: 103699472-103699472 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs545840795 Chr.14: 103699480-103699480 on Build GRCh38
rs373720646 Chr.14: 103699462-103699462 on Build GRCh38
rs999097958 Chr.14: 103699136-103699136 on Build GRCh38
rs1038174944 Chr.14: 103699298-103699298 on Build GRCh38
rs150986165 Chr.14: 103699310-103699310 on Build GRCh38
rs145063633 Chr.14: 103699167-103699167 on Build GRCh38
rs774160691 Chr.14: 103699128-103699128 on Build GRCh38
rs17101777 Chr.14: 103699074-103699074 on Build GRCh38
rs538640569 Chr.14: 103699123-103699123 on Build GRCh38
rs3212114 Chr.14: 103699259-103699259 on Build GRCh38
rs3212113 Chr.14: 103699303-103699303 on Build GRCh38
rs575587037 Chr.14: 103699407-103699407 on Build GRCh38
rs757909834 Chr.14: 103699183-103699183 on Build GRCh38
rs991885466 Chr.14: 103699250-103699250 on Build GRCh38
rs752714158 Chr.14: 103699387-103699388 on Build GRCh38
rs376400797 Chr.14: 103699210-103699210 on Build GRCh38
rs1028019465 Chr.14: 103699165-103699165 on Build GRCh38
rs777515106 Chr.14: 103699396-103699396 on Build GRCh38
rs748493439 Chr.14: 103699116-103699116 on Build GRCh38
rs745449993 Chr.14: 103699274-103699274 on Build GRCh38
rs1047962571 Chr.14: 103699125-103699125 on Build GRCh38
rs45565335 Chr.14: 103699090-103699090 on Build GRCh38
rs201886982 Chr.14: 103699476-103699476 on Build GRCh38
rs531515916 Chr.14: 103699291-103699291 on Build GRCh38
rs546723375 Chr.14: 103699322-103699322 on Build GRCh38
rs989165390 Chr.14: 103699246-103699246 on Build GRCh38
rs890853307 Chr.14: 103699166-103699166 on Build GRCh38
rs56225499 Chr.14: 103699046-103699046 on Build GRCh38
rs771788141 Chr.14: 103699226-103699226 on Build GRCh38
rs752896646 Chr.14: 103699058-103699058 on Build GRCh38
rs766900735 Chr.14: 103699419-103699419 on Build GRCh38
rs758701199 Chr.14: 103699064-103699064 on Build GRCh38
rs752896177 Chr.14: 103699164-103699164 on Build GRCh38
rs191281642 Chr.14: 103699262-103699262 on Build GRCh38
rs544850281 Chr.14: 103699244-103699244 on Build GRCh38
rs749728676 Chr.14: 103699227-103699227 on Build GRCh38
rs771174982 Chr.14: 103699095-103699095 on Build GRCh38
rs778626661 Chr.14: 103699218-103699218 on Build GRCh38
rs374774780 Chr.14: 103699119-103699119 on Build GRCh38
rs79874791 Chr.14: 103699415-103699415 on Build GRCh38
rs560276298 Chr.14: 103699261-103699261 on Build GRCh38
rs777604903 Chr.14: 103699453-103699453 on Build GRCh38
rs759615397 Chr.14: 103699144-103699144 on Build GRCh38
rs900479421 Chr.14: 103699151-103699151 on Build GRCh38
rs373638417 Chr.14: 103699373-103699373 on Build GRCh38
rs3212115 Chr.14: 103699251-103699251 on Build GRCh38
rs755604906 Chr.14: 103699200-103699200 on Build GRCh38
rs56393055 Chr.14: 103699212-103699212 on Build GRCh38
rs45475405 Chr.14: 103699282-103699282 on Build GRCh38
rs138142727 Chr.14: 103699427-103699427 on Build GRCh38
rs778161228 Chr.14: 103699065-103699065 on Build GRCh38
rs969202747 Chr.14: 103699406-103699406 on Build GRCh38
rs188768970 Chr.14: 103699421-103699421 on Build GRCh38
rs938878023 Chr.14: 103699265-103699265 on Build GRCh38
rs765853123 Chr.14: 103699359-103699359 on Build GRCh38
rs369703243 Chr.14: 103699352-103699352 on Build GRCh38
rs200657624 Chr.14: 103699382-103699382 on Build GRCh38
rs56034293 Chr.14: 103699264-103699264 on Build GRCh38
rs959531828 Chr.14: 103699355-103699355 on Build GRCh38
rs754653129 Chr.14: 103699367-103699367 on Build GRCh38
rs778540307 Chr.14: 103699376-103699376 on Build GRCh38
rs768365128 Chr.14: 103699126-103699126 on Build GRCh38
rs543072564 Chr.14: 103699336-103699337 on Build GRCh38
rs780906908 Chr.14: 103699083-103699083 on Build GRCh38
rs773686312 Chr.14: 103699417-103699417 on Build GRCh38
rs749394060 Chr.14: 103699411-103699411 on Build GRCh38
rs1026835746 Chr.14: 103699242-103699242 on Build GRCh38
rs750906208 Chr.14: 103699191-103699191 on Build GRCh38
rs566710190 Chr.14: 103699403-103699403 on Build GRCh38
rs28903080 Chr.14: 103699143-103699143 on Build GRCh38
rs756935758 Chr.14: 103699455-103699455 on Build GRCh38
rs772408011 Chr.14: 103699479-103699479 on Build GRCh38
rs376831498 Chr.14: 103699460-103699460 on Build GRCh38
rs3212112 Chr.14: 103699345-103699345 on Build GRCh38
rs779590080 Chr.14: 103699121-103699121 on Build GRCh38
rs753371151 Chr.14: 103699360-103699360 on Build GRCh38
rs990300669 Chr.14: 103699442-103699442 on Build GRCh38
rs754529357 Chr.14: 103699425-103699425 on Build GRCh38
rs766482540 Chr.14: 103699045-103699045 on Build GRCh38
rs757719690 Chr.14: 103699435-103699435 on Build GRCh38
rs757817573 Chr.14: 103699069-103699069 on Build GRCh38
rs376047069 Chr.14: 103699260-103699260 on Build GRCh38
rs768565346 Chr.14: 103699413-103699413 on Build GRCh38
rs989356677 Chr.14: 103699337-103699337 on Build GRCh38
rs746816769 Chr.14: 103699454-103699454 on Build GRCh38
rs767691806 Chr.14: 103699372-103699373 on Build GRCh38
rs77381814 Chr.14: 103699410-103699410 on Build GRCh38
rs771442715 Chr.14: 103699231-103699231 on Build GRCh38
rs761784037 Chr.14: 103699138-103699138 on Build GRCh38
rs756610731 Chr.14: 103699350-103699350 on Build GRCh38
rs996215859 Chr.14: 103699152-103699152 on Build GRCh38
rs781206798 Chr.14: 103699458-103699458 on Build GRCh38
rs758025766 Chr.14: 103699450-103699450 on Build GRCh38
rs183521431 Chr.14: 103699318-103699318 on Build GRCh38
rs199549072 Chr.14: 103699043-103699043 on Build GRCh38
rs368501131 Chr.14: 103699067-103699067 on Build GRCh38
rs763255103 Chr.14: 103699055-103699057 on Build GRCh38
rs749414375 Chr.14: 103699127-103699127 on Build GRCh38
rs368831796 Chr.14: 103699472-103699472 on Build GRCh38
rs774598205 Chr.14: 103699321-103699322 on Build GRCh38
rs861539 Chr.14: 103699416-103699416 on Build GRCh38
rs547011404 Chr.14: 103699326-103699326 on Build GRCh38
rs950190832 Chr.14: 103699102-103699102 on Build GRCh38
rs947763538 Chr.14: 103699073-103699073 on Build GRCh38
rs1008315699 Chr.14: 103699311-103699311 on Build GRCh38
rs942196380 Chr.14: 103699283-103699283 on Build GRCh38
rs764416609 Chr.14: 103699171-103699171 on Build GRCh38
rs765373047 Chr.14: 103699049-103699049 on Build GRCh38
rs770889808 Chr.14: 103699408-103699408 on Build GRCh38
rs767947314 Chr.14: 103699342-103699342 on Build GRCh38
rs771142980 Chr.14: 103699492-103699492 on Build GRCh38
rs762286206 Chr.14: 103699330-103699330 on Build GRCh38
rs745370909 Chr.14: 103699106-103699106 on Build GRCh38
rs567960077 Chr.14: 103699314-103699314 on Build GRCh38
rs771984433 Chr.14: 103699140-103699140 on Build GRCh38
rs765302014 Chr.14: 103699153-103699153 on Build GRCh38
rs201759596 Chr.14: 103699329-103699329 on Build GRCh38
rs769807134 Chr.14: 103699316-103699316 on Build GRCh38
rs751959603 Chr.14: 103699180-103699180 on Build GRCh38
rs761813245 Chr.14: 103699451-103699456 on Build GRCh38
rs768891111 Chr.14: 103699474-103699474 on Build GRCh38
rs781520263 Chr.14: 103699114-103699114 on Build GRCh38
rs761101817 Chr.14: 103699489-103699489 on Build GRCh38
rs748931165 Chr.14: 103699211-103699211 on Build GRCh38

View Full Product Details