Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 23686241-23686734 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDN
Entrez Gene ID
4692
Gene Name
necdin, MAGE family member
Gene Aliases
HsT16328, PWCR
UniGene
Hs.50130
Gene Chromosome Location
Chr.15: 23685407-23687303 on Build GRCh38
Transcripts
NM_002487.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1678298 c.659G>A p.R220Q Chr.15: 23686559-23686559 on Build GRCh38
6723511 c.856G>T p.D286Y Chr.15: 23686362-23686362 on Build GRCh38
3500039 c.960G>A p.E320E Chr.15: 23686258-23686258 on Build GRCh38
1707938 c.828G>A p.M276I Chr.15: 23686390-23686390 on Build GRCh38
6025546 c.553A>G p.M185V Chr.15: 23686665-23686665 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780869401 Chr.15: 23686575-23686575 on Build GRCh38
rs868081273 Chr.15: 23686340-23686340 on Build GRCh38
rs761172221 Chr.15: 23686625-23686625 on Build GRCh38
rs377398058 Chr.15: 23686316-23686316 on Build GRCh38
rs867008265 Chr.15: 23686577-23686577 on Build GRCh38
rs978719509 Chr.15: 23686550-23686550 on Build GRCh38
rs148944452 Chr.15: 23686570-23686570 on Build GRCh38
rs769908982 Chr.15: 23686605-23686605 on Build GRCh38
rs372364421 Chr.15: 23686462-23686462 on Build GRCh38
rs749762250 Chr.15: 23686355-23686356 on Build GRCh38
rs371472034 Chr.15: 23686395-23686395 on Build GRCh38
rs376175102 Chr.15: 23686389-23686389 on Build GRCh38
rs765322757 Chr.15: 23686560-23686560 on Build GRCh38
rs532203832 Chr.15: 23686305-23686305 on Build GRCh38
rs143663868 Chr.15: 23686659-23686659 on Build GRCh38
rs866169143 Chr.15: 23686334-23686334 on Build GRCh38
rs762933700 Chr.15: 23686678-23686678 on Build GRCh38
rs748090305 Chr.15: 23686435-23686435 on Build GRCh38
rs772907017 Chr.15: 23686278-23686278 on Build GRCh38
rs755074055 Chr.15: 23686427-23686427 on Build GRCh38
rs748887378 Chr.15: 23686714-23686714 on Build GRCh38
rs770123594 Chr.15: 23686297-23686297 on Build GRCh38
rs371636740 Chr.15: 23686699-23686699 on Build GRCh38
rs766766336 Chr.15: 23686522-23686522 on Build GRCh38
rs114077338 Chr.15: 23686265-23686265 on Build GRCh38
rs761117087 Chr.15: 23686516-23686516 on Build GRCh38
rs770500531 Chr.15: 23686455-23686455 on Build GRCh38
rs778906913 Chr.15: 23686366-23686366 on Build GRCh38
rs368282875 Chr.15: 23686417-23686417 on Build GRCh38
rs771425671 Chr.15: 23686402-23686403 on Build GRCh38
rs777026374 Chr.15: 23686648-23686648 on Build GRCh38
rs753543868 Chr.15: 23686532-23686532 on Build GRCh38
rs745500582 Chr.15: 23686581-23686581 on Build GRCh38
rs767688029 Chr.15: 23686343-23686343 on Build GRCh38
rs750649069 Chr.15: 23686351-23686351 on Build GRCh38
rs145763085 Chr.15: 23686569-23686569 on Build GRCh38
rs777277983 Chr.15: 23686298-23686315 on Build GRCh38
rs751473607 Chr.15: 23686684-23686684 on Build GRCh38
rs762857502 Chr.15: 23686519-23686519 on Build GRCh38
rs367977212 Chr.15: 23686595-23686595 on Build GRCh38
rs757030070 Chr.15: 23686574-23686574 on Build GRCh38
rs565889752 Chr.15: 23686380-23686380 on Build GRCh38
rs776066263 Chr.15: 23686388-23686388 on Build GRCh38
rs750065030 Chr.15: 23686695-23686695 on Build GRCh38
rs762132398 Chr.15: 23686393-23686393 on Build GRCh38
rs200117077 Chr.15: 23686301-23686301 on Build GRCh38
rs771393294 Chr.15: 23686629-23686629 on Build GRCh38
rs766110712 Chr.15: 23686639-23686639 on Build GRCh38
rs149718513 Chr.15: 23686534-23686534 on Build GRCh38
rs750704224 Chr.15: 23686415-23686415 on Build GRCh38
rs748616399 Chr.15: 23686606-23686606 on Build GRCh38
rs774758735 Chr.15: 23686495-23686495 on Build GRCh38
rs769323151 Chr.15: 23686486-23686486 on Build GRCh38
rs142210120 Chr.15: 23686450-23686450 on Build GRCh38
rs771690730 Chr.15: 23686272-23686272 on Build GRCh38
rs371136148 Chr.15: 23686353-23686353 on Build GRCh38
rs752759758 Chr.15: 23686677-23686677 on Build GRCh38
rs549788947 Chr.15: 23686285-23686285 on Build GRCh38
rs772783094 Chr.15: 23686383-23686383 on Build GRCh38
rs780814235 Chr.15: 23686461-23686461 on Build GRCh38
rs774718730 Chr.15: 23686621-23686628 on Build GRCh38
rs765847311 Chr.15: 23686290-23686290 on Build GRCh38
rs901682735 Chr.15: 23686582-23686582 on Build GRCh38
rs757634514 Chr.15: 23686317-23686317 on Build GRCh38
rs764073249 Chr.15: 23686679-23686679 on Build GRCh38
rs779567210 Chr.15: 23686601-23686601 on Build GRCh38
rs770453718 Chr.15: 23686385-23686385 on Build GRCh38
rs773843519 Chr.15: 23686615-23686615 on Build GRCh38
rs778767538 Chr.15: 23686429-23686429 on Build GRCh38
rs924606829 Chr.15: 23686563-23686563 on Build GRCh38
rs2192206 Chr.15: 23686360-23686360 on Build GRCh38
rs757081401 Chr.15: 23686687-23686687 on Build GRCh38
rs758161696 Chr.15: 23686438-23686438 on Build GRCh38
rs112774241 Chr.15: 23686668-23686668 on Build GRCh38
rs994158712 Chr.15: 23686688-23686688 on Build GRCh38
rs267604134 Chr.15: 23686326-23686326 on Build GRCh38
rs780922737 Chr.15: 23686690-23686690 on Build GRCh38
rs754040091 Chr.15: 23686424-23686424 on Build GRCh38
rs756301395 Chr.15: 23686352-23686352 on Build GRCh38
rs145312445 Chr.15: 23686269-23686269 on Build GRCh38
rs202231164 Chr.15: 23686528-23686528 on Build GRCh38
rs769217515 Chr.15: 23686590-23686590 on Build GRCh38
rs776854811 Chr.15: 23686275-23686275 on Build GRCh38
rs749259531 Chr.15: 23686357-23686357 on Build GRCh38
rs764572296 Chr.15: 23686392-23686392 on Build GRCh38
rs746627311 Chr.15: 23686384-23686384 on Build GRCh38
rs766572018 Chr.15: 23686422-23686422 on Build GRCh38
rs267604135 Chr.15: 23686333-23686333 on Build GRCh38
rs543532099 Chr.15: 23686494-23686494 on Build GRCh38
rs556576161 Chr.15: 23686680-23686680 on Build GRCh38
rs752601934 Chr.15: 23686564-23686564 on Build GRCh38
rs764528624 Chr.15: 23686312-23686312 on Build GRCh38
rs772344835 Chr.15: 23686506-23686506 on Build GRCh38
rs1005917965 Chr.15: 23686651-23686651 on Build GRCh38
rs773653514 Chr.15: 23686515-23686515 on Build GRCh38
rs765248776 Chr.15: 23686667-23686667 on Build GRCh38
rs772773476 Chr.15: 23686608-23686608 on Build GRCh38
rs549529148 Chr.15: 23686500-23686500 on Build GRCh38

View Full Product Details